Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3496636-3496861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOHH
Entrez Gene ID
83475
Gene Name
deoxyhypusine hydroxylase/monooxygenase
Gene Aliases
HLRC1, hDOHH
UniGene
Hs.515064
Gene Chromosome Location
Chr.19: 3490821-3500940 on Build GRCh38
Transcripts
NM_001145165.1 NM_031304.4 XM_011528341.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3196823 c.38C>T p.T13M Chr.19: 3496777-3496777 on Build GRCh38
4903434 c.102C>T p.L34L Chr.19: 3496713-3496713 on Build GRCh38
4972612 c.71G>T p.R24L Chr.19: 3496744-3496744 on Build GRCh38
3422665 c.115G>A p.A39T Chr.19: 3496700-3496700 on Build GRCh38
4939794 c.22G>A p.D8N Chr.19: 3496793-3496793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751828553 Chr.19: 3496709-3496709 on Build GRCh38
rs775063975 Chr.19: 3496819-3496819 on Build GRCh38
rs750062548 Chr.19: 3496833-3496833 on Build GRCh38
rs778229924 Chr.19: 3496721-3496721 on Build GRCh38
rs756904433 Chr.19: 3496762-3496762 on Build GRCh38
rs753559247 Chr.19: 3496761-3496761 on Build GRCh38
rs919015897 Chr.19: 3496729-3496729 on Build GRCh38
rs754040779 Chr.19: 3496692-3496692 on Build GRCh38
rs767989996 Chr.19: 3496757-3496757 on Build GRCh38
rs765369562 Chr.19: 3496739-3496739 on Build GRCh38
rs766450742 Chr.19: 3496816-3496816 on Build GRCh38
rs377527596 Chr.19: 3496787-3496787 on Build GRCh38
rs778426380 Chr.19: 3496767-3496767 on Build GRCh38
rs769502127 Chr.19: 3496659-3496659 on Build GRCh38
rs774386764 Chr.19: 3496663-3496663 on Build GRCh38
rs371544653 Chr.19: 3496674-3496674 on Build GRCh38
rs751506501 Chr.19: 3496836-3496836 on Build GRCh38
rs60925578 Chr.19: 3496835-3496835 on Build GRCh38
rs745823611 Chr.19: 3496725-3496725 on Build GRCh38
rs372170801 Chr.19: 3496732-3496732 on Build GRCh38
rs773095684 Chr.19: 3496742-3496742 on Build GRCh38
rs755687932 Chr.19: 3496736-3496736 on Build GRCh38
rs141419837 Chr.19: 3496710-3496710 on Build GRCh38
rs1015003548 Chr.19: 3496829-3496829 on Build GRCh38
rs148032865 Chr.19: 3496777-3496777 on Build GRCh38
rs374807566 Chr.19: 3496661-3496661 on Build GRCh38
rs766767851 Chr.19: 3496706-3496706 on Build GRCh38
rs764253867 Chr.19: 3496682-3496682 on Build GRCh38
rs899812826 Chr.19: 3496815-3496815 on Build GRCh38
rs766745007 Chr.19: 3496745-3496745 on Build GRCh38
rs757916302 Chr.19: 3496834-3496834 on Build GRCh38
rs765301122 Chr.19: 3496700-3496700 on Build GRCh38
rs775380231 Chr.19: 3496728-3496728 on Build GRCh38
rs367741024 Chr.19: 3496667-3496667 on Build GRCh38
rs758770567 Chr.19: 3496702-3496702 on Build GRCh38
rs377309810 Chr.19: 3496713-3496713 on Build GRCh38
rs763545312 Chr.19: 3496743-3496743 on Build GRCh38
rs779492500 Chr.19: 3496788-3496788 on Build GRCh38
rs761925007 Chr.19: 3496699-3496699 on Build GRCh38
rs376432012 Chr.19: 3496662-3496662 on Build GRCh38
rs750992757 Chr.19: 3496701-3496701 on Build GRCh38
rs753545824 Chr.19: 3496830-3496830 on Build GRCh38
rs778518630 Chr.19: 3496772-3496772 on Build GRCh38
rs373818669 Chr.19: 3496806-3496806 on Build GRCh38
rs534676072 Chr.19: 3496752-3496752 on Build GRCh38
rs761551443 Chr.19: 3496826-3496826 on Build GRCh38
rs753073269 Chr.19: 3496825-3496825 on Build GRCh38
rs762989647 Chr.19: 3496811-3496811 on Build GRCh38
rs375607519 Chr.19: 3496830-3496830 on Build GRCh38
rs368504297 Chr.19: 3496726-3496726 on Build GRCh38
rs767998437 Chr.19: 3496822-3496822 on Build GRCh38
rs777455189 Chr.19: 3496660-3496660 on Build GRCh38
rs375206434 Chr.19: 3496793-3496793 on Build GRCh38
rs372090911 Chr.19: 3496776-3496776 on Build GRCh38
rs765992958 Chr.19: 3496810-3496810 on Build GRCh38
rs868596860 Chr.19: 3496708-3496708 on Build GRCh38
rs140835573 Chr.19: 3496785-3496785 on Build GRCh38
rs200328323 Chr.19: 3496807-3496807 on Build GRCh38
rs376491163 Chr.19: 3496734-3496734 on Build GRCh38
rs760106934 Chr.19: 3496820-3496820 on Build GRCh38
rs754127896 Chr.19: 3496653-3496653 on Build GRCh38
rs748917492 Chr.19: 3496711-3496711 on Build GRCh38
rs113976052 Chr.19: 3496773-3496773 on Build GRCh38
rs755433371 Chr.19: 3496754-3496754 on Build GRCh38
rs771915905 Chr.19: 3496673-3496673 on Build GRCh38
rs762004928 Chr.19: 3496738-3496738 on Build GRCh38
rs147778126 Chr.19: 3496668-3496668 on Build GRCh38

View Full Product Details