Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33405430-33405667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIFC1
Entrez Gene ID
3833
Gene Name
kinesin family member C1
Gene Aliases
HSET, KNSL2
UniGene
Hs.436912
Gene Chromosome Location
Chr.6: 33391524-33410226 on Build GRCh38
Transcripts
NM_002263.3 XM_011514585.1 XM_011514587.2 XM_017010836.1 XM_017010837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7170871 c.1344G>C p.L448L Chr.6: 33405439-33405439 on Build GRCh38
4161069 c.1419G>T p.L473L Chr.6: 33405514-33405514 on Build GRCh38
1078030 c.1408C>T p.R470W Chr.6: 33405503-33405503 on Build GRCh38
6852239 c.1346G>T p.S449I Chr.6: 33405441-33405441 on Build GRCh38
742331 c.1523C>T p.S508F Chr.6: 33405618-33405618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770142434 Chr.6: 33405533-33405533 on Build GRCh38
rs756209540 Chr.6: 33405597-33405597 on Build GRCh38
rs369930694 Chr.6: 33405545-33405545 on Build GRCh38
rs780188691 Chr.6: 33405600-33405600 on Build GRCh38
rs764537054 Chr.6: 33405479-33405479 on Build GRCh38
rs771178651 Chr.6: 33405526-33405526 on Build GRCh38
rs754762568 Chr.6: 33405514-33405514 on Build GRCh38
rs752546936 Chr.6: 33405461-33405461 on Build GRCh38
rs766981533 Chr.6: 33405500-33405500 on Build GRCh38
rs199869318 Chr.6: 33405528-33405528 on Build GRCh38
rs145495302 Chr.6: 33405527-33405527 on Build GRCh38
rs777044732 Chr.6: 33405456-33405456 on Build GRCh38
rs140038474 Chr.6: 33405467-33405467 on Build GRCh38
rs555069021 Chr.6: 33405588-33405588 on Build GRCh38
rs770998169 Chr.6: 33405449-33405449 on Build GRCh38
rs774762509 Chr.6: 33405616-33405616 on Build GRCh38
rs775737916 Chr.6: 33405537-33405537 on Build GRCh38
rs776190697 Chr.6: 33405648-33405648 on Build GRCh38
rs767079494 Chr.6: 33405554-33405555 on Build GRCh38
rs981774834 Chr.6: 33405603-33405603 on Build GRCh38
rs148425649 Chr.6: 33405561-33405561 on Build GRCh38
rs750141338 Chr.6: 33405503-33405503 on Build GRCh38
rs200810007 Chr.6: 33405524-33405524 on Build GRCh38
rs144332063 Chr.6: 33405478-33405478 on Build GRCh38
rs778908482 Chr.6: 33405486-33405486 on Build GRCh38
rs201620772 Chr.6: 33405511-33405511 on Build GRCh38
rs755752726 Chr.6: 33405508-33405508 on Build GRCh38
rs779625685 Chr.6: 33405509-33405509 on Build GRCh38
rs749494078 Chr.6: 33405610-33405610 on Build GRCh38
rs1054446623 Chr.6: 33405577-33405577 on Build GRCh38
rs765052988 Chr.6: 33405576-33405576 on Build GRCh38
rs749636190 Chr.6: 33405542-33405542 on Build GRCh38
rs774758891 Chr.6: 33405547-33405547 on Build GRCh38
rs7770412 Chr.6: 33405564-33405564 on Build GRCh38
rs745973522 Chr.6: 33405596-33405596 on Build GRCh38
rs993223554 Chr.6: 33405459-33405459 on Build GRCh38
rs760201775 Chr.6: 33405642-33405642 on Build GRCh38
rs773669400 Chr.6: 33405636-33405636 on Build GRCh38
rs368268322 Chr.6: 33405460-33405460 on Build GRCh38
rs772574358 Chr.6: 33405625-33405625 on Build GRCh38
rs754781888 Chr.6: 33405574-33405574 on Build GRCh38
rs767073513 Chr.6: 33405560-33405560 on Build GRCh38
rs372177506 Chr.6: 33405480-33405480 on Build GRCh38
rs748456384 Chr.6: 33405618-33405618 on Build GRCh38
rs776006873 Chr.6: 33405469-33405469 on Build GRCh38
rs9278048 Chr.6: 33405647-33405647 on Build GRCh38
rs536745198 Chr.6: 33405586-33405586 on Build GRCh38
rs761214732 Chr.6: 33405537-33405537 on Build GRCh38
rs368995550 Chr.6: 33405558-33405558 on Build GRCh38
rs753485302 Chr.6: 33405566-33405566 on Build GRCh38
rs768967939 Chr.6: 33405611-33405611 on Build GRCh38
rs778742940 Chr.6: 33405515-33405515 on Build GRCh38
rs756902516 Chr.6: 33405615-33405615 on Build GRCh38
rs576528702 Chr.6: 33405592-33405592 on Build GRCh38
rs761278092 Chr.6: 33405489-33405489 on Build GRCh38

View Full Product Details