Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108209053-108209280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG4
Entrez Gene ID
3981
Gene Name
DNA ligase 4
Gene Aliases
LIG4S
UniGene
Hs.166091
Gene Chromosome Location
Chr.13: 108207442-108218368 on Build GRCh38
Transcripts
NM_001098268.1 NM_002312.3 NM_206937.1 XM_005254056.1 XM_005254057.4 XM_005254058.3 XM_006719951.3 XM_006719952.1 XM_011521091.2 XM_011521092.2 XM_017020563.1 XM_017020564.1 XM_017020565.1 XM_017020566.1 XM_017020567.1 XM_017020568.1 XM_017020569.1 XM_017020570.1 XM_017020571.1 XM_017020572.1 XM_017020573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
696075 c.2059G>T p.G687C Chr.13: 108209210-108209210 on Build GRCh38
2262534 c.2155G>A p.D719N Chr.13: 108209114-108209114 on Build GRCh38
1365367 c.2042G>T p.R681I Chr.13: 108209227-108209227 on Build GRCh38
5929484 c.2201G>A p.S734N Chr.13: 108209068-108209068 on Build GRCh38
1128425 c.2166G>A p.K722K Chr.13: 108209103-108209103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746539398 Chr.13: 108209203-108209203 on Build GRCh38
rs902025450 Chr.13: 108209197-108209197 on Build GRCh38
rs769579059 Chr.13: 108209227-108209227 on Build GRCh38
rs996239540 Chr.13: 108209202-108209202 on Build GRCh38
rs371171917 Chr.13: 108209182-108209182 on Build GRCh38
rs866407813 Chr.13: 108209126-108209126 on Build GRCh38
rs772911320 Chr.13: 108209228-108209228 on Build GRCh38
rs370991186 Chr.13: 108209150-108209150 on Build GRCh38
rs957569908 Chr.13: 108209234-108209234 on Build GRCh38
rs184527984 Chr.13: 108209178-108209178 on Build GRCh38
rs778050176 Chr.13: 108209159-108209159 on Build GRCh38
rs937572927 Chr.13: 108209089-108209089 on Build GRCh38
rs754546751 Chr.13: 108209118-108209121 on Build GRCh38
rs780866587 Chr.13: 108209146-108209147 on Build GRCh38
rs766525115 Chr.13: 108209074-108209074 on Build GRCh38
rs746688093 Chr.13: 108209115-108209133 on Build GRCh38
rs377032117 Chr.13: 108209123-108209123 on Build GRCh38
rs375816915 Chr.13: 108209225-108209225 on Build GRCh38
rs373938258 Chr.13: 108209175-108209175 on Build GRCh38
rs142144659 Chr.13: 108209253-108209253 on Build GRCh38
rs866357875 Chr.13: 108209191-108209191 on Build GRCh38
rs372457480 Chr.13: 108209108-108209108 on Build GRCh38
rs780787673 Chr.13: 108209224-108209224 on Build GRCh38
rs751692239 Chr.13: 108209087-108209087 on Build GRCh38
rs768218603 Chr.13: 108209219-108209219 on Build GRCh38
rs996038028 Chr.13: 108209230-108209230 on Build GRCh38
rs779365107 Chr.13: 108209085-108209085 on Build GRCh38
rs1019110835 Chr.13: 108209173-108209173 on Build GRCh38

View Full Product Details