Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69237664-69237917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD2
Entrez Gene ID
55218
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
C14orf114, EXDL2
UniGene
Hs.533878
Gene Chromosome Location
Chr.14: 69191477-69244020 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_001193362.1 NM_001193363.1 NM_018199.3 NR_034164.1 NR_034165.1 XM_005267817.3 XM_011536908.2 XM_017021421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3497349 c.1022C>T p.S341F Chr.14: 69237679-69237679 on Build GRCh38
3497347 c.1007C>T p.S336F Chr.14: 69237664-69237664 on Build GRCh38
2135498 c.1255G>T p.A419S Chr.14: 69237912-69237912 on Build GRCh38
5437441 c.1092G>A p.E364E Chr.14: 69237749-69237749 on Build GRCh38
4681967 c.1560G>T p.K520N Chr.14: 69237842-69237842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146099727 Chr.14: 69237804-69237804 on Build GRCh38
rs764363640 Chr.14: 69237686-69237686 on Build GRCh38
rs745891692 Chr.14: 69237756-69237756 on Build GRCh38
rs772556810 Chr.14: 69237864-69237864 on Build GRCh38
rs774538134 Chr.14: 69237777-69237777 on Build GRCh38
rs757107117 Chr.14: 69237749-69237749 on Build GRCh38
rs940641073 Chr.14: 69237731-69237731 on Build GRCh38
rs749441521 Chr.14: 69237761-69237761 on Build GRCh38
rs200000291 Chr.14: 69237754-69237754 on Build GRCh38
rs567418308 Chr.14: 69237815-69237815 on Build GRCh38
rs36018174 Chr.14: 69237687-69237687 on Build GRCh38
rs200113172 Chr.14: 69237688-69237688 on Build GRCh38
rs764806085 Chr.14: 69237789-69237789 on Build GRCh38
rs753330721 Chr.14: 69237699-69237699 on Build GRCh38
rs768692297 Chr.14: 69237837-69237837 on Build GRCh38
rs780175594 Chr.14: 69237760-69237760 on Build GRCh38
rs138804268 Chr.14: 69237833-69237833 on Build GRCh38
rs771585469 Chr.14: 69237880-69237880 on Build GRCh38
rs932776109 Chr.14: 69237730-69237730 on Build GRCh38
rs376323223 Chr.14: 69237887-69237887 on Build GRCh38
rs762241953 Chr.14: 69237778-69237778 on Build GRCh38
rs573844426 Chr.14: 69237780-69237780 on Build GRCh38
rs768943200 Chr.14: 69237773-69237773 on Build GRCh38
rs752389023 Chr.14: 69237725-69237725 on Build GRCh38
rs769852607 Chr.14: 69237757-69237757 on Build GRCh38
rs368454564 Chr.14: 69237817-69237817 on Build GRCh38
rs764893919 Chr.14: 69237719-69237719 on Build GRCh38
rs144689124 Chr.14: 69237869-69237869 on Build GRCh38
rs371199925 Chr.14: 69237737-69237737 on Build GRCh38
rs150337966 Chr.14: 69237738-69237738 on Build GRCh38
rs142848069 Chr.14: 69237792-69237792 on Build GRCh38
rs912313083 Chr.14: 69237740-69237740 on Build GRCh38
rs763294878 Chr.14: 69237685-69237685 on Build GRCh38
rs779208543 Chr.14: 69237838-69237838 on Build GRCh38
rs8007859 Chr.14: 69237836-69237836 on Build GRCh38
rs890229608 Chr.14: 69237771-69237771 on Build GRCh38
rs534330100 Chr.14: 69237781-69237781 on Build GRCh38
rs768098924 Chr.14: 69237697-69237697 on Build GRCh38
rs372787434 Chr.14: 69237818-69237818 on Build GRCh38
rs751210628 Chr.14: 69237896-69237896 on Build GRCh38
rs773747812 Chr.14: 69237866-69237866 on Build GRCh38
rs45557240 Chr.14: 69237717-69237717 on Build GRCh38
rs775702891 Chr.14: 69237684-69237684 on Build GRCh38
rs146185094 Chr.14: 69237859-69237859 on Build GRCh38
rs761996376 Chr.14: 69237794-69237795 on Build GRCh38
rs138036015 Chr.14: 69237790-69237790 on Build GRCh38
rs763600120 Chr.14: 69237891-69237891 on Build GRCh38
rs765495890 Chr.14: 69237864-69237865 on Build GRCh38
rs763841058 Chr.14: 69237793-69237793 on Build GRCh38
rs761327770 Chr.14: 69237786-69237786 on Build GRCh38
rs780087461 Chr.14: 69237835-69237835 on Build GRCh38

View Full Product Details