Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76498181-76498406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRB
Entrez Gene ID
2103
Gene Name
estrogen related receptor beta
Gene Aliases
DFNB35, ERR2, ERRb, ESRL2, NR3B2
UniGene
Hs.435845
Gene Chromosome Location
Chr.14: 76371347-76501839 on Build GRCh38
Transcripts
NM_004452.3 XM_011536547.2 XM_011536550.2 XM_011536553.2 XM_011536554.2 XM_017021085.1 XM_017021086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
699333 c.1157C>T p.P386L Chr.14: 76498313-76498313 on Build GRCh38
4429980 c.1179G>A p.L393L Chr.14: 76498335-76498335 on Build GRCh38
3956229 c.1081G>T p.E361* Chr.14: 76498237-76498237 on Build GRCh38
5563657 c.1141C>G p.Q381E Chr.14: 76498297-76498297 on Build GRCh38
6857112 c.1145G>A p.R382H Chr.14: 76498301-76498301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367778284 Chr.14: 76498323-76498323 on Build GRCh38
rs149094398 Chr.14: 76498362-76498362 on Build GRCh38
rs557399970 Chr.14: 76498301-76498301 on Build GRCh38
rs372520422 Chr.14: 76498256-76498256 on Build GRCh38
rs727504577 Chr.14: 76498361-76498365 on Build GRCh38
rs878956760 Chr.14: 76498369-76498369 on Build GRCh38
rs544297700 Chr.14: 76498350-76498350 on Build GRCh38
rs948871827 Chr.14: 76498354-76498354 on Build GRCh38
rs976657167 Chr.14: 76498287-76498287 on Build GRCh38
rs761581388 Chr.14: 76498247-76498247 on Build GRCh38
rs954746224 Chr.14: 76498207-76498207 on Build GRCh38
rs746427886 Chr.14: 76498347-76498347 on Build GRCh38
rs727503041 Chr.14: 76498317-76498317 on Build GRCh38
rs765852928 Chr.14: 76498375-76498375 on Build GRCh38
rs876657807 Chr.14: 76498352-76498352 on Build GRCh38
rs773374701 Chr.14: 76498220-76498220 on Build GRCh38
rs772231682 Chr.14: 76498206-76498206 on Build GRCh38
rs35774316 Chr.14: 76498312-76498312 on Build GRCh38
rs764505962 Chr.14: 76498356-76498356 on Build GRCh38
rs777803891 Chr.14: 76498320-76498320 on Build GRCh38
rs766117009 Chr.14: 76498260-76498261 on Build GRCh38
rs755637136 Chr.14: 76498279-76498279 on Build GRCh38
rs563575581 Chr.14: 76498365-76498365 on Build GRCh38
rs61742642 Chr.14: 76498312-76498312 on Build GRCh38
rs201714970 Chr.14: 76498322-76498322 on Build GRCh38
rs373131497 Chr.14: 76498300-76498300 on Build GRCh38
rs553650212 Chr.14: 76498240-76498240 on Build GRCh38
rs771486766 Chr.14: 76498226-76498226 on Build GRCh38
rs370193367 Chr.14: 76498269-76498269 on Build GRCh38
rs535904835 Chr.14: 76498258-76498258 on Build GRCh38
rs760608250 Chr.14: 76498222-76498222 on Build GRCh38
rs770377549 Chr.14: 76498348-76498348 on Build GRCh38
rs747244800 Chr.14: 76498321-76498321 on Build GRCh38
rs945120466 Chr.14: 76498305-76498305 on Build GRCh38
rs777066644 Chr.14: 76498227-76498227 on Build GRCh38
rs762761026 Chr.14: 76498248-76498248 on Build GRCh38
rs778699630 Chr.14: 76498308-76498308 on Build GRCh38
rs921080294 Chr.14: 76498293-76498293 on Build GRCh38
rs760083999 Chr.14: 76498228-76498228 on Build GRCh38
rs983156725 Chr.14: 76498310-76498310 on Build GRCh38
rs747893418 Chr.14: 76498270-76498270 on Build GRCh38
rs765737772 Chr.14: 76498231-76498231 on Build GRCh38
rs372544730 Chr.14: 76498291-76498291 on Build GRCh38
rs748786865 Chr.14: 76498374-76498374 on Build GRCh38

View Full Product Details