Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77336083-77336356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAK1
Entrez Gene ID
5058
Gene Name
p21 (RAC1) activated kinase 1
Gene Aliases
PAKalpha
UniGene
Hs.435714
Gene Chromosome Location
Chr.11: 77322015-77474063 on Build GRCh38
Transcripts
NM_001128620.1 NM_002576.4 XM_011545080.2 XM_011545081.2 XM_011545082.2 XM_011545083.2 XM_011545084.2 XM_017017846.1 XM_017017847.1 XM_017017848.1 XM_017017849.1 XM_017017850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7123915 c.1249G>C p.E417Q Chr.11: 77336250-77336250 on Build GRCh38
7224106 c.1290G>A p.W430* Chr.11: 77336209-77336209 on Build GRCh38
6907190 c.1409T>A p.L470Q Chr.11: 77336090-77336090 on Build GRCh38
6907189 c.1409T>A p.L470Q Chr.11: 77336090-77336090 on Build GRCh38
6944103 c.1387_1388CC>TT p.P463L Chr.11: 77336111-77336112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189183289 Chr.11: 77336313-77336313 on Build GRCh38
rs747098916 Chr.11: 77336187-77336187 on Build GRCh38
rs772588967 Chr.11: 77336296-77336296 on Build GRCh38
rs764494214 Chr.11: 77336303-77336303 on Build GRCh38
rs537450989 Chr.11: 77336322-77336322 on Build GRCh38
rs769876664 Chr.11: 77336218-77336218 on Build GRCh38
rs747237588 Chr.11: 77336328-77336328 on Build GRCh38
rs754801007 Chr.11: 77336290-77336290 on Build GRCh38
rs912418858 Chr.11: 77336257-77336257 on Build GRCh38
rs752969852 Chr.11: 77336167-77336167 on Build GRCh38
rs775404881 Chr.11: 77336245-77336245 on Build GRCh38
rs757273330 Chr.11: 77336199-77336199 on Build GRCh38
rs778201116 Chr.11: 77336181-77336181 on Build GRCh38
rs142211771 Chr.11: 77336215-77336215 on Build GRCh38
rs376509369 Chr.11: 77336269-77336269 on Build GRCh38
rs760412763 Chr.11: 77336297-77336297 on Build GRCh38
rs781239784 Chr.11: 77336202-77336202 on Build GRCh38
rs768747783 Chr.11: 77336287-77336287 on Build GRCh38
rs750033937 Chr.11: 77336118-77336118 on Build GRCh38
rs1011158625 Chr.11: 77336143-77336143 on Build GRCh38
rs765695347 Chr.11: 77336161-77336161 on Build GRCh38
rs761420351 Chr.11: 77336288-77336288 on Build GRCh38
rs147615438 Chr.11: 77336176-77336176 on Build GRCh38
rs780365101 Chr.11: 77336301-77336304 on Build GRCh38
rs55939840 Chr.11: 77336191-77336191 on Build GRCh38
rs977742227 Chr.11: 77336254-77336254 on Build GRCh38
rs112479067 Chr.11: 77336289-77336289 on Build GRCh38
rs972319037 Chr.11: 77336238-77336238 on Build GRCh38
rs753125629 Chr.11: 77336326-77336326 on Build GRCh38
rs267603202 Chr.11: 77336134-77336134 on Build GRCh38

View Full Product Details