Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137232518-137232789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC34A3
Entrez Gene ID
142680
Gene Name
solute carrier family 34 member 3
Gene Aliases
HHRH, NPTIIc
UniGene
Hs.432442
Gene Chromosome Location
Chr.9: 137230757-137236554 on Build GRCh38
Transcripts
NM_001177316.1 NM_001177317.1 NM_080877.2 XM_011518257.2 XM_011518258.1 XM_011518259.1 XM_011518260.1 XM_011518261.2 XM_017014290.1 XM_017014291.1 XM_017014292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624053 c.197T>A p.L66Q Chr.9: 137232596-137232596 on Build GRCh38
1107080 c.252_254delCTT p.F86delF Chr.9: 137232651-137232653 on Build GRCh38
1107081 c.277C>A p.L93I Chr.9: 137232676-137232676 on Build GRCh38
5624051 c.195G>C p.R65S Chr.9: 137232594-137232594 on Build GRCh38
6781565 c.231C>T p.C77C Chr.9: 137232630-137232630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373774399 Chr.9: 137232543-137232543 on Build GRCh38
rs531529369 Chr.9: 137232576-137232576 on Build GRCh38
rs999388892 Chr.9: 137232570-137232570 on Build GRCh38
rs780222614 Chr.9: 137232598-137232598 on Build GRCh38
rs367687903 Chr.9: 137232559-137232559 on Build GRCh38
rs758814501 Chr.9: 137232753-137232753 on Build GRCh38
rs780602942 Chr.9: 137232633-137232633 on Build GRCh38
rs777537074 Chr.9: 137232724-137232724 on Build GRCh38
rs199624248 Chr.9: 137232616-137232616 on Build GRCh38
rs776608897 Chr.9: 137232684-137232684 on Build GRCh38
rs910111613 Chr.9: 137232634-137232634 on Build GRCh38
rs143930538 Chr.9: 137232589-137232589 on Build GRCh38
rs765837541 Chr.9: 137232751-137232751 on Build GRCh38
rs941222172 Chr.9: 137232614-137232615 on Build GRCh38
rs770093905 Chr.9: 137232551-137232551 on Build GRCh38
rs372061694 Chr.9: 137232748-137232748 on Build GRCh38
rs760634105 Chr.9: 137232605-137232605 on Build GRCh38
rs34710159 Chr.9: 137232602-137232602 on Build GRCh38
rs140559114 Chr.9: 137232603-137232603 on Build GRCh38
rs771867611 Chr.9: 137232665-137232665 on Build GRCh38
rs199652389 Chr.9: 137232723-137232723 on Build GRCh38
rs770388562 Chr.9: 137232675-137232675 on Build GRCh38
rs775952487 Chr.9: 137232554-137232554 on Build GRCh38
rs756241784 Chr.9: 137232631-137232631 on Build GRCh38
rs756183651 Chr.9: 137232713-137232713 on Build GRCh38
rs201273897 Chr.9: 137232641-137232641 on Build GRCh38
rs777729650 Chr.9: 137232735-137232735 on Build GRCh38
rs757499807 Chr.9: 137232622-137232622 on Build GRCh38
rs769631018 Chr.9: 137232734-137232734 on Build GRCh38
rs767710880 Chr.9: 137232706-137232706 on Build GRCh38
rs753879514 Chr.9: 137232746-137232746 on Build GRCh38
rs556022435 Chr.9: 137232701-137232701 on Build GRCh38
rs201662577 Chr.9: 137232752-137232752 on Build GRCh38
rs781180635 Chr.9: 137232625-137232625 on Build GRCh38
rs201293634 Chr.9: 137232705-137232705 on Build GRCh38
rs1050390872 Chr.9: 137232762-137232762 on Build GRCh38
rs749421435 Chr.9: 137232576-137232596 on Build GRCh38
rs755237394 Chr.9: 137232609-137232609 on Build GRCh38
rs749796750 Chr.9: 137232639-137232639 on Build GRCh38
rs750340368 Chr.9: 137232580-137232580 on Build GRCh38
rs755447222 Chr.9: 137232718-137232718 on Build GRCh38
rs781272303 Chr.9: 137232594-137232594 on Build GRCh38
rs754789761 Chr.9: 137232583-137232583 on Build GRCh38
rs763818136 Chr.9: 137232561-137232561 on Build GRCh38
rs138218664 Chr.9: 137232731-137232731 on Build GRCh38
rs772857187 Chr.9: 137232604-137232604 on Build GRCh38
rs387907509 Chr.9: 137232644-137232644 on Build GRCh38
rs376991926 Chr.9: 137232640-137232640 on Build GRCh38
rs387907511 Chr.9: 137232617-137232617 on Build GRCh38
rs766590721 Chr.9: 137232582-137232582 on Build GRCh38
rs532224704 Chr.9: 137232579-137232599 on Build GRCh38
rs542402543 Chr.9: 137232700-137232700 on Build GRCh38
rs772397254 Chr.9: 137232651-137232653 on Build GRCh38
rs761399770 Chr.9: 137232575-137232575 on Build GRCh38
rs794729659 Chr.9: 137232627-137232627 on Build GRCh38
rs566604583 Chr.9: 137232717-137232717 on Build GRCh38
rs778304907 Chr.9: 137232757-137232757 on Build GRCh38
rs745749883 Chr.9: 137232733-137232733 on Build GRCh38
rs774035480 Chr.9: 137232601-137232601 on Build GRCh38
rs770425926 Chr.9: 137232730-137232730 on Build GRCh38
rs756864193 Chr.9: 137232763-137232763 on Build GRCh38
rs760842230 Chr.9: 137232739-137232739 on Build GRCh38
rs570528948 Chr.9: 137232610-137232610 on Build GRCh38
rs779089703 Chr.9: 137232578-137232579 on Build GRCh38
rs890109311 Chr.9: 137232699-137232699 on Build GRCh38
rs200633744 Chr.9: 137232716-137232716 on Build GRCh38
rs748680563 Chr.9: 137232611-137232613 on Build GRCh38
rs367866718 Chr.9: 137232742-137232742 on Build GRCh38
rs370472204 Chr.9: 137232588-137232588 on Build GRCh38
rs1030903618 Chr.9: 137232584-137232584 on Build GRCh38
rs145677050 Chr.9: 137232672-137232672 on Build GRCh38
rs373581939 Chr.9: 137232615-137232615 on Build GRCh38
rs368604037 Chr.9: 137232719-137232719 on Build GRCh38
rs548021746 Chr.9: 137232581-137232581 on Build GRCh38
rs758185208 Chr.9: 137232712-137232712 on Build GRCh38
rs759412716 Chr.9: 137232685-137232685 on Build GRCh38
rs746632675 Chr.9: 137232760-137232760 on Build GRCh38
rs200584216 Chr.9: 137232596-137232596 on Build GRCh38
rs749877122 Chr.9: 137232645-137232645 on Build GRCh38
rs746709828 Chr.9: 137232728-137232728 on Build GRCh38
rs201285169 Chr.9: 137232540-137232540 on Build GRCh38
rs576714301 Chr.9: 137232673-137232673 on Build GRCh38
rs748511004 Chr.9: 137232721-137232721 on Build GRCh38
rs773929889 Chr.9: 137232563-137232563 on Build GRCh38
rs34372115 Chr.9: 137232599-137232599 on Build GRCh38
rs375828996 Chr.9: 137232736-137232736 on Build GRCh38
rs879005445 Chr.9: 137232709-137232709 on Build GRCh38
rs750512803 Chr.9: 137232630-137232630 on Build GRCh38
rs532181536 Chr.9: 137232732-137232732 on Build GRCh38
rs746422757 Chr.9: 137232550-137232550 on Build GRCh38

View Full Product Details