Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 122817233-122817488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX2
Entrez Gene ID
6643
Gene Name
sorting nexin 2
Gene Aliases
TRG-9
UniGene
Hs.125352
Gene Chromosome Location
Chr.5: 122774996-122834539 on Build GRCh38
Transcripts
NM_001278199.1 NM_003100.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357839 c.950A>G p.D317G Chr.5: 122817317-122817317 on Build GRCh38
1060104 c.977G>A p.S326N Chr.5: 122817344-122817344 on Build GRCh38
5774041 c.1006+10C>T p.? Chr.5: 122817383-122817383 on Build GRCh38
1060103 c.930G>A p.Q310Q Chr.5: 122817297-122817297 on Build GRCh38
1486239 c.971A>C p.H324P Chr.5: 122817338-122817338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766997236 Chr.5: 122817410-122817410 on Build GRCh38
rs540880558 Chr.5: 122817414-122817414 on Build GRCh38
rs747090144 Chr.5: 122817345-122817345 on Build GRCh38
rs373778409 Chr.5: 122817301-122817301 on Build GRCh38
rs1127846 Chr.5: 122817358-122817358 on Build GRCh38
rs976562266 Chr.5: 122817408-122817408 on Build GRCh38
rs1046742373 Chr.5: 122817441-122817441 on Build GRCh38
rs752351692 Chr.5: 122817393-122817394 on Build GRCh38
rs776897787 Chr.5: 122817352-122817352 on Build GRCh38
rs200130772 Chr.5: 122817366-122817366 on Build GRCh38
rs931199975 Chr.5: 122817428-122817428 on Build GRCh38
rs919853366 Chr.5: 122817423-122817423 on Build GRCh38
rs754096951 Chr.5: 122817285-122817285 on Build GRCh38
rs746141631 Chr.5: 122817365-122817365 on Build GRCh38
rs780905648 Chr.5: 122817263-122817263 on Build GRCh38
rs376654483 Chr.5: 122817399-122817399 on Build GRCh38
rs759029508 Chr.5: 122817267-122817268 on Build GRCh38
rs368367334 Chr.5: 122817418-122817418 on Build GRCh38
rs764888674 Chr.5: 122817392-122817402 on Build GRCh38
rs77998331 Chr.5: 122817390-122817390 on Build GRCh38
rs200594938 Chr.5: 122817315-122817315 on Build GRCh38
rs755768250 Chr.5: 122817416-122817416 on Build GRCh38
rs762907221 Chr.5: 122817346-122817346 on Build GRCh38
rs755360049 Chr.5: 122817297-122817297 on Build GRCh38
rs778437567 Chr.5: 122817424-122817424 on Build GRCh38
rs777956116 Chr.5: 122817316-122817316 on Build GRCh38
rs762829791 Chr.5: 122817396-122817396 on Build GRCh38
rs189518915 Chr.5: 122817371-122817371 on Build GRCh38
rs761404128 Chr.5: 122817406-122817406 on Build GRCh38
rs372930896 Chr.5: 122817395-122817395 on Build GRCh38
rs762697216 Chr.5: 122817423-122817424 on Build GRCh38
rs79671754 Chr.5: 122817268-122817268 on Build GRCh38
rs748639812 Chr.5: 122817303-122817303 on Build GRCh38
rs758904917 Chr.5: 122817420-122817420 on Build GRCh38
rs367637115 Chr.5: 122817347-122817347 on Build GRCh38
rs765645555 Chr.5: 122817417-122817417 on Build GRCh38
rs982191229 Chr.5: 122817362-122817362 on Build GRCh38

View Full Product Details