Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123907162-123907397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8D4
Entrez Gene ID
338662
Gene Name
olfactory receptor family 8 subfamily D member 4
Gene Aliases
OR11-275
UniGene
Hs.449688
Gene Chromosome Location
Chr.11: 123906432-123907376 on Build GRCh38
Transcripts
NM_001005197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5922916 c.935C>T p.S312F Chr.11: 123907366-123907366 on Build GRCh38
4455541 c.814A>T p.K272* Chr.11: 123907245-123907245 on Build GRCh38
6067766 c.757G>T p.G253W Chr.11: 123907188-123907188 on Build GRCh38
6067765 c.928T>A p.S310T Chr.11: 123907359-123907359 on Build GRCh38
1604236 c.752T>A p.F251Y Chr.11: 123907183-123907183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749430558 Chr.11: 123907296-123907297 on Build GRCh38
rs745689178 Chr.11: 123907198-123907198 on Build GRCh38
rs781212400 Chr.11: 123907263-123907263 on Build GRCh38
rs1018399296 Chr.11: 123907278-123907278 on Build GRCh38
rs774107622 Chr.11: 123907191-123907191 on Build GRCh38
rs763444148 Chr.11: 123907356-123907356 on Build GRCh38
rs146310490 Chr.11: 123907332-123907332 on Build GRCh38
rs760392980 Chr.11: 123907205-123907205 on Build GRCh38
rs978160525 Chr.11: 123907216-123907216 on Build GRCh38
rs573839478 Chr.11: 123907305-123907305 on Build GRCh38
rs200968079 Chr.11: 123907214-123907214 on Build GRCh38
rs765799707 Chr.11: 123907236-123907236 on Build GRCh38
rs771963445 Chr.11: 123907203-123907203 on Build GRCh38
rs759500251 Chr.11: 123907342-123907343 on Build GRCh38
rs369728924 Chr.11: 123907301-123907301 on Build GRCh38
rs149621046 Chr.11: 123907212-123907212 on Build GRCh38
rs761174077 Chr.11: 123907233-123907233 on Build GRCh38
rs755231455 Chr.11: 123907249-123907249 on Build GRCh38
rs7927385 Chr.11: 123907324-123907324 on Build GRCh38
rs750894709 Chr.11: 123907238-123907238 on Build GRCh38
rs779034358 Chr.11: 123907282-123907282 on Build GRCh38
rs796639654 Chr.11: 123907200-123907200 on Build GRCh38
rs773763437 Chr.11: 123907350-123907350 on Build GRCh38
rs775423940 Chr.11: 123907299-123907299 on Build GRCh38
rs748282165 Chr.11: 123907270-123907270 on Build GRCh38
rs751933829 Chr.11: 123907362-123907362 on Build GRCh38
rs201836268 Chr.11: 123907206-123907206 on Build GRCh38
rs7942047 Chr.11: 123907279-123907279 on Build GRCh38
rs199888408 Chr.11: 123907190-123907190 on Build GRCh38
rs772051731 Chr.11: 123907296-123907296 on Build GRCh38
rs766803470 Chr.11: 123907358-123907358 on Build GRCh38
rs758808062 Chr.11: 123907242-123907242 on Build GRCh38
rs375243595 Chr.11: 123907208-123907208 on Build GRCh38
rs867276686 Chr.11: 123907310-123907310 on Build GRCh38
rs148753800 Chr.11: 123907319-123907319 on Build GRCh38
rs543598139 Chr.11: 123907248-123907248 on Build GRCh38
rs866199393 Chr.11: 123907290-123907290 on Build GRCh38
rs1048445399 Chr.11: 123907202-123907202 on Build GRCh38
rs745779154 Chr.11: 123907292-123907292 on Build GRCh38

View Full Product Details