Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93131429-93131658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD9L
Entrez Gene ID
219285
Gene Name
sterile alpha motif domain containing 9 like
Gene Aliases
C7orf6, DRIF2, UEF1
UniGene
Hs.489118
Gene Chromosome Location
Chr.7: 93130054-93148401 on Build GRCh38
Transcripts
NM_001303496.1 NM_001303497.1 NM_001303498.1 NM_001303500.1 NM_152703.3 XM_005250193.3 XM_006715890.1 XM_011515903.2 XM_017011822.1 XM_017011823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093129 c.4503A>C p.Q1501H Chr.7: 93131469-93131469 on Build GRCh38
747525 c.4482G>T p.E1494D Chr.7: 93131490-93131490 on Build GRCh38
5505220 c.4409A>G p.K1470R Chr.7: 93131563-93131563 on Build GRCh38
6771780 c.4422A>G p.T1474T Chr.7: 93131550-93131550 on Build GRCh38
1093130 c.4393C>T p.R1465C Chr.7: 93131579-93131579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781060659 Chr.7: 93131540-93131540 on Build GRCh38
rs765115425 Chr.7: 93131513-93131513 on Build GRCh38
rs200615225 Chr.7: 93131460-93131460 on Build GRCh38
rs267601632 Chr.7: 93131599-93131599 on Build GRCh38
rs756342171 Chr.7: 93131543-93131543 on Build GRCh38
rs781121824 Chr.7: 93131536-93131536 on Build GRCh38
rs778102246 Chr.7: 93131617-93131617 on Build GRCh38
rs763687130 Chr.7: 93131582-93131582 on Build GRCh38
rs776292434 Chr.7: 93131583-93131583 on Build GRCh38
rs201797999 Chr.7: 93131598-93131598 on Build GRCh38
rs202145032 Chr.7: 93131490-93131490 on Build GRCh38
rs149666036 Chr.7: 93131538-93131538 on Build GRCh38
rs372802865 Chr.7: 93131464-93131464 on Build GRCh38
rs144026608 Chr.7: 93131604-93131604 on Build GRCh38
rs750287709 Chr.7: 93131593-93131593 on Build GRCh38
rs775122107 Chr.7: 93131480-93131480 on Build GRCh38
rs750294178 Chr.7: 93131517-93131517 on Build GRCh38
rs749620937 Chr.7: 93131550-93131550 on Build GRCh38
rs778159444 Chr.7: 93131544-93131544 on Build GRCh38
rs764992405 Chr.7: 93131592-93131592 on Build GRCh38
rs190393069 Chr.7: 93131578-93131578 on Build GRCh38
rs759047710 Chr.7: 93131463-93131463 on Build GRCh38
rs758407828 Chr.7: 93131535-93131535 on Build GRCh38
rs139201196 Chr.7: 93131589-93131589 on Build GRCh38
rs771454708 Chr.7: 93131551-93131551 on Build GRCh38
rs181466556 Chr.7: 93131579-93131579 on Build GRCh38
rs774821462 Chr.7: 93131559-93131559 on Build GRCh38
rs772700651 Chr.7: 93131575-93131575 on Build GRCh38
rs968445739 Chr.7: 93131547-93131547 on Build GRCh38
rs139478067 Chr.7: 93131510-93131510 on Build GRCh38
rs752665363 Chr.7: 93131601-93131601 on Build GRCh38
rs1005620157 Chr.7: 93131454-93131454 on Build GRCh38
rs572977789 Chr.7: 93131502-93131502 on Build GRCh38
rs368871404 Chr.7: 93131600-93131600 on Build GRCh38
rs200166375 Chr.7: 93131481-93131481 on Build GRCh38
rs746341432 Chr.7: 93131561-93131561 on Build GRCh38

View Full Product Details