Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92385629-92385876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 XM_011518689.1 NM_014057.4 NM_024416.4 NM_033014.3

Gene Symbol
OGN
Entrez Gene ID
4969
Gene Name
osteoglycin
Gene Aliases
OG, OIF, SLRR3A
UniGene
Hs.109439
Gene Chromosome Location
Chr.9: 92383271-92404699 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 XM_011518689.1 NM_014057.4 NM_024416.4 NM_033014.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934853 c.878C>G p.P293R Chr.9: 92385639-92385639 on Build GRCh38
456235 c.839G>C p.G280A Chr.9: 92385678-92385678 on Build GRCh38
219974 c.774T>A p.N258K Chr.9: 92385743-92385743 on Build GRCh38
6235331 c.831C>A p.I277I Chr.9: 92385686-92385686 on Build GRCh38
339929 c.790C>A p.R264R Chr.9: 92385727-92385727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766769286 Chr.9: 92385721-92385721 on Build GRCh38
rs946099759 Chr.9: 92385812-92385812 on Build GRCh38
rs770580940 Chr.9: 92385720-92385720 on Build GRCh38
rs371372601 Chr.9: 92385701-92385701 on Build GRCh38
rs142523561 Chr.9: 92385680-92385680 on Build GRCh38
rs755382157 Chr.9: 92385782-92385782 on Build GRCh38
rs147797196 Chr.9: 92385786-92385786 on Build GRCh38
rs758144658 Chr.9: 92385831-92385838 on Build GRCh38
rs150923091 Chr.9: 92385726-92385726 on Build GRCh38
rs777501525 Chr.9: 92385717-92385717 on Build GRCh38
rs750625278 Chr.9: 92385683-92385683 on Build GRCh38
rs771916853 Chr.9: 92385742-92385742 on Build GRCh38
rs2236529 Chr.9: 92385802-92385802 on Build GRCh38
rs139749001 Chr.9: 92385670-92385670 on Build GRCh38
rs367640138 Chr.9: 92385743-92385743 on Build GRCh38
rs758663034 Chr.9: 92385685-92385685 on Build GRCh38
rs1046079460 Chr.9: 92385706-92385706 on Build GRCh38
rs754252404 Chr.9: 92385763-92385763 on Build GRCh38
rs879172047 Chr.9: 92385805-92385805 on Build GRCh38
rs749276412 Chr.9: 92385718-92385718 on Build GRCh38
rs763692574 Chr.9: 92385761-92385761 on Build GRCh38
rs765557463 Chr.9: 92385773-92385773 on Build GRCh38
rs1017029541 Chr.9: 92385658-92385658 on Build GRCh38
rs758609787 Chr.9: 92385794-92385794 on Build GRCh38
rs760037955 Chr.9: 92385751-92385751 on Build GRCh38
rs754435795 Chr.9: 92385780-92385781 on Build GRCh38
rs750857725 Chr.9: 92385792-92385792 on Build GRCh38
rs140783624 Chr.9: 92385727-92385727 on Build GRCh38
rs754968978 Chr.9: 92385839-92385839 on Build GRCh38
rs765711204 Chr.9: 92385793-92385794 on Build GRCh38
rs375943971 Chr.9: 92385721-92385721 on Build GRCh38
rs754374602 Chr.9: 92385668-92385668 on Build GRCh38
rs149681015 Chr.9: 92385772-92385772 on Build GRCh38
rs752333372 Chr.9: 92385794-92385794 on Build GRCh38
rs552552673 Chr.9: 92385667-92385667 on Build GRCh38
rs367913185 Chr.9: 92385705-92385705 on Build GRCh38
rs780107933 Chr.9: 92385696-92385696 on Build GRCh38

View Full Product Details