Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1950859-1951084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL3L
Entrez Gene ID
6123
Gene Name
ribosomal protein L3 like
Gene Aliases
-
UniGene
Hs.657266
Gene Chromosome Location
Chr.16: 1944571-1955123 on Build GRCh38
Transcripts
NM_005061.2 XM_011522571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5384381 c.441G>A p.Q147Q Chr.16: 1950904-1950904 on Build GRCh38
5579483 c.426G>A p.G142G Chr.16: 1950919-1950919 on Build GRCh38
3506830 c.429A>T p.K143N Chr.16: 1950916-1950916 on Build GRCh38
4884542 c.374G>T p.S125I Chr.16: 1950971-1950971 on Build GRCh38
4496751 c.481C>T p.R161W Chr.16: 1950864-1950864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913371063 Chr.16: 1951065-1951065 on Build GRCh38
rs749717183 Chr.16: 1950937-1950937 on Build GRCh38
rs560164168 Chr.16: 1950984-1950984 on Build GRCh38
rs1021322167 Chr.16: 1950936-1950936 on Build GRCh38
rs529244017 Chr.16: 1950985-1950985 on Build GRCh38
rs769175427 Chr.16: 1950942-1950942 on Build GRCh38
rs961881573 Chr.16: 1950949-1950949 on Build GRCh38
rs371249692 Chr.16: 1950922-1950922 on Build GRCh38
rs967129764 Chr.16: 1950965-1950965 on Build GRCh38
rs373443605 Chr.16: 1950901-1950901 on Build GRCh38
rs555349951 Chr.16: 1950891-1950891 on Build GRCh38
rs367870263 Chr.16: 1951016-1951016 on Build GRCh38
rs770836963 Chr.16: 1950935-1950936 on Build GRCh38
rs929392984 Chr.16: 1951040-1951040 on Build GRCh38
rs761976111 Chr.16: 1950879-1950879 on Build GRCh38
rs749178872 Chr.16: 1950881-1950881 on Build GRCh38
rs184860646 Chr.16: 1950910-1950910 on Build GRCh38
rs371438665 Chr.16: 1951020-1951020 on Build GRCh38
rs187547533 Chr.16: 1950998-1950998 on Build GRCh38
rs897893004 Chr.16: 1951034-1951034 on Build GRCh38
rs199584535 Chr.16: 1951009-1951009 on Build GRCh38
rs758322610 Chr.16: 1950995-1950995 on Build GRCh38
rs758052886 Chr.16: 1950906-1950906 on Build GRCh38
rs774643139 Chr.16: 1950955-1950955 on Build GRCh38
rs977039952 Chr.16: 1950990-1950990 on Build GRCh38
rs767741903 Chr.16: 1950882-1950882 on Build GRCh38
rs148464643 Chr.16: 1950889-1950889 on Build GRCh38
rs201908489 Chr.16: 1950900-1950900 on Build GRCh38
rs751529301 Chr.16: 1951031-1951031 on Build GRCh38
rs150997667 Chr.16: 1950895-1950895 on Build GRCh38
rs779464047 Chr.16: 1951018-1951018 on Build GRCh38
rs942165554 Chr.16: 1951029-1951029 on Build GRCh38
rs773367998 Chr.16: 1950971-1950971 on Build GRCh38
rs760790279 Chr.16: 1950972-1950972 on Build GRCh38
rs747967218 Chr.16: 1950902-1950902 on Build GRCh38
rs762170719 Chr.16: 1950961-1950961 on Build GRCh38
rs765268036 Chr.16: 1950986-1950986 on Build GRCh38
rs140887504 Chr.16: 1950932-1950932 on Build GRCh38
rs746514363 Chr.16: 1950911-1950911 on Build GRCh38
rs140731138 Chr.16: 1951056-1951056 on Build GRCh38
rs142682876 Chr.16: 1950892-1950892 on Build GRCh38
rs375794746 Chr.16: 1950968-1950968 on Build GRCh38
rs753985680 Chr.16: 1950982-1950982 on Build GRCh38
rs569187579 Chr.16: 1951019-1951019 on Build GRCh38
rs771210878 Chr.16: 1951030-1951030 on Build GRCh38
rs752531898 Chr.16: 1950988-1950988 on Build GRCh38
rs756963325 Chr.16: 1950999-1950999 on Build GRCh38
rs540319038 Chr.16: 1950976-1950976 on Build GRCh38
rs200927090 Chr.16: 1950894-1950894 on Build GRCh38
rs745398676 Chr.16: 1951010-1951010 on Build GRCh38
rs775726501 Chr.16: 1950960-1950961 on Build GRCh38

View Full Product Details