Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44141178-44141421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL7
Entrez Gene ID
58498
Gene Name
myosin light chain 7
Gene Aliases
MYL2A, MYLC2A
UniGene
Hs.75636
Gene Chromosome Location
Chr.7: 44138864-44141392 on Build GRCh38
Transcripts
XM_005249817.4 XM_011515463.2 XM_011515464.2 XM_011515465.2 XM_017012478.1 XM_017012479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1089918 c.3+8C>T p.? Chr.7: 44141295-44141295 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758826658 Chr.7: 44141353-44141353 on Build GRCh38
rs757548980 Chr.7: 44141219-44141219 on Build GRCh38
rs760672044 Chr.7: 44141300-44141300 on Build GRCh38
rs759377363 Chr.7: 44141279-44141279 on Build GRCh38
rs2908278 Chr.7: 44141333-44141333 on Build GRCh38
rs762487962 Chr.7: 44141266-44141266 on Build GRCh38
rs747580801 Chr.7: 44141257-44141257 on Build GRCh38
rs756198688 Chr.7: 44141209-44141209 on Build GRCh38
rs964107731 Chr.7: 44141351-44141351 on Build GRCh38
rs779202551 Chr.7: 44141343-44141343 on Build GRCh38
rs772395394 Chr.7: 44141240-44141240 on Build GRCh38
rs764137092 Chr.7: 44141301-44141301 on Build GRCh38
rs192113493 Chr.7: 44141272-44141272 on Build GRCh38
rs374644501 Chr.7: 44141245-44141245 on Build GRCh38
rs201572348 Chr.7: 44141327-44141327 on Build GRCh38
rs779100561 Chr.7: 44141221-44141221 on Build GRCh38
rs572358786 Chr.7: 44141332-44141332 on Build GRCh38
rs375947803 Chr.7: 44141263-44141263 on Build GRCh38
rs746277231 Chr.7: 44141237-44141237 on Build GRCh38
rs538817433 Chr.7: 44141273-44141273 on Build GRCh38
rs761884213 Chr.7: 44141222-44141222 on Build GRCh38
rs941181956 Chr.7: 44141202-44141202 on Build GRCh38
rs1046111816 Chr.7: 44141248-44141248 on Build GRCh38
rs767383892 Chr.7: 44141280-44141280 on Build GRCh38
rs769428596 Chr.7: 44141259-44141259 on Build GRCh38
rs777886881 Chr.7: 44141214-44141214 on Build GRCh38
rs752617022 Chr.7: 44141287-44141287 on Build GRCh38
rs556312224 Chr.7: 44141216-44141216 on Build GRCh38
rs750697614 Chr.7: 44141344-44141344 on Build GRCh38
rs1014353393 Chr.7: 44141235-44141235 on Build GRCh38

View Full Product Details