Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49962767-49963016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6821
Entrez Gene ID
102465493
Gene Name
microRNA 6821
Gene Aliases
hsa-mir-6821
UniGene
Gene Chromosome Location
Chr.22: 49962866-49962939 on Build GRCh38
Transcripts
NR_106879.1 NM_001001852.3

Gene Symbol
PIM3
Entrez Gene ID
415116
Gene Name
Pim-3 proto-oncogene, serine/threonine kinase
Gene Aliases
pim-3
UniGene
Hs.530381
Gene Chromosome Location
Chr.22: 49960495-49964072 on Build GRCh38
Transcripts
NR_106879.1 NM_001001852.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5366527 c.794A>G p.E265G Chr.22: 49962940-49962940 on Build GRCh38
5393862 c.705C>T p.L235L Chr.22: 49962777-49962777 on Build GRCh38
1416958 c.738G>A p.E246E Chr.22: 49962810-49962810 on Build GRCh38
5515806 c.840G>A p.E280E Chr.22: 49962986-49962986 on Build GRCh38
5617067 c.794-4A>G p.? Chr.22: 49962936-49962936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755459715 Chr.22: 49962896-49962896 on Build GRCh38
rs773177526 Chr.22: 49962864-49962864 on Build GRCh38
rs761757255 Chr.22: 49962885-49962885 on Build GRCh38
rs537004314 Chr.22: 49962934-49962934 on Build GRCh38
rs1027267181 Chr.22: 49962987-49962987 on Build GRCh38
rs901127837 Chr.22: 49962796-49962796 on Build GRCh38
rs1036969611 Chr.22: 49962944-49962944 on Build GRCh38
rs762412577 Chr.22: 49962929-49962929 on Build GRCh38
rs961249882 Chr.22: 49962843-49962843 on Build GRCh38
rs368530413 Chr.22: 49962932-49962932 on Build GRCh38
rs769318310 Chr.22: 49962812-49962812 on Build GRCh38
rs897950950 Chr.22: 49962972-49962972 on Build GRCh38
rs754335791 Chr.22: 49962950-49962950 on Build GRCh38
rs377265889 Chr.22: 49962937-49962937 on Build GRCh38
rs746080812 Chr.22: 49962910-49962910 on Build GRCh38
rs756692068 Chr.22: 49962899-49962899 on Build GRCh38
rs775149141 Chr.22: 49962838-49962838 on Build GRCh38
rs555006458 Chr.22: 49962914-49962914 on Build GRCh38
rs770713831 Chr.22: 49962976-49962976 on Build GRCh38
rs370175675 Chr.22: 49962893-49962893 on Build GRCh38
rs139983531 Chr.22: 49962825-49962825 on Build GRCh38
rs749323042 Chr.22: 49962869-49962869 on Build GRCh38
rs771018838 Chr.22: 49962905-49962905 on Build GRCh38
rs891126389 Chr.22: 49962808-49962808 on Build GRCh38
rs118127603 Chr.22: 49962849-49962849 on Build GRCh38
rs1015894680 Chr.22: 49962834-49962834 on Build GRCh38
rs147698351 Chr.22: 49962840-49962840 on Build GRCh38
rs779066356 Chr.22: 49962958-49962958 on Build GRCh38
rs773091453 Chr.22: 49962971-49962971 on Build GRCh38
rs753142995 Chr.22: 49962933-49962933 on Build GRCh38
rs751313726 Chr.22: 49962931-49962931 on Build GRCh38
rs567243898 Chr.22: 49962801-49962801 on Build GRCh38
rs747332665 Chr.22: 49962810-49962810 on Build GRCh38
rs764038607 Chr.22: 49962841-49962841 on Build GRCh38
rs749895033 Chr.22: 49962850-49962850 on Build GRCh38
rs745885702 Chr.22: 49962959-49962959 on Build GRCh38
rs763073107 Chr.22: 49962835-49962835 on Build GRCh38
rs773179821 Chr.22: 49962927-49962927 on Build GRCh38
rs889176956 Chr.22: 49962819-49962819 on Build GRCh38
rs780300012 Chr.22: 49962809-49962809 on Build GRCh38
rs762933246 Chr.22: 49962975-49962975 on Build GRCh38
rs757994179 Chr.22: 49962852-49962852 on Build GRCh38
rs781774947 Chr.22: 49962860-49962860 on Build GRCh38
rs1028316890 Chr.22: 49962798-49962798 on Build GRCh38
rs778434698 Chr.22: 49962848-49962848 on Build GRCh38
rs376408360 Chr.22: 49962870-49962870 on Build GRCh38
rs772227615 Chr.22: 49962911-49962911 on Build GRCh38
rs774002494 Chr.22: 49962981-49962981 on Build GRCh38
rs926809294 Chr.22: 49962884-49962884 on Build GRCh38
rs747355686 Chr.22: 49962856-49962856 on Build GRCh38
rs148936816 Chr.22: 49962817-49962817 on Build GRCh38
rs780201470 Chr.22: 49962968-49962968 on Build GRCh38
rs1028190314 Chr.22: 49962807-49962807 on Build GRCh38
rs772488592 Chr.22: 49962791-49962791 on Build GRCh38
rs372643241 Chr.22: 49962930-49962930 on Build GRCh38
rs778189349 Chr.22: 49962942-49962942 on Build GRCh38
rs4078443 Chr.22: 49962907-49962907 on Build GRCh38
rs757341562 Chr.22: 49962956-49962956 on Build GRCh38
rs760508730 Chr.22: 49962879-49962879 on Build GRCh38
rs745505989 Chr.22: 49962799-49962799 on Build GRCh38
rs773633337 Chr.22: 49962826-49962826 on Build GRCh38
rs145509761 Chr.22: 49962881-49962881 on Build GRCh38
rs201559644 Chr.22: 49962858-49962858 on Build GRCh38
rs201620023 Chr.22: 49962855-49962855 on Build GRCh38
rs749345331 Chr.22: 49962900-49962900 on Build GRCh38
rs374423701 Chr.22: 49962922-49962922 on Build GRCh38
rs749933111 Chr.22: 49962889-49962889 on Build GRCh38
rs547561980 Chr.22: 49962880-49962880 on Build GRCh38
rs767296630 Chr.22: 49962991-49962991 on Build GRCh38
rs1049567483 Chr.22: 49962923-49962923 on Build GRCh38
rs778580867 Chr.22: 49962789-49962789 on Build GRCh38
rs766565240 Chr.22: 49962837-49962837 on Build GRCh38
rs765096620 Chr.22: 49962888-49962888 on Build GRCh38
rs759077126 Chr.22: 49962986-49962986 on Build GRCh38
rs539004244 Chr.22: 49962970-49962970 on Build GRCh38
rs758242581 Chr.22: 49962931-49962932 on Build GRCh38
rs200639371 Chr.22: 49962876-49962876 on Build GRCh38
rs757876078 Chr.22: 49962890-49962890 on Build GRCh38
rs777606422 Chr.22: 49962967-49962968 on Build GRCh38
rs142498730 Chr.22: 49962847-49962847 on Build GRCh38
rs377375610 Chr.22: 49962863-49962863 on Build GRCh38
rs752489243 Chr.22: 49962874-49962875 on Build GRCh38
rs369976635 Chr.22: 49962962-49962962 on Build GRCh38
rs901074036 Chr.22: 49962916-49962916 on Build GRCh38
rs15682 Chr.22: 49962980-49962980 on Build GRCh38
rs751194119 Chr.22: 49962895-49962895 on Build GRCh38

View Full Product Details