Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72603022-72603289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHKA1
Entrez Gene ID
5255
Gene Name
phosphorylase kinase regulatory subunit alpha 1
Gene Aliases
PHKA
UniGene
Hs.201379
Gene Chromosome Location
Chr.X: 72578814-72714181 on Build GRCh38
Transcripts
NM_001122670.1 NM_001172436.1 NM_002637.3 XM_006724661.2

SNP Detail

SNP ID Chromosome Location
rs782702439 Chr.X: 72603124-72603124 on Build GRCh38
rs371624021 Chr.X: 72603226-72603226 on Build GRCh38
rs782477256 Chr.X: 72603165-72603165 on Build GRCh38
rs782415920 Chr.X: 72603068-72603068 on Build GRCh38
rs781958738 Chr.X: 72603106-72603106 on Build GRCh38
rs782455291 Chr.X: 72603201-72603201 on Build GRCh38
rs782102345 Chr.X: 72603120-72603120 on Build GRCh38
rs367798015 Chr.X: 72603146-72603146 on Build GRCh38
rs782155843 Chr.X: 72603196-72603196 on Build GRCh38
rs782752386 Chr.X: 72603087-72603088 on Build GRCh38
rs782119322 Chr.X: 72603090-72603090 on Build GRCh38
rs782036365 Chr.X: 72603088-72603088 on Build GRCh38
rs782674118 Chr.X: 72603197-72603197 on Build GRCh38
rs963566964 Chr.X: 72603181-72603181 on Build GRCh38
rs782772541 Chr.X: 72603095-72603095 on Build GRCh38
rs782602098 Chr.X: 72603180-72603180 on Build GRCh38
rs782690754 Chr.X: 72603245-72603245 on Build GRCh38

View Full Product Details