Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37927556-37927781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICALL1
Entrez Gene ID
85377
Gene Name
MICAL like 1
Gene Aliases
MICAL-L1, MIRAB13
UniGene
Hs.517610
Gene Chromosome Location
Chr.22: 37906148-37942458 on Build GRCh38
Transcripts
NM_033386.3 XM_005261791.3 XM_005261792.3 XM_011530471.2 XM_011530472.2 XM_011530473.2 XM_011530474.2 XM_011530475.2 XM_017029026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5555763 c.1739G>A p.G580D Chr.22: 37927684-37927684 on Build GRCh38
7134826 c.1613C>T p.S538L Chr.22: 37927558-37927558 on Build GRCh38
6712623 c.1792A>T p.S598C Chr.22: 37927737-37927737 on Build GRCh38
1318768 c.1761C>A p.G587G Chr.22: 37927706-37927706 on Build GRCh38
6276446 c.1752G>T p.R584S Chr.22: 37927697-37927697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140312542 Chr.22: 37927743-37927743 on Build GRCh38
rs933723588 Chr.22: 37927601-37927601 on Build GRCh38
rs373880028 Chr.22: 37927644-37927644 on Build GRCh38
rs558153807 Chr.22: 37927754-37927754 on Build GRCh38
rs747113274 Chr.22: 37927640-37927640 on Build GRCh38
rs567955945 Chr.22: 37927589-37927589 on Build GRCh38
rs776672075 Chr.22: 37927659-37927659 on Build GRCh38
rs369177724 Chr.22: 37927619-37927619 on Build GRCh38
rs778018992 Chr.22: 37927726-37927726 on Build GRCh38
rs781121592 Chr.22: 37927740-37927740 on Build GRCh38
rs751483044 Chr.22: 37927761-37927763 on Build GRCh38
rs907397105 Chr.22: 37927753-37927753 on Build GRCh38
rs775163590 Chr.22: 37927684-37927684 on Build GRCh38
rs772284925 Chr.22: 37927638-37927638 on Build GRCh38
rs775532370 Chr.22: 37927751-37927751 on Build GRCh38
rs138003560 Chr.22: 37927722-37927722 on Build GRCh38
rs754423523 Chr.22: 37927714-37927714 on Build GRCh38
rs747201365 Chr.22: 37927734-37927734 on Build GRCh38
rs147148915 Chr.22: 37927738-37927738 on Build GRCh38
rs897655503 Chr.22: 37927744-37927744 on Build GRCh38
rs759388082 Chr.22: 37927662-37927662 on Build GRCh38
rs554001124 Chr.22: 37927628-37927628 on Build GRCh38
rs141260509 Chr.22: 37927711-37927711 on Build GRCh38
rs144172126 Chr.22: 37927758-37927758 on Build GRCh38
rs376904774 Chr.22: 37927618-37927618 on Build GRCh38
rs752463321 Chr.22: 37927585-37927585 on Build GRCh38
rs758713733 Chr.22: 37927725-37927725 on Build GRCh38
rs769691240 Chr.22: 37927669-37927669 on Build GRCh38
rs78227046 Chr.22: 37927604-37927604 on Build GRCh38
rs773383148 Chr.22: 37927639-37927639 on Build GRCh38
rs745905166 Chr.22: 37927741-37927741 on Build GRCh38
rs762866949 Chr.22: 37927752-37927752 on Build GRCh38
rs761444701 Chr.22: 37927709-37927709 on Build GRCh38
rs780017679 Chr.22: 37927633-37927633 on Build GRCh38
rs756791925 Chr.22: 37927605-37927605 on Build GRCh38
rs41280823 Chr.22: 37927656-37927656 on Build GRCh38
rs763783825 Chr.22: 37927599-37927599 on Build GRCh38
rs2272829 Chr.22: 37927693-37927693 on Build GRCh38
rs545892604 Chr.22: 37927694-37927694 on Build GRCh38
rs764129404 Chr.22: 37927584-37927585 on Build GRCh38
rs751231683 Chr.22: 37927697-37927697 on Build GRCh38
rs149483529 Chr.22: 37927723-37927723 on Build GRCh38
rs755451444 Chr.22: 37927716-37927716 on Build GRCh38

View Full Product Details