Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81834795-81835020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R27
Entrez Gene ID
116729
Gene Name
protein phosphatase 1 regulatory subunit 27
Gene Aliases
DYSFIP1
UniGene
Hs.454720
Gene Chromosome Location
Chr.17: 81833492-81835050 on Build GRCh38
Transcripts
NM_001007533.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6754053 c.79C>T p.R27C Chr.17: 81834875-81834875 on Build GRCh38
4887597 c.121G>A p.G41S Chr.17: 81834833-81834833 on Build GRCh38
6754052 c.74G>C p.S25T Chr.17: 81834880-81834880 on Build GRCh38
4717101 c.141delG p.R48fs*33 Chr.17: 81834813-81834813 on Build GRCh38
6754049 c.152G>A p.R51Q Chr.17: 81834802-81834802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373055230 Chr.17: 81834831-81834831 on Build GRCh38
rs771561258 Chr.17: 81834828-81834828 on Build GRCh38
rs776322601 Chr.17: 81834899-81834899 on Build GRCh38
rs375799258 Chr.17: 81834905-81834905 on Build GRCh38
rs759372826 Chr.17: 81834841-81834841 on Build GRCh38
rs756399012 Chr.17: 81834926-81834926 on Build GRCh38
rs748651166 Chr.17: 81834936-81834936 on Build GRCh38
rs192958732 Chr.17: 81834960-81834960 on Build GRCh38
rs148935616 Chr.17: 81834914-81834914 on Build GRCh38
rs369002344 Chr.17: 81834812-81834812 on Build GRCh38
rs767634444 Chr.17: 81834985-81834985 on Build GRCh38
rs535491596 Chr.17: 81834840-81834840 on Build GRCh38
rs756272222 Chr.17: 81834989-81834989 on Build GRCh38
rs750635325 Chr.17: 81834925-81834925 on Build GRCh38
rs745446292 Chr.17: 81834954-81834954 on Build GRCh38
rs746553122 Chr.17: 81834896-81834896 on Build GRCh38
rs777476929 Chr.17: 81834897-81834899 on Build GRCh38
rs763837929 Chr.17: 81834961-81834961 on Build GRCh38
rs763503378 Chr.17: 81834821-81834821 on Build GRCh38
rs1052798862 Chr.17: 81834915-81834915 on Build GRCh38
rs543979508 Chr.17: 81834913-81834913 on Build GRCh38
rs558362319 Chr.17: 81834893-81834893 on Build GRCh38
rs757369949 Chr.17: 81834869-81834869 on Build GRCh38
rs372589539 Chr.17: 81834935-81834935 on Build GRCh38
rs756031327 Chr.17: 81834828-81834829 on Build GRCh38
rs780331549 Chr.17: 81834929-81834929 on Build GRCh38
rs769210919 Chr.17: 81834884-81834884 on Build GRCh38
rs372728244 Chr.17: 81834934-81834934 on Build GRCh38
rs776347342 Chr.17: 81834948-81834948 on Build GRCh38
rs775373280 Chr.17: 81834959-81834959 on Build GRCh38
rs369174976 Chr.17: 81834883-81834883 on Build GRCh38
rs761719447 Chr.17: 81834970-81834970 on Build GRCh38
rs781512031 Chr.17: 81834874-81834874 on Build GRCh38
rs894621524 Chr.17: 81834992-81834992 on Build GRCh38
rs750506957 Chr.17: 81834987-81834987 on Build GRCh38
rs770458996 Chr.17: 81834943-81834943 on Build GRCh38
rs769580300 Chr.17: 81834958-81834958 on Build GRCh38
rs950737148 Chr.17: 81834838-81834838 on Build GRCh38
rs370229834 Chr.17: 81834908-81834908 on Build GRCh38
rs533762365 Chr.17: 81834864-81834864 on Build GRCh38
rs749139905 Chr.17: 81834907-81834908 on Build GRCh38
rs138820775 Chr.17: 81834813-81834813 on Build GRCh38
rs752668582 Chr.17: 81834847-81834847 on Build GRCh38
rs147812243 Chr.17: 81834839-81834839 on Build GRCh38
rs775949807 Chr.17: 81834816-81834816 on Build GRCh38
rs546095206 Chr.17: 81834865-81834865 on Build GRCh38
rs145719058 Chr.17: 81834907-81834907 on Build GRCh38
rs750675769 Chr.17: 81834904-81834904 on Build GRCh38
rs772541862 Chr.17: 81834888-81834888 on Build GRCh38
rs911043795 Chr.17: 81834837-81834837 on Build GRCh38
rs756452367 Chr.17: 81834875-81834875 on Build GRCh38
rs758476192 Chr.17: 81834858-81834858 on Build GRCh38
rs372449567 Chr.17: 81834846-81834846 on Build GRCh38
rs905727675 Chr.17: 81834976-81834976 on Build GRCh38
rs765118856 Chr.17: 81834902-81834902 on Build GRCh38
rs770386678 Chr.17: 81834898-81834898 on Build GRCh38
rs946851998 Chr.17: 81834901-81834901 on Build GRCh38
rs541368350 Chr.17: 81834963-81834963 on Build GRCh38
rs780458528 Chr.17: 81834878-81834878 on Build GRCh38

View Full Product Details