Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245915423-245915694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMYD3
Entrez Gene ID
64754
Gene Name
SET and MYND domain containing 3
Gene Aliases
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1
UniGene
Hs.567571
Gene Chromosome Location
Chr.1: 245749340-246507342 on Build GRCh38
Transcripts
NM_001167740.1 NM_022743.2 XM_011544253.1 XM_011544254.2 XM_011544257.1 XM_011544258.2 XM_011544259.2 XM_011544260.1 XM_011544261.2 XM_017002094.1 XM_017002095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4509000 c.793C>T p.R265C Chr.1: 245915550-245915550 on Build GRCh38
4960037 c.741G>A p.E247E Chr.1: 245915602-245915602 on Build GRCh38
6785835 c.617G>A p.R206H Chr.1: 245915549-245915549 on Build GRCh38
4953468 c.779A>T p.E260V Chr.1: 245915564-245915564 on Build GRCh38
906833 c.636+2T>C p.? Chr.1: 245915528-245915528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756254559 Chr.1: 245915570-245915570 on Build GRCh38
rs754089601 Chr.1: 245915574-245915574 on Build GRCh38
rs115557356 Chr.1: 245915509-245915509 on Build GRCh38
rs758096229 Chr.1: 245915500-245915500 on Build GRCh38
rs749588870 Chr.1: 245915498-245915498 on Build GRCh38
rs143716511 Chr.1: 245915634-245915634 on Build GRCh38
rs769353259 Chr.1: 245915533-245915533 on Build GRCh38
rs143461843 Chr.1: 245915659-245915659 on Build GRCh38
rs370744880 Chr.1: 245915501-245915501 on Build GRCh38
rs757450128 Chr.1: 245915580-245915580 on Build GRCh38
rs764309201 Chr.1: 245915571-245915571 on Build GRCh38
rs61750995 Chr.1: 245915545-245915545 on Build GRCh38
rs952048801 Chr.1: 245915552-245915552 on Build GRCh38
rs375605550 Chr.1: 245915480-245915480 on Build GRCh38
rs987211168 Chr.1: 245915594-245915594 on Build GRCh38
rs61762672 Chr.1: 245915549-245915549 on Build GRCh38
rs748774049 Chr.1: 245915631-245915631 on Build GRCh38
rs12065652 Chr.1: 245915522-245915522 on Build GRCh38
rs370162934 Chr.1: 245915483-245915483 on Build GRCh38
rs769267565 Chr.1: 245915618-245915618 on Build GRCh38
rs367551555 Chr.1: 245915668-245915668 on Build GRCh38
rs753310553 Chr.1: 245915569-245915569 on Build GRCh38
rs761313433 Chr.1: 245915665-245915665 on Build GRCh38
rs202173734 Chr.1: 245915560-245915560 on Build GRCh38
rs911579163 Chr.1: 245915648-245915648 on Build GRCh38
rs781096043 Chr.1: 245915597-245915597 on Build GRCh38
rs533568761 Chr.1: 245915534-245915534 on Build GRCh38
rs966881550 Chr.1: 245915568-245915568 on Build GRCh38
rs762638036 Chr.1: 245915544-245915544 on Build GRCh38
rs115624121 Chr.1: 245915494-245915494 on Build GRCh38
rs746799686 Chr.1: 245915506-245915506 on Build GRCh38
rs41305945 Chr.1: 245915666-245915666 on Build GRCh38
rs779033365 Chr.1: 245915586-245915586 on Build GRCh38
rs759928395 Chr.1: 245915638-245915638 on Build GRCh38
rs768007152 Chr.1: 245915551-245915551 on Build GRCh38
rs1034902054 Chr.1: 245915539-245915539 on Build GRCh38
rs150058397 Chr.1: 245915550-245915550 on Build GRCh38
rs746753364 Chr.1: 245915589-245915589 on Build GRCh38
rs747600087 Chr.1: 245915525-245915525 on Build GRCh38
rs762396175 Chr.1: 245915487-245915488 on Build GRCh38
rs748154181 Chr.1: 245915617-245915617 on Build GRCh38
rs368134398 Chr.1: 245915495-245915495 on Build GRCh38
rs145023938 Chr.1: 245915595-245915595 on Build GRCh38
rs772382627 Chr.1: 245915642-245915642 on Build GRCh38
rs148947059 Chr.1: 245915630-245915630 on Build GRCh38

View Full Product Details