Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 575922-576157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGQ
Entrez Gene ID
9091
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
Gene Aliases
GPI1, c407A10.1
UniGene
Hs.741878
Gene Chromosome Location
Chr.16: 569968-584136 on Build GRCh38
Transcripts
NM_004204.3 NM_148920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7320185 c.797C>T p.A266V Chr.16: 575946-575946 on Build GRCh38
4952878 c.823A>G p.K275E Chr.16: 576135-576135 on Build GRCh38
7320186 c.797C>T p.A266V Chr.16: 575946-575946 on Build GRCh38
3377922 c.834G>A p.T278T Chr.16: 576146-576146 on Build GRCh38
4631614 c.804C>A p.N268K Chr.16: 575953-575953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375362170 Chr.16: 576113-576113 on Build GRCh38
rs553494816 Chr.16: 576119-576119 on Build GRCh38
rs780448367 Chr.16: 576109-576109 on Build GRCh38
rs747036362 Chr.16: 575953-575953 on Build GRCh38
rs552888936 Chr.16: 576097-576097 on Build GRCh38
rs571151338 Chr.16: 576110-576110 on Build GRCh38
rs923446263 Chr.16: 576133-576133 on Build GRCh38
rs368848682 Chr.16: 576005-576005 on Build GRCh38
rs539479546 Chr.16: 576078-576078 on Build GRCh38
rs900266144 Chr.16: 576114-576114 on Build GRCh38
rs760673037 Chr.16: 576032-576032 on Build GRCh38
rs781235827 Chr.16: 576079-576079 on Build GRCh38
rs777308943 Chr.16: 576007-576007 on Build GRCh38
rs758694346 Chr.16: 576056-576056 on Build GRCh38
rs767564412 Chr.16: 576020-576020 on Build GRCh38
rs865869018 Chr.16: 576010-576010 on Build GRCh38
rs568568536 Chr.16: 576125-576125 on Build GRCh38
rs751908436 Chr.16: 576035-576035 on Build GRCh38
rs946717882 Chr.16: 576034-576034 on Build GRCh38
rs147905228 Chr.16: 575947-575947 on Build GRCh38
rs978014471 Chr.16: 576096-576096 on Build GRCh38
rs867980784 Chr.16: 576044-576044 on Build GRCh38
rs781614768 Chr.16: 575963-575963 on Build GRCh38
rs145382882 Chr.16: 576067-576067 on Build GRCh38
rs976527210 Chr.16: 576123-576123 on Build GRCh38
rs746245143 Chr.16: 575968-575968 on Build GRCh38
rs895239301 Chr.16: 576052-576052 on Build GRCh38
rs753545975 Chr.16: 576041-576041 on Build GRCh38
rs371380018 Chr.16: 576037-576037 on Build GRCh38
rs772843213 Chr.16: 576134-576134 on Build GRCh38
rs1006649526 Chr.16: 576038-576038 on Build GRCh38
rs966972915 Chr.16: 576090-576090 on Build GRCh38
rs528942784 Chr.16: 576028-576028 on Build GRCh38
rs749014095 Chr.16: 575994-575994 on Build GRCh38
rs764171566 Chr.16: 576057-576057 on Build GRCh38
rs199735408 Chr.16: 575973-575973 on Build GRCh38
rs773807126 Chr.16: 576118-576118 on Build GRCh38
rs528254518 Chr.16: 575946-575946 on Build GRCh38
rs761186033 Chr.16: 576006-576006 on Build GRCh38
rs35227916 Chr.16: 575959-575959 on Build GRCh38
rs374974911 Chr.16: 575977-575977 on Build GRCh38
rs1044100051 Chr.16: 576050-576050 on Build GRCh38
rs768173307 Chr.16: 576003-576003 on Build GRCh38
rs372589976 Chr.16: 576100-576100 on Build GRCh38

View Full Product Details