Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 7587182-7587415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBFOX1
Entrez Gene ID
54715
Gene Name
RNA binding protein, fox-1 homolog 1
Gene Aliases
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
UniGene
Hs.459842
Gene Chromosome Location
Chr.16: 5239749-7713343 on Build GRCh38
Transcripts
NM_001142333.1 NM_001142334.1 NM_001308117.1 NM_018723.3 NM_145891.2 NM_145892.2 NM_145893.2 XM_005255386.3 XM_005255387.3 XM_005255388.4 XM_005255390.3 XM_005255391.3 XM_005255394.4 XM_011522546.2 XM_011522547.2 XM_011522548.2 XM_017023318.1 XM_017023319.1 XM_017023320.1 XM_017023321.1 XM_017023322.1 XM_017023323.1 XM_017023324.1 XM_017023325.1 XM_017023326.1 XM_017023327.1 XM_017023328.1 XM_017023329.1 XM_017023330.1 XM_017023331.1 XM_017023332.1 XM_017023333.1 XM_017023334.1 XM_017023335.1 XM_017023336.1 XM_017023337.1 XM_017023338.1 XM_017023339.1 XM_017023340.1 XM_017023341.1 XM_017023342.1 XM_017023343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6145475 c.581A>G p.N194S Chr.16: 7587284-7587284 on Build GRCh38
5515586 c.429C>A p.I143I Chr.16: 7587261-7587261 on Build GRCh38
6412485 c.475C>A p.Q159K Chr.16: 7587247-7587247 on Build GRCh38
5470572 c.597+9C>A p.? Chr.16: 7587309-7587309 on Build GRCh38
6145470 c.520G>A p.G174S Chr.16: 7587292-7587292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760123563 Chr.16: 7587222-7587222 on Build GRCh38
rs766827645 Chr.16: 7587233-7587233 on Build GRCh38
rs778780786 Chr.16: 7587330-7587330 on Build GRCh38
rs781375390 Chr.16: 7587311-7587311 on Build GRCh38
rs949451562 Chr.16: 7587369-7587369 on Build GRCh38
rs763706934 Chr.16: 7587223-7587223 on Build GRCh38
rs745738130 Chr.16: 7587217-7587217 on Build GRCh38
rs527688499 Chr.16: 7587357-7587357 on Build GRCh38
rs771994011 Chr.16: 7587351-7587351 on Build GRCh38
rs35434610 Chr.16: 7587215-7587215 on Build GRCh38
rs116419633 Chr.16: 7587327-7587327 on Build GRCh38
rs947183826 Chr.16: 7587213-7587213 on Build GRCh38
rs375917267 Chr.16: 7587347-7587347 on Build GRCh38
rs1045279266 Chr.16: 7587387-7587387 on Build GRCh38
rs775022018 Chr.16: 7587219-7587219 on Build GRCh38
rs750960761 Chr.16: 7587358-7587358 on Build GRCh38
rs746576201 Chr.16: 7587222-7587226 on Build GRCh38
rs750800481 Chr.16: 7587230-7587230 on Build GRCh38
rs953365504 Chr.16: 7587321-7587321 on Build GRCh38
rs962229700 Chr.16: 7587220-7587220 on Build GRCh38
rs768579064 Chr.16: 7587310-7587311 on Build GRCh38
rs751611395 Chr.16: 7587235-7587235 on Build GRCh38
rs776233193 Chr.16: 7587226-7587226 on Build GRCh38
rs763416978 Chr.16: 7587388-7587388 on Build GRCh38
rs925343353 Chr.16: 7587224-7587224 on Build GRCh38
rs761520199 Chr.16: 7587228-7587228 on Build GRCh38
rs748380046 Chr.16: 7587324-7587324 on Build GRCh38
rs758874318 Chr.16: 7587231-7587231 on Build GRCh38
rs562053699 Chr.16: 7587212-7587212 on Build GRCh38
rs754993274 Chr.16: 7587260-7587260 on Build GRCh38
rs746001735 Chr.16: 7587353-7587353 on Build GRCh38

View Full Product Details