Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 20251544-20251815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUM2
Entrez Gene ID
23369
Gene Name
pumilio RNA binding family member 2
Gene Aliases
PUMH2, PUML2
UniGene
Hs.467824
Gene Chromosome Location
Chr.2: 20248692-20352423 on Build GRCh38
Transcripts
NM_001282752.1 NM_001282790.1 NM_001282791.1 NM_015317.2 XM_005262607.1 XM_005262609.1 XM_005262610.1 XM_006711973.1 XM_006711974.2 XM_006711975.2 XM_011532719.1 XM_011532720.2 XM_011532721.2 XM_011532723.1 XM_017003701.1 XM_017003702.1 XM_017003703.1 XM_017003704.1 XM_017003705.1 XM_017003706.1 XM_017003707.1 XM_017003708.1 XM_017003709.1 XM_017003710.1 XM_017003711.1 XM_017003712.1 XM_017003713.1 XM_017003714.1 XM_017003715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4719475 c.3102C>T p.Y1034Y Chr.2: 20251678-20251678 on Build GRCh38
6010004 c.3150G>A p.K1050K Chr.2: 20251636-20251636 on Build GRCh38
1720196 c.3170G>A p.G1057E Chr.2: 20251610-20251610 on Build GRCh38
1404604 c.3089G>A p.R1030H Chr.2: 20251691-20251691 on Build GRCh38
5829367 c.3095G>A p.R1032H Chr.2: 20251691-20251691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373056312 Chr.2: 20251712-20251712 on Build GRCh38
rs771643233 Chr.2: 20251735-20251735 on Build GRCh38
rs759196214 Chr.2: 20251733-20251733 on Build GRCh38
rs986179946 Chr.2: 20251779-20251779 on Build GRCh38
rs551248991 Chr.2: 20251615-20251615 on Build GRCh38
rs760608492 Chr.2: 20251609-20251609 on Build GRCh38
rs932142911 Chr.2: 20251783-20251783 on Build GRCh38
rs940451183 Chr.2: 20251731-20251731 on Build GRCh38
rs1050646105 Chr.2: 20251785-20251785 on Build GRCh38
rs976576283 Chr.2: 20251716-20251716 on Build GRCh38
rs143577873 Chr.2: 20251628-20251628 on Build GRCh38
rs554815019 Chr.2: 20251637-20251637 on Build GRCh38
rs767318598 Chr.2: 20251607-20251607 on Build GRCh38
rs773682519 Chr.2: 20251724-20251724 on Build GRCh38
rs749741584 Chr.2: 20251704-20251704 on Build GRCh38
rs540817312 Chr.2: 20251681-20251681 on Build GRCh38
rs751840310 Chr.2: 20251670-20251670 on Build GRCh38
rs747228156 Chr.2: 20251748-20251749 on Build GRCh38
rs376088574 Chr.2: 20251765-20251765 on Build GRCh38
rs770281539 Chr.2: 20251691-20251691 on Build GRCh38
rs958440757 Chr.2: 20251599-20251599 on Build GRCh38
rs755202484 Chr.2: 20251678-20251678 on Build GRCh38
rs1040562792 Chr.2: 20251739-20251739 on Build GRCh38
rs778404461 Chr.2: 20251697-20251697 on Build GRCh38
rs72785292 Chr.2: 20251684-20251684 on Build GRCh38
rs764158447 Chr.2: 20251614-20251614 on Build GRCh38
rs780176333 Chr.2: 20251580-20251581 on Build GRCh38
rs977712711 Chr.2: 20251646-20251648 on Build GRCh38
rs932038044 Chr.2: 20251747-20251747 on Build GRCh38
rs191721728 Chr.2: 20251627-20251627 on Build GRCh38
rs965166711 Chr.2: 20251690-20251690 on Build GRCh38
rs779965127 Chr.2: 20251630-20251630 on Build GRCh38
rs775115684 Chr.2: 20251745-20251745 on Build GRCh38
rs765579891 Chr.2: 20251668-20251668 on Build GRCh38

View Full Product Details