Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155295512-155295785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKLR
Entrez Gene ID
5313
Gene Name
pyruvate kinase, liver and RBC
Gene Aliases
PK1, PKL, PKR, PKRL, RPK
UniGene
Hs.95990
Gene Chromosome Location
Chr.1: 155289293-155301434 on Build GRCh38
Transcripts
NM_000298.5 NM_181871.3 XM_006711386.3 XM_011509640.2 XM_017001493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5779319 c.376-3C>A p.? Chr.1: 155295571-155295571 on Build GRCh38
896875 c.399C>T p.N133N Chr.1: 155295545-155295545 on Build GRCh38
305412 c.369C>T p.S123S Chr.1: 155295671-155295671 on Build GRCh38
1334629 c.346C>T p.R116* Chr.1: 155295694-155295694 on Build GRCh38
3976195 c.307C>A p.R103S Chr.1: 155295733-155295733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918627824 Chr.1: 155295709-155295709 on Build GRCh38
rs781353201 Chr.1: 155295605-155295605 on Build GRCh38
rs772979589 Chr.1: 155295573-155295573 on Build GRCh38
rs998374005 Chr.1: 155295571-155295571 on Build GRCh38
rs753658012 Chr.1: 155295582-155295582 on Build GRCh38
rs770802374 Chr.1: 155295648-155295648 on Build GRCh38
rs374177297 Chr.1: 155295599-155295599 on Build GRCh38
rs779517988 Chr.1: 155295699-155295699 on Build GRCh38
rs749817706 Chr.1: 155295545-155295545 on Build GRCh38
rs375544947 Chr.1: 155295652-155295652 on Build GRCh38
rs764337069 Chr.1: 155295686-155295686 on Build GRCh38
rs776268137 Chr.1: 155295740-155295740 on Build GRCh38
rs370958480 Chr.1: 155295650-155295650 on Build GRCh38
rs886045351 Chr.1: 155295551-155295553 on Build GRCh38
rs920274820 Chr.1: 155295749-155295749 on Build GRCh38
rs762937077 Chr.1: 155295752-155295752 on Build GRCh38
rs772680904 Chr.1: 155295659-155295660 on Build GRCh38
rs779765565 Chr.1: 155295733-155295733 on Build GRCh38
rs767598564 Chr.1: 155295666-155295666 on Build GRCh38
rs377371135 Chr.1: 155295626-155295626 on Build GRCh38
rs778268791 Chr.1: 155295627-155295627 on Build GRCh38
rs1033033245 Chr.1: 155295613-155295613 on Build GRCh38
rs754441130 Chr.1: 155295693-155295693 on Build GRCh38
rs752905261 Chr.1: 155295667-155295667 on Build GRCh38
rs775301408 Chr.1: 155295577-155295577 on Build GRCh38
rs754282523 Chr.1: 155295690-155295690 on Build GRCh38
rs765267717 Chr.1: 155295659-155295659 on Build GRCh38
rs372561238 Chr.1: 155295588-155295588 on Build GRCh38
rs770337533 Chr.1: 155295546-155295546 on Build GRCh38
rs770175821 Chr.1: 155295541-155295541 on Build GRCh38
rs762661462 Chr.1: 155295742-155295742 on Build GRCh38
rs200472280 Chr.1: 155295636-155295636 on Build GRCh38
rs1018490282 Chr.1: 155295551-155295553 on Build GRCh38
rs752921668 Chr.1: 155295609-155295609 on Build GRCh38
rs746377052 Chr.1: 155295706-155295706 on Build GRCh38
rs148636159 Chr.1: 155295745-155295745 on Build GRCh38
rs534895384 Chr.1: 155295744-155295744 on Build GRCh38
rs118204089 Chr.1: 155295555-155295555 on Build GRCh38
rs8177971 Chr.1: 155295655-155295655 on Build GRCh38
rs574051756 Chr.1: 155295543-155295543 on Build GRCh38
rs771734912 Chr.1: 155295647-155295647 on Build GRCh38
rs756378131 Chr.1: 155295611-155295611 on Build GRCh38
rs749758002 Chr.1: 155295625-155295625 on Build GRCh38
rs773154027 Chr.1: 155295660-155295660 on Build GRCh38
rs371471086 Chr.1: 155295732-155295732 on Build GRCh38
rs760501654 Chr.1: 155295578-155295578 on Build GRCh38
rs757921751 Chr.1: 155295725-155295725 on Build GRCh38
rs771904185 Chr.1: 155295575-155295575 on Build GRCh38
rs148340437 Chr.1: 155295590-155295590 on Build GRCh38
rs971962553 Chr.1: 155295694-155295694 on Build GRCh38
rs763951356 Chr.1: 155295581-155295581 on Build GRCh38
rs761866493 Chr.1: 155295583-155295583 on Build GRCh38
rs759546000 Chr.1: 155295661-155295661 on Build GRCh38
rs143984223 Chr.1: 155295739-155295739 on Build GRCh38
rs759358280 Chr.1: 155295755-155295755 on Build GRCh38
rs773981429 Chr.1: 155295559-155295559 on Build GRCh38
rs756428691 Chr.1: 155295672-155295672 on Build GRCh38
rs1030651618 Chr.1: 155295577-155295577 on Build GRCh38
rs778259300 Chr.1: 155295618-155295618 on Build GRCh38

View Full Product Details