Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82048768-82049031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFNG
Entrez Gene ID
5986
Gene Name
RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
-
UniGene
Hs.569700
Gene Chromosome Location
Chr.17: 82047902-82051774 on Build GRCh38
Transcripts
NM_002917.1 XM_011523587.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189628 c.949A>G p.T317A Chr.17: 82048773-82048773 on Build GRCh38
4513147 c.928C>T p.H310Y Chr.17: 82048794-82048794 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569956624 Chr.17: 82048822-82048822 on Build GRCh38
rs1030443927 Chr.17: 82048946-82048946 on Build GRCh38
rs770258658 Chr.17: 82048820-82048820 on Build GRCh38
rs577952720 Chr.17: 82048900-82048900 on Build GRCh38
rs745956782 Chr.17: 82049005-82049005 on Build GRCh38
rs567824071 Chr.17: 82049009-82049009 on Build GRCh38
rs78233003 Chr.17: 82049001-82049001 on Build GRCh38
rs769116581 Chr.17: 82048980-82048980 on Build GRCh38
rs868734611 Chr.17: 82048903-82048903 on Build GRCh38
rs751606612 Chr.17: 82049000-82049000 on Build GRCh38
rs990513974 Chr.17: 82048972-82048972 on Build GRCh38
rs201923333 Chr.17: 82048837-82048837 on Build GRCh38
rs372996555 Chr.17: 82048855-82048855 on Build GRCh38
rs962154536 Chr.17: 82048814-82048814 on Build GRCh38
rs1014900528 Chr.17: 82048919-82048919 on Build GRCh38
rs762093760 Chr.17: 82048849-82048879 on Build GRCh38
rs543653992 Chr.17: 82048948-82048948 on Build GRCh38
rs550914623 Chr.17: 82048978-82048978 on Build GRCh38
rs1055867338 Chr.17: 82048886-82048886 on Build GRCh38
rs146534665 Chr.17: 82048907-82048907 on Build GRCh38
rs776953180 Chr.17: 82048817-82048817 on Build GRCh38
rs533861256 Chr.17: 82048949-82048949 on Build GRCh38
rs887984927 Chr.17: 82049008-82049008 on Build GRCh38
rs549333049 Chr.17: 82049002-82049002 on Build GRCh38
rs533533309 Chr.17: 82048961-82048961 on Build GRCh38
rs777143304 Chr.17: 82048831-82048861 on Build GRCh38
rs758174620 Chr.17: 82048861-82048861 on Build GRCh38
rs753586779 Chr.17: 82048834-82048834 on Build GRCh38
rs771239830 Chr.17: 82048797-82048797 on Build GRCh38
rs989838740 Chr.17: 82048867-82048867 on Build GRCh38
rs552116504 Chr.17: 82048879-82048879 on Build GRCh38
rs758363257 Chr.17: 82048838-82048838 on Build GRCh38
rs767487836 Chr.17: 82048990-82048990 on Build GRCh38
rs200202089 Chr.17: 82048825-82048825 on Build GRCh38
rs558082669 Chr.17: 82048909-82048909 on Build GRCh38
rs140019414 Chr.17: 82048994-82048994 on Build GRCh38
rs369573534 Chr.17: 82048849-82048849 on Build GRCh38
rs759174298 Chr.17: 82048992-82048992 on Build GRCh38
rs888042304 Chr.17: 82048897-82048897 on Build GRCh38
rs913143164 Chr.17: 82048976-82048976 on Build GRCh38
rs1024538484 Chr.17: 82048931-82048931 on Build GRCh38
rs371704868 Chr.17: 82048823-82048823 on Build GRCh38
rs778706831 Chr.17: 82048836-82048836 on Build GRCh38
rs945732283 Chr.17: 82048869-82048869 on Build GRCh38
rs1015311117 Chr.17: 82048812-82048812 on Build GRCh38
rs937534900 Chr.17: 82048882-82048882 on Build GRCh38
rs374884083 Chr.17: 82048833-82048833 on Build GRCh38
rs866851454 Chr.17: 82048885-82048885 on Build GRCh38
rs775874966 Chr.17: 82048858-82048858 on Build GRCh38
rs778770186 Chr.17: 82048854-82048854 on Build GRCh38
rs759804427 Chr.17: 82048818-82048818 on Build GRCh38
rs559121215 Chr.17: 82048967-82048967 on Build GRCh38
rs763334584 Chr.17: 82048859-82048859 on Build GRCh38
rs765983830 Chr.17: 82048989-82048989 on Build GRCh38
rs771186696 Chr.17: 82048844-82048844 on Build GRCh38
rs541098958 Chr.17: 82048962-82048962 on Build GRCh38
rs769921589 Chr.17: 82048856-82048856 on Build GRCh38
rs1045925774 Chr.17: 82048875-82048875 on Build GRCh38
rs867836835 Chr.17: 82048922-82048922 on Build GRCh38
rs894506960 Chr.17: 82048914-82048914 on Build GRCh38
rs910878388 Chr.17: 82048965-82048965 on Build GRCh38
rs985302082 Chr.17: 82048952-82048952 on Build GRCh38
rs756316120 Chr.17: 82049006-82049006 on Build GRCh38
rs781107605 Chr.17: 82048826-82048826 on Build GRCh38
rs539782564 Chr.17: 82048906-82048906 on Build GRCh38
rs1027827367 Chr.17: 82048951-82048951 on Build GRCh38
rs755250241 Chr.17: 82048788-82048788 on Build GRCh38
rs535087847 Chr.17: 82048863-82048863 on Build GRCh38
rs553117599 Chr.17: 82048868-82048868 on Build GRCh38
rs201190708 Chr.17: 82049011-82049011 on Build GRCh38
rs532758119 Chr.17: 82048968-82048968 on Build GRCh38
rs774884692 Chr.17: 82048985-82048985 on Build GRCh38
rs200040630 Chr.17: 82048828-82048828 on Build GRCh38
rs8069068 Chr.17: 82048936-82048936 on Build GRCh38
rs906950885 Chr.17: 82048877-82048877 on Build GRCh38
rs764102847 Chr.17: 82048998-82048998 on Build GRCh38

View Full Product Details