Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47784881-47785146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP160
Entrez Gene ID
23279
Gene Name
nucleoporin 160
Gene Aliases
-
UniGene
Hs.643526
Gene Chromosome Location
Chr.11: 47778083-47848544 on Build GRCh38
Transcripts
NM_015231.2 NR_134636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1236098 c.3867G>A p.W1289* Chr.11: 47785045-47785045 on Build GRCh38
4033253 c.3975T>C p.L1325L Chr.11: 47784937-47784937 on Build GRCh38
6731461 c.3852T>C p.A1284A Chr.11: 47785060-47785060 on Build GRCh38
415590 c.3953G>C p.G1318A Chr.11: 47784959-47784959 on Build GRCh38
6868154 c.3983G>A p.S1328N Chr.11: 47784929-47784929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758095287 Chr.11: 47784963-47784963 on Build GRCh38
rs748389104 Chr.11: 47785067-47785068 on Build GRCh38
rs754350286 Chr.11: 47785103-47785104 on Build GRCh38
rs767001109 Chr.11: 47785109-47785109 on Build GRCh38
rs1055426340 Chr.11: 47785038-47785038 on Build GRCh38
rs752234613 Chr.11: 47785111-47785111 on Build GRCh38
rs747178084 Chr.11: 47785057-47785057 on Build GRCh38
rs200105376 Chr.11: 47784916-47784916 on Build GRCh38
rs751230133 Chr.11: 47785107-47785107 on Build GRCh38
rs978560129 Chr.11: 47785012-47785012 on Build GRCh38
rs773490396 Chr.11: 47785097-47785097 on Build GRCh38
rs936515260 Chr.11: 47784956-47784956 on Build GRCh38
rs373842346 Chr.11: 47784954-47784954 on Build GRCh38
rs190828883 Chr.11: 47785100-47785100 on Build GRCh38
rs186108278 Chr.11: 47785043-47785043 on Build GRCh38
rs770313573 Chr.11: 47785091-47785091 on Build GRCh38
rs1044873153 Chr.11: 47784918-47784918 on Build GRCh38
rs761480915 Chr.11: 47785104-47785109 on Build GRCh38
rs1018187095 Chr.11: 47784923-47784923 on Build GRCh38
rs371709395 Chr.11: 47785090-47785090 on Build GRCh38
rs758629949 Chr.11: 47785107-47785109 on Build GRCh38
rs768836811 Chr.11: 47785033-47785033 on Build GRCh38
rs767023066 Chr.11: 47785104-47785105 on Build GRCh38
rs763781654 Chr.11: 47785101-47785102 on Build GRCh38
rs768868483 Chr.11: 47785073-47785073 on Build GRCh38
rs778039579 Chr.11: 47785106-47785107 on Build GRCh38
rs772240034 Chr.11: 47785068-47785068 on Build GRCh38
rs762700858 Chr.11: 47785102-47785108 on Build GRCh38
rs765589083 Chr.11: 47784945-47784945 on Build GRCh38
rs768566049 Chr.11: 47785115-47785118 on Build GRCh38
rs146411765 Chr.11: 47784925-47784925 on Build GRCh38
rs772406263 Chr.11: 47784912-47784912 on Build GRCh38
rs777483075 Chr.11: 47785034-47785034 on Build GRCh38
rs901884459 Chr.11: 47785075-47785075 on Build GRCh38
rs749153781 Chr.11: 47785115-47785150 on Build GRCh38
rs996120280 Chr.11: 47785112-47785112 on Build GRCh38
rs780697313 Chr.11: 47785007-47785007 on Build GRCh38
rs759124767 Chr.11: 47785108-47785108 on Build GRCh38
rs202140901 Chr.11: 47784943-47784943 on Build GRCh38
rs201748590 Chr.11: 47784972-47784972 on Build GRCh38
rs777830119 Chr.11: 47785098-47785099 on Build GRCh38
rs751177088 Chr.11: 47784986-47784986 on Build GRCh38
rs780453055 Chr.11: 47785056-47785056 on Build GRCh38
rs769707637 Chr.11: 47785115-47785136 on Build GRCh38
rs754509063 Chr.11: 47784999-47784999 on Build GRCh38
rs765988996 Chr.11: 47784984-47784984 on Build GRCh38
rs554894200 Chr.11: 47785105-47785105 on Build GRCh38
rs748038025 Chr.11: 47785015-47785015 on Build GRCh38
rs945806123 Chr.11: 47784973-47784973 on Build GRCh38
rs369448926 Chr.11: 47785084-47785084 on Build GRCh38
rs756052407 Chr.11: 47785115-47785115 on Build GRCh38
rs201258904 Chr.11: 47785088-47785088 on Build GRCh38
rs775438638 Chr.11: 47785115-47785123 on Build GRCh38
rs1019367232 Chr.11: 47785065-47785065 on Build GRCh38
rs148288371 Chr.11: 47785031-47785031 on Build GRCh38
rs775316788 Chr.11: 47785101-47785149 on Build GRCh38
rs756070737 Chr.11: 47785018-47785018 on Build GRCh38
rs538421016 Chr.11: 47785104-47785104 on Build GRCh38
rs371080808 Chr.11: 47784915-47784915 on Build GRCh38

View Full Product Details