Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153991469-153991726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS27
Entrez Gene ID
6232
Gene Name
ribosomal protein S27
Gene Aliases
MPS-1, MPS1, S27
UniGene
Hs.546291
Gene Chromosome Location
Chr.1: 153990763-153992155 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7262950 c.127A>T p.I43F Chr.1: 153991577-153991577 on Build GRCh38
7324267 c.184G>A p.V62I Chr.1: 153991634-153991634 on Build GRCh38
2208466 c.156G>A p.T52T Chr.1: 153991606-153991606 on Build GRCh38
896486 c.130A>G p.T44A Chr.1: 153991580-153991580 on Build GRCh38
6346259 c.131C>A p.T44N Chr.1: 153991581-153991581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147042648 Chr.1: 153991540-153991540 on Build GRCh38
rs758495666 Chr.1: 153991596-153991596 on Build GRCh38
rs767972625 Chr.1: 153991554-153991554 on Build GRCh38
rs372202101 Chr.1: 153991588-153991588 on Build GRCh38
rs900800386 Chr.1: 153991525-153991525 on Build GRCh38
rs757498323 Chr.1: 153991585-153991585 on Build GRCh38
rs35506293 Chr.1: 153991520-153991521 on Build GRCh38
rs1006565022 Chr.1: 153991672-153991672 on Build GRCh38
rs892368669 Chr.1: 153991570-153991570 on Build GRCh38
rs777914816 Chr.1: 153991521-153991521 on Build GRCh38
rs2633 Chr.1: 153991514-153991514 on Build GRCh38
rs753234083 Chr.1: 153991559-153991559 on Build GRCh38
rs1050891984 Chr.1: 153991581-153991581 on Build GRCh38
rs770500545 Chr.1: 153991636-153991636 on Build GRCh38
rs769511722 Chr.1: 153991621-153991621 on Build GRCh38
rs769613499 Chr.1: 153991542-153991542 on Build GRCh38
rs371349094 Chr.1: 153991558-153991558 on Build GRCh38
rs114120114 Chr.1: 153991507-153991507 on Build GRCh38
rs772783073 Chr.1: 153991544-153991544 on Build GRCh38
rs748951108 Chr.1: 153991633-153991633 on Build GRCh38
rs367550840 Chr.1: 153991550-153991550 on Build GRCh38
rs748090283 Chr.1: 153991619-153991619 on Build GRCh38
rs775899772 Chr.1: 153991688-153991688 on Build GRCh38
rs753564465 Chr.1: 153991677-153991678 on Build GRCh38
rs549879404 Chr.1: 153991639-153991639 on Build GRCh38
rs760724908 Chr.1: 153991521-153991522 on Build GRCh38
rs909804068 Chr.1: 153991513-153991513 on Build GRCh38
rs772283808 Chr.1: 153991551-153991551 on Build GRCh38
rs998131135 Chr.1: 153991536-153991536 on Build GRCh38
rs1039395581 Chr.1: 153991524-153991524 on Build GRCh38
rs942420547 Chr.1: 153991511-153991511 on Build GRCh38
rs759104303 Chr.1: 153991659-153991659 on Build GRCh38
rs2632 Chr.1: 153991545-153991545 on Build GRCh38
rs397713349 Chr.1: 153991522-153991523 on Build GRCh38
rs375509510 Chr.1: 153991694-153991694 on Build GRCh38
rs768647872 Chr.1: 153991534-153991534 on Build GRCh38
rs772939307 Chr.1: 153991632-153991632 on Build GRCh38
rs140257179 Chr.1: 153991519-153991520 on Build GRCh38
rs764487569 Chr.1: 153991563-153991563 on Build GRCh38
rs760036479 Chr.1: 153991553-153991553 on Build GRCh38
rs763952144 Chr.1: 153991559-153991559 on Build GRCh38
rs749497079 Chr.1: 153991528-153991528 on Build GRCh38
rs779087764 Chr.1: 153991530-153991530 on Build GRCh38
rs767031397 Chr.1: 153991675-153991675 on Build GRCh38
rs746959195 Chr.1: 153991532-153991532 on Build GRCh38
rs781044293 Chr.1: 153991607-153991607 on Build GRCh38
rs567792813 Chr.1: 153991689-153991689 on Build GRCh38
rs762555280 Chr.1: 153991548-153991548 on Build GRCh38
rs150298203 Chr.1: 153991591-153991591 on Build GRCh38

View Full Product Details