Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43008186-43008421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEA1
Entrez Gene ID
4201
Gene Name
male-enhanced antigen 1
Gene Aliases
HYS, MEA
UniGene
Hs.278362
Gene Chromosome Location
Chr.6: 42980344-43016886 on Build GRCh38
Transcripts
XM_017010868.1 NM_001270476.1 NM_006245.3 NM_180976.2 NM_180977.2

Gene Symbol
PPP2R5D
Entrez Gene ID
5528
Gene Name
protein phosphatase 2 regulatory subunit B'delta
Gene Aliases
B56D, MRD35
UniGene
Hs.533308
Gene Chromosome Location
Chr.6: 42984499-43012345 on Build GRCh38
Transcripts
XM_017010868.1 NM_001270476.1 NM_006245.3 NM_180976.2 NM_180977.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586767 c.930C>T p.G310G Chr.6: 43008379-43008379 on Build GRCh38
5746582 c.920T>A p.I307N Chr.6: 43008369-43008369 on Build GRCh38
6887187 c.921C>T p.I307I Chr.6: 43008370-43008370 on Build GRCh38
4161204 c.866A>C p.Y289S Chr.6: 43008209-43008209 on Build GRCh38
3874165 c.858-2A>G p.? Chr.6: 43008199-43008199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775123223 Chr.6: 43008225-43008225 on Build GRCh38
rs768102492 Chr.6: 43008228-43008228 on Build GRCh38
rs781052890 Chr.6: 43008306-43008306 on Build GRCh38
rs765413854 Chr.6: 43008391-43008391 on Build GRCh38
rs982726510 Chr.6: 43008350-43008350 on Build GRCh38
rs752770370 Chr.6: 43008275-43008275 on Build GRCh38
rs777042845 Chr.6: 43008379-43008379 on Build GRCh38
rs199597619 Chr.6: 43008269-43008269 on Build GRCh38
rs768307179 Chr.6: 43008325-43008325 on Build GRCh38
rs762616717 Chr.6: 43008227-43008227 on Build GRCh38
rs746980244 Chr.6: 43008296-43008296 on Build GRCh38
rs866594047 Chr.6: 43008386-43008386 on Build GRCh38
rs770562697 Chr.6: 43008297-43008297 on Build GRCh38
rs370250232 Chr.6: 43008270-43008270 on Build GRCh38
rs1019801370 Chr.6: 43008313-43008313 on Build GRCh38
rs531787162 Chr.6: 43008388-43008388 on Build GRCh38
rs769480815 Chr.6: 43008311-43008311 on Build GRCh38
rs899469077 Chr.6: 43008332-43008332 on Build GRCh38
rs145452516 Chr.6: 43008216-43008216 on Build GRCh38
rs761251273 Chr.6: 43008349-43008349 on Build GRCh38
rs372472214 Chr.6: 43008276-43008276 on Build GRCh38
rs999616194 Chr.6: 43008237-43008237 on Build GRCh38
rs374394486 Chr.6: 43008318-43008318 on Build GRCh38
rs745704580 Chr.6: 43008310-43008310 on Build GRCh38
rs755243916 Chr.6: 43008271-43008271 on Build GRCh38
rs766597810 Chr.6: 43008362-43008362 on Build GRCh38
rs368393024 Chr.6: 43008323-43008323 on Build GRCh38
rs751013445 Chr.6: 43008268-43008268 on Build GRCh38
rs748782415 Chr.6: 43008324-43008324 on Build GRCh38
rs777637869 Chr.6: 43008288-43008288 on Build GRCh38

View Full Product Details