Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119335853-119336116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF26
Entrez Gene ID
79102
Gene Name
ring finger protein 26
Gene Aliases
-
UniGene
Hs.524084
Gene Chromosome Location
Chr.11: 119334500-119337314 on Build GRCh38
Transcripts
NM_032015.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6838133 c.841G>C p.A281P Chr.11: 119335963-119335963 on Build GRCh38
3868564 c.971G>A p.R324K Chr.11: 119336093-119336093 on Build GRCh38
923836 c.894G>A p.A298A Chr.11: 119336016-119336016 on Build GRCh38
6996261 c.865C>T p.R289* Chr.11: 119335987-119335987 on Build GRCh38
6448702 c.866G>A p.R289Q Chr.11: 119335988-119335988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751974458 Chr.11: 119336048-119336048 on Build GRCh38
rs35503635 Chr.11: 119335914-119335914 on Build GRCh38
rs749925094 Chr.11: 119336039-119336039 on Build GRCh38
rs375540920 Chr.11: 119335912-119335912 on Build GRCh38
rs759453904 Chr.11: 119335948-119335948 on Build GRCh38
rs749407842 Chr.11: 119336020-119336020 on Build GRCh38
rs992580092 Chr.11: 119335934-119335934 on Build GRCh38
rs778509045 Chr.11: 119335972-119335972 on Build GRCh38
rs752932597 Chr.11: 119335929-119335929 on Build GRCh38
rs202120371 Chr.11: 119336090-119336090 on Build GRCh38
rs767506805 Chr.11: 119335951-119335951 on Build GRCh38
rs777328293 Chr.11: 119335893-119335893 on Build GRCh38
rs755183725 Chr.11: 119335903-119335904 on Build GRCh38
rs866047176 Chr.11: 119336022-119336022 on Build GRCh38
rs564974788 Chr.11: 119335913-119335913 on Build GRCh38
rs202238538 Chr.11: 119336029-119336029 on Build GRCh38
rs773272418 Chr.11: 119336000-119336000 on Build GRCh38
rs778016899 Chr.11: 119336070-119336070 on Build GRCh38
rs867982099 Chr.11: 119336078-119336078 on Build GRCh38
rs904616153 Chr.11: 119336085-119336085 on Build GRCh38
rs548319846 Chr.11: 119335899-119335899 on Build GRCh38
rs150176910 Chr.11: 119335957-119335957 on Build GRCh38
rs989949038 Chr.11: 119336016-119336016 on Build GRCh38
rs761546321 Chr.11: 119336033-119336033 on Build GRCh38
rs200106607 Chr.11: 119336035-119336035 on Build GRCh38
rs781394481 Chr.11: 119336066-119336066 on Build GRCh38
rs776499628 Chr.11: 119336032-119336032 on Build GRCh38
rs948588585 Chr.11: 119335964-119335964 on Build GRCh38
rs780469665 Chr.11: 119335985-119335985 on Build GRCh38
rs754542537 Chr.11: 119335963-119335963 on Build GRCh38
rs748309223 Chr.11: 119335992-119335992 on Build GRCh38
rs141362166 Chr.11: 119335980-119335980 on Build GRCh38
rs752504351 Chr.11: 119335953-119335953 on Build GRCh38
rs758034366 Chr.11: 119336042-119336042 on Build GRCh38
rs981345539 Chr.11: 119336052-119336052 on Build GRCh38
rs755518635 Chr.11: 119336061-119336061 on Build GRCh38
rs756432249 Chr.11: 119336089-119336089 on Build GRCh38
rs765940789 Chr.11: 119336043-119336043 on Build GRCh38
rs755785802 Chr.11: 119335891-119335891 on Build GRCh38
rs770055202 Chr.11: 119335999-119335999 on Build GRCh38
rs753366689 Chr.11: 119335900-119335900 on Build GRCh38
rs1037118881 Chr.11: 119335987-119335987 on Build GRCh38
rs776771535 Chr.11: 119335927-119335927 on Build GRCh38
rs760434486 Chr.11: 119335956-119335956 on Build GRCh38
rs146952991 Chr.11: 119335986-119335986 on Build GRCh38
rs889281790 Chr.11: 119336051-119336051 on Build GRCh38
rs370172683 Chr.11: 119335933-119335933 on Build GRCh38
rs143529789 Chr.11: 119336050-119336050 on Build GRCh38
rs151036748 Chr.11: 119335882-119335882 on Build GRCh38
rs752489469 Chr.11: 119335887-119335887 on Build GRCh38
rs145726404 Chr.11: 119336096-119336096 on Build GRCh38
rs145563235 Chr.11: 119335971-119335971 on Build GRCh38
rs746864647 Chr.11: 119336093-119336093 on Build GRCh38
rs771972535 Chr.11: 119336028-119336028 on Build GRCh38
rs768894128 Chr.11: 119335922-119335922 on Build GRCh38
rs373389286 Chr.11: 119335984-119335984 on Build GRCh38
rs753391065 Chr.11: 119335958-119335958 on Build GRCh38
rs137974898 Chr.11: 119335988-119335988 on Build GRCh38
rs748575385 Chr.11: 119336069-119336069 on Build GRCh38
rs747402374 Chr.11: 119335920-119335920 on Build GRCh38
rs144186824 Chr.11: 119335877-119335877 on Build GRCh38
rs144743618 Chr.11: 119336030-119336030 on Build GRCh38
rs769795405 Chr.11: 119335928-119335928 on Build GRCh38
rs749532471 Chr.11: 119336083-119336083 on Build GRCh38

View Full Product Details