Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 123401548-123401821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRY1
Entrez Gene ID
10252
Gene Name
sprouty RTK signaling antagonist 1
Gene Aliases
hSPRY1
UniGene
Hs.744854
Gene Chromosome Location
Chr.4: 123396791-123403760 on Build GRCh38
Transcripts
NM_001258038.1 NM_001258039.1 NM_005841.2 NM_199327.2 XM_005262686.1 XM_017007635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227435 c.197G>A p.R66Q Chr.4: 123401788-123401788 on Build GRCh38
1050764 c.35C>T p.S12L Chr.4: 123401626-123401626 on Build GRCh38
6256676 c.63G>T p.L21F Chr.4: 123401654-123401654 on Build GRCh38
6214659 c.1A>G p.M1V Chr.4: 123401592-123401592 on Build GRCh38
3009030 c.193C>T p.P65S Chr.4: 123401784-123401784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769362140 Chr.4: 123401703-123401703 on Build GRCh38
rs759365951 Chr.4: 123401730-123401730 on Build GRCh38
rs115001600 Chr.4: 123401609-123401609 on Build GRCh38
rs772531944 Chr.4: 123401704-123401704 on Build GRCh38
rs748168416 Chr.4: 123401589-123401589 on Build GRCh38
rs781253997 Chr.4: 123401588-123401588 on Build GRCh38
rs748601164 Chr.4: 123401586-123401586 on Build GRCh38
rs112240508 Chr.4: 123401680-123401680 on Build GRCh38
rs766226331 Chr.4: 123401649-123401649 on Build GRCh38
rs776404987 Chr.4: 123401636-123401636 on Build GRCh38
rs764619025 Chr.4: 123401639-123401639 on Build GRCh38
rs770444253 Chr.4: 123401784-123401784 on Build GRCh38
rs746288824 Chr.4: 123401686-123401686 on Build GRCh38
rs757800382 Chr.4: 123401745-123401745 on Build GRCh38
rs778412478 Chr.4: 123401605-123401605 on Build GRCh38
rs149918227 Chr.4: 123401774-123401774 on Build GRCh38
rs905885817 Chr.4: 123401610-123401610 on Build GRCh38
rs1032412417 Chr.4: 123401688-123401688 on Build GRCh38
rs772538327 Chr.4: 123401691-123401691 on Build GRCh38
rs537774102 Chr.4: 123401734-123401734 on Build GRCh38
rs115106991 Chr.4: 123401700-123401700 on Build GRCh38
rs776496321 Chr.4: 123401692-123401692 on Build GRCh38
rs751828074 Chr.4: 123401572-123401572 on Build GRCh38
rs779689163 Chr.4: 123401569-123401569 on Build GRCh38
rs114172721 Chr.4: 123401744-123401744 on Build GRCh38
rs745654554 Chr.4: 123401787-123401787 on Build GRCh38
rs747809146 Chr.4: 123401769-123401769 on Build GRCh38
rs762749150 Chr.4: 123401648-123401648 on Build GRCh38
rs780510324 Chr.4: 123401766-123401766 on Build GRCh38
rs979389180 Chr.4: 123401679-123401679 on Build GRCh38
rs112734942 Chr.4: 123401602-123401602 on Build GRCh38
rs774339078 Chr.4: 123401786-123401786 on Build GRCh38
rs942674208 Chr.4: 123401713-123401713 on Build GRCh38
rs1012523568 Chr.4: 123401644-123401644 on Build GRCh38
rs746376752 Chr.4: 123401764-123401764 on Build GRCh38
rs141161959 Chr.4: 123401677-123401677 on Build GRCh38
rs764236200 Chr.4: 123401743-123401743 on Build GRCh38
rs775197149 Chr.4: 123401792-123401792 on Build GRCh38
rs758894580 Chr.4: 123401765-123401765 on Build GRCh38
rs971313977 Chr.4: 123401646-123401646 on Build GRCh38
rs760438293 Chr.4: 123401620-123401620 on Build GRCh38
rs772802512 Chr.4: 123401778-123401778 on Build GRCh38
rs201409774 Chr.4: 123401662-123401662 on Build GRCh38
rs749729525 Chr.4: 123401607-123401607 on Build GRCh38
rs755152264 Chr.4: 123401581-123401581 on Build GRCh38
rs200501686 Chr.4: 123401715-123401715 on Build GRCh38
rs557398664 Chr.4: 123401741-123401741 on Build GRCh38
rs546122038 Chr.4: 123401753-123401753 on Build GRCh38
rs371347844 Chr.4: 123401660-123401660 on Build GRCh38
rs147613282 Chr.4: 123401779-123401779 on Build GRCh38
rs769852534 Chr.4: 123401590-123401590 on Build GRCh38
rs752972858 Chr.4: 123401736-123401736 on Build GRCh38
rs756230838 Chr.4: 123401590-123401591 on Build GRCh38
rs140546097 Chr.4: 123401788-123401788 on Build GRCh38
rs376428204 Chr.4: 123401727-123401727 on Build GRCh38
rs146763160 Chr.4: 123401685-123401685 on Build GRCh38
rs761473092 Chr.4: 123401638-123401638 on Build GRCh38
rs774249491 Chr.4: 123401729-123401729 on Build GRCh38
rs756227593 Chr.4: 123401668-123401668 on Build GRCh38
rs774533854 Chr.4: 123401611-123401611 on Build GRCh38
rs757634424 Chr.4: 123401683-123401683 on Build GRCh38
rs1020791706 Chr.4: 123401671-123401671 on Build GRCh38
rs181355842 Chr.4: 123401754-123401754 on Build GRCh38
rs535040315 Chr.4: 123401626-123401626 on Build GRCh38
rs369054680 Chr.4: 123401645-123401645 on Build GRCh38
rs898813706 Chr.4: 123401627-123401627 on Build GRCh38
rs755205655 Chr.4: 123401661-123401661 on Build GRCh38
rs201890938 Chr.4: 123401723-123401723 on Build GRCh38
rs1035810825 Chr.4: 123401657-123401657 on Build GRCh38
rs760985314 Chr.4: 123401796-123401796 on Build GRCh38
rs940446674 Chr.4: 123401793-123401793 on Build GRCh38
rs374778958 Chr.4: 123401578-123401578 on Build GRCh38

View Full Product Details