Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 177336165-177336390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEIL3
Entrez Gene ID
55247
Gene Name
nei like DNA glycosylase 3
Gene Aliases
FGP2, FPG2, NEI3, ZGRF3, hFPG2, hNEI3
UniGene
Hs.405467
Gene Chromosome Location
Chr.4: 177309837-177362938 on Build GRCh38
Transcripts
NM_018248.2 XM_017008360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6099848 c.572G>T p.G191V Chr.4: 177336266-177336266 on Build GRCh38
5518562 c.495A>G p.A165A Chr.4: 177336189-177336189 on Build GRCh38
6724359 c.583delA p.N196fs*18 Chr.4: 177336277-177336277 on Build GRCh38
4997635 c.535G>A p.D179N Chr.4: 177336229-177336229 on Build GRCh38
3674141 c.617C>A p.P206Q Chr.4: 177336311-177336311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149191538 Chr.4: 177336301-177336301 on Build GRCh38
rs1023250712 Chr.4: 177336202-177336202 on Build GRCh38
rs975138442 Chr.4: 177336311-177336311 on Build GRCh38
rs142427084 Chr.4: 177336313-177336313 on Build GRCh38
rs35455003 Chr.4: 177336334-177336334 on Build GRCh38
rs534859801 Chr.4: 177336298-177336298 on Build GRCh38
rs754521126 Chr.4: 177336217-177336217 on Build GRCh38
rs767793726 Chr.4: 177336220-177336220 on Build GRCh38
rs201257637 Chr.4: 177336218-177336218 on Build GRCh38
rs761968037 Chr.4: 177336194-177336194 on Build GRCh38
rs367778438 Chr.4: 177336293-177336293 on Build GRCh38
rs201390371 Chr.4: 177336361-177336361 on Build GRCh38
rs765433921 Chr.4: 177336198-177336198 on Build GRCh38
rs766585379 Chr.4: 177336276-177336276 on Build GRCh38
rs995579729 Chr.4: 177336201-177336201 on Build GRCh38
rs755893929 Chr.4: 177336222-177336222 on Build GRCh38
rs770578974 Chr.4: 177336233-177336233 on Build GRCh38
rs372830444 Chr.4: 177336257-177336257 on Build GRCh38
rs777534860 Chr.4: 177336229-177336229 on Build GRCh38
rs774380757 Chr.4: 177336241-177336241 on Build GRCh38
rs565542222 Chr.4: 177336216-177336216 on Build GRCh38
rs749139541 Chr.4: 177336232-177336232 on Build GRCh38
rs145273162 Chr.4: 177336250-177336250 on Build GRCh38
rs771970862 Chr.4: 177336337-177336337 on Build GRCh38
rs150006122 Chr.4: 177336249-177336249 on Build GRCh38
rs530222180 Chr.4: 177336347-177336347 on Build GRCh38
rs752120186 Chr.4: 177336282-177336282 on Build GRCh38
rs527920923 Chr.4: 177336225-177336225 on Build GRCh38
rs775375636 Chr.4: 177336342-177336342 on Build GRCh38
rs765485707 Chr.4: 177336261-177336261 on Build GRCh38
rs761878205 Chr.4: 177336258-177336258 on Build GRCh38
rs2271102 Chr.4: 177336362-177336362 on Build GRCh38
rs763398465 Chr.4: 177336271-177336271 on Build GRCh38
rs941793688 Chr.4: 177336351-177336351 on Build GRCh38
rs116410768 Chr.4: 177336289-177336289 on Build GRCh38
rs778518433 Chr.4: 177336332-177336332 on Build GRCh38
rs755658378 Chr.4: 177336297-177336297 on Build GRCh38
rs758660043 Chr.4: 177336215-177336215 on Build GRCh38
rs771171873 Chr.4: 177336368-177336368 on Build GRCh38
rs897692951 Chr.4: 177336363-177336363 on Build GRCh38
rs763026175 Chr.4: 177336365-177336365 on Build GRCh38
rs17064658 Chr.4: 177336210-177336210 on Build GRCh38
rs752651163 Chr.4: 177336266-177336266 on Build GRCh38

View Full Product Details