Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64769243-64769478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SF1
Entrez Gene ID
7536
Gene Name
splicing factor 1
Gene Aliases
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162
UniGene
Hs.502829
Gene Chromosome Location
Chr.11: 64764604-64779043 on Build GRCh38
Transcripts
NM_001178030.1 NM_001178031.1 NM_004630.3 NM_201995.2 NM_201997.2 NM_201998.2 NR_033649.1 NR_033650.1 XM_011545244.1 XM_011545245.1 XM_011545246.1 XM_011545247.1 XM_011545248.1 XM_017018244.1 XM_017018245.1 XM_017018246.1 XM_017018247.1 XM_017018248.1 XM_017018249.1 XM_017018250.1 XM_017018251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7416701 c.743C>T p.A248V Chr.11: 64769259-64769259 on Build GRCh38
6069919 c.742G>A p.A248T Chr.11: 64769260-64769260 on Build GRCh38
544454 c.718C>T p.R240W Chr.11: 64769284-64769284 on Build GRCh38
1676155 c.682C>T p.Q228* Chr.11: 64769320-64769320 on Build GRCh38
6054412 c.700G>T p.E234* Chr.11: 64769302-64769302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs484886 Chr.11: 64769394-64769394 on Build GRCh38
rs377713077 Chr.11: 64769406-64769406 on Build GRCh38
rs764352170 Chr.11: 64769357-64769358 on Build GRCh38
rs776368614 Chr.11: 64769360-64769360 on Build GRCh38
rs759244399 Chr.11: 64769398-64769398 on Build GRCh38
rs757136423 Chr.11: 64769418-64769418 on Build GRCh38
rs775643647 Chr.11: 64769366-64769366 on Build GRCh38
rs188599860 Chr.11: 64769297-64769297 on Build GRCh38
rs906490473 Chr.11: 64769444-64769444 on Build GRCh38
rs777682941 Chr.11: 64769344-64769344 on Build GRCh38
rs774974011 Chr.11: 64769391-64769391 on Build GRCh38
rs111552365 Chr.11: 64769269-64769269 on Build GRCh38
rs143262056 Chr.11: 64769438-64769438 on Build GRCh38
rs770692604 Chr.11: 64769354-64769354 on Build GRCh38
rs372747905 Chr.11: 64769353-64769353 on Build GRCh38
rs551156250 Chr.11: 64769327-64769327 on Build GRCh38
rs757834421 Chr.11: 64769409-64769409 on Build GRCh38
rs373299928 Chr.11: 64769400-64769400 on Build GRCh38
rs764751023 Chr.11: 64769384-64769384 on Build GRCh38
rs752817802 Chr.11: 64769374-64769374 on Build GRCh38
rs561102220 Chr.11: 64769399-64769399 on Build GRCh38
rs750771343 Chr.11: 64769397-64769397 on Build GRCh38
rs369419135 Chr.11: 64769326-64769326 on Build GRCh38
rs371348522 Chr.11: 64769416-64769416 on Build GRCh38
rs148293463 Chr.11: 64769456-64769456 on Build GRCh38
rs150432446 Chr.11: 64769312-64769312 on Build GRCh38
rs1002073728 Chr.11: 64769279-64769279 on Build GRCh38
rs148875917 Chr.11: 64769420-64769420 on Build GRCh38
rs751745205 Chr.11: 64769394-64769396 on Build GRCh38
rs766180684 Chr.11: 64769273-64769273 on Build GRCh38
rs758362181 Chr.11: 64769341-64769341 on Build GRCh38
rs749430585 Chr.11: 64769447-64769447 on Build GRCh38
rs776101875 Chr.11: 64769446-64769446 on Build GRCh38
rs71583707 Chr.11: 64769361-64769361 on Build GRCh38
rs986300843 Chr.11: 64769370-64769370 on Build GRCh38
rs762306570 Chr.11: 64769393-64769393 on Build GRCh38
rs530490514 Chr.11: 64769324-64769324 on Build GRCh38
rs757256486 Chr.11: 64769401-64769403 on Build GRCh38
rs769850081 Chr.11: 64769365-64769365 on Build GRCh38
rs759657783 Chr.11: 64769330-64769330 on Build GRCh38
rs368385290 Chr.11: 64769417-64769417 on Build GRCh38
rs746866349 Chr.11: 64769350-64769350 on Build GRCh38
rs780977526 Chr.11: 64769419-64769419 on Build GRCh38

View Full Product Details