Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 39012739-39012984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER1
Entrez Gene ID
80209
Gene Name
proline and serine rich 1
Gene Aliases
C13orf23
UniGene
Hs.318526
Gene Chromosome Location
Chr.13: 39009865-39038115 on Build GRCh38
Transcripts
NM_025138.4 XM_005266545.3 XM_011535239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3384839 c.2394C>T p.S798S Chr.13: 39012858-39012858 on Build GRCh38
5042099 c.2278G>A p.A760T Chr.13: 39012974-39012974 on Build GRCh38
4963444 c.2450C>T p.A817V Chr.13: 39012802-39012802 on Build GRCh38
947319 c.2420C>T p.P807L Chr.13: 39012832-39012832 on Build GRCh38
4652395 c.2433G>A p.A811A Chr.13: 39012819-39012819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761563804 Chr.13: 39012788-39012788 on Build GRCh38
rs368546956 Chr.13: 39012831-39012831 on Build GRCh38
rs770081587 Chr.13: 39012839-39012839 on Build GRCh38
rs754985997 Chr.13: 39012860-39012860 on Build GRCh38
rs8181819 Chr.13: 39012917-39012917 on Build GRCh38
rs200475947 Chr.13: 39012769-39012769 on Build GRCh38
rs756436560 Chr.13: 39012776-39012776 on Build GRCh38
rs771669084 Chr.13: 39012899-39012899 on Build GRCh38
rs553511129 Chr.13: 39012950-39012950 on Build GRCh38
rs760454269 Chr.13: 39012904-39012904 on Build GRCh38
rs767070879 Chr.13: 39012789-39012789 on Build GRCh38
rs781402133 Chr.13: 39012883-39012883 on Build GRCh38
rs771254099 Chr.13: 39012820-39012820 on Build GRCh38
rs145378291 Chr.13: 39012906-39012906 on Build GRCh38
rs1024868552 Chr.13: 39012800-39012800 on Build GRCh38
rs752513888 Chr.13: 39012935-39012935 on Build GRCh38
rs1048693937 Chr.13: 39012771-39012771 on Build GRCh38
rs757393065 Chr.13: 39012939-39012939 on Build GRCh38
rs773975514 Chr.13: 39012846-39012846 on Build GRCh38
rs150563182 Chr.13: 39012798-39012798 on Build GRCh38
rs147842168 Chr.13: 39012847-39012847 on Build GRCh38
rs765236866 Chr.13: 39012929-39012929 on Build GRCh38
rs374708257 Chr.13: 39012873-39012873 on Build GRCh38
rs369279863 Chr.13: 39012889-39012889 on Build GRCh38
rs746037879 Chr.13: 39012886-39012886 on Build GRCh38
rs762477554 Chr.13: 39012774-39012774 on Build GRCh38
rs886960130 Chr.13: 39012952-39012952 on Build GRCh38
rs200279376 Chr.13: 39012783-39012783 on Build GRCh38
rs765330700 Chr.13: 39012761-39012762 on Build GRCh38
rs775921403 Chr.13: 39012765-39012765 on Build GRCh38
rs751008212 Chr.13: 39012782-39012782 on Build GRCh38
rs747000435 Chr.13: 39012928-39012928 on Build GRCh38
rs746249118 Chr.13: 39012832-39012832 on Build GRCh38
rs141413915 Chr.13: 39012888-39012888 on Build GRCh38
rs776637595 Chr.13: 39012920-39012920 on Build GRCh38
rs199766590 Chr.13: 39012806-39012806 on Build GRCh38
rs201323836 Chr.13: 39012858-39012858 on Build GRCh38
rs773021456 Chr.13: 39012900-39012900 on Build GRCh38
rs17058957 Chr.13: 39012840-39012840 on Build GRCh38
rs375193710 Chr.13: 39012763-39012763 on Build GRCh38
rs771038527 Chr.13: 39012758-39012758 on Build GRCh38
rs760578951 Chr.13: 39012857-39012857 on Build GRCh38
rs41286143 Chr.13: 39012842-39012842 on Build GRCh38
rs751722075 Chr.13: 39012870-39012870 on Build GRCh38
rs1008025726 Chr.13: 39012879-39012879 on Build GRCh38
rs756392723 Chr.13: 39012958-39012958 on Build GRCh38
rs1022195630 Chr.13: 39012845-39012845 on Build GRCh38
rs762275816 Chr.13: 39012760-39012771 on Build GRCh38
rs769262042 Chr.13: 39012891-39012891 on Build GRCh38
rs559540995 Chr.13: 39012796-39012796 on Build GRCh38
rs777960567 Chr.13: 39012816-39012816 on Build GRCh38
rs371650897 Chr.13: 39012964-39012964 on Build GRCh38
rs763606760 Chr.13: 39012778-39012778 on Build GRCh38
rs767629281 Chr.13: 39012941-39012941 on Build GRCh38
rs750299331 Chr.13: 39012791-39012791 on Build GRCh38
rs773093191 Chr.13: 39012894-39012894 on Build GRCh38
rs763559956 Chr.13: 39012841-39012841 on Build GRCh38
rs758430040 Chr.13: 39012812-39012812 on Build GRCh38
rs753787902 Chr.13: 39012807-39012807 on Build GRCh38
rs771795032 Chr.13: 39012863-39012863 on Build GRCh38
rs747030436 Chr.13: 39012819-39012819 on Build GRCh38

View Full Product Details