Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140351400-140351661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
-
UniGene
Hs.446021
Gene Chromosome Location
Chr.7: 140333752-140404352 on Build GRCh38
Transcripts
NM_001287498.1 NM_032295.3 NM_207113.2 XM_011516626.2 XM_011516627.2 XM_017012712.1 XM_017012713.1 XM_017012714.1 XM_017012715.1 XM_017012716.1 XM_017012717.1 XM_017012718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6781919 c.732C>A p.N244K Chr.7: 140351423-140351423 on Build GRCh38
6781916 c.710C>T p.S237L Chr.7: 140351445-140351445 on Build GRCh38
1699447 c.755C>T p.P252L Chr.7: 140351400-140351400 on Build GRCh38
6781918 c.727G>T p.E243* Chr.7: 140351428-140351428 on Build GRCh38
6781920 c.732C>A p.N244K Chr.7: 140351423-140351423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199603017 Chr.7: 140351422-140351422 on Build GRCh38
rs34391142 Chr.7: 140351452-140351452 on Build GRCh38
rs140597201 Chr.7: 140351445-140351445 on Build GRCh38
rs1055569751 Chr.7: 140351569-140351569 on Build GRCh38
rs775652777 Chr.7: 140351453-140351454 on Build GRCh38
rs755623368 Chr.7: 140351465-140351465 on Build GRCh38
rs777242798 Chr.7: 140351445-140351446 on Build GRCh38
rs760696568 Chr.7: 140351439-140351439 on Build GRCh38
rs758348166 Chr.7: 140351453-140351453 on Build GRCh38
rs770563997 Chr.7: 140351464-140351464 on Build GRCh38
rs913094724 Chr.7: 140351444-140351444 on Build GRCh38
rs905945754 Chr.7: 140351575-140351575 on Build GRCh38
rs763993092 Chr.7: 140351457-140351457 on Build GRCh38
rs769186281 Chr.7: 140351494-140351494 on Build GRCh38
rs764253635 Chr.7: 140351456-140351457 on Build GRCh38
rs751345197 Chr.7: 140351458-140351458 on Build GRCh38
rs766459870 Chr.7: 140351440-140351440 on Build GRCh38
rs1053034406 Chr.7: 140351517-140351517 on Build GRCh38
rs767054133 Chr.7: 140351579-140351579 on Build GRCh38
rs925846936 Chr.7: 140351480-140351480 on Build GRCh38
rs748808645 Chr.7: 140351468-140351468 on Build GRCh38
rs950175738 Chr.7: 140351553-140351553 on Build GRCh38
rs1045848660 Chr.7: 140351568-140351568 on Build GRCh38
rs746423466 Chr.7: 140351452-140351453 on Build GRCh38
rs544294335 Chr.7: 140351521-140351521 on Build GRCh38
rs530503242 Chr.7: 140351610-140351610 on Build GRCh38
rs577466422 Chr.7: 140351495-140351495 on Build GRCh38
rs370484205 Chr.7: 140351466-140351467 on Build GRCh38
rs774590007 Chr.7: 140351470-140351471 on Build GRCh38
rs369739327 Chr.7: 140351586-140351586 on Build GRCh38
rs779706538 Chr.7: 140351466-140351466 on Build GRCh38
rs757035931 Chr.7: 140351461-140351461 on Build GRCh38
rs763392222 Chr.7: 140351454-140351455 on Build GRCh38
rs544437038 Chr.7: 140351505-140351505 on Build GRCh38

View Full Product Details