Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1240362-1240635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A18
Entrez Gene ID
348932
Gene Name
solute carrier family 6 member 18
Gene Aliases
Xtrp2
UniGene
Hs.213284
Gene Chromosome Location
Chr.5: 1225355-1246189 on Build GRCh38
Transcripts
NM_182632.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979807 c.854G>A p.C285Y Chr.5: 1240539-1240539 on Build GRCh38
6783252 c.877G>A p.A293T Chr.5: 1240562-1240562 on Build GRCh38
4141389 c.919G>T p.V307F Chr.5: 1240604-1240604 on Build GRCh38
402556 c.855C>A p.C285* Chr.5: 1240540-1240540 on Build GRCh38
7390744 c.919G>A p.V307I Chr.5: 1240604-1240604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764525558 Chr.5: 1240484-1240484 on Build GRCh38
rs570043403 Chr.5: 1240447-1240447 on Build GRCh38
rs778837090 Chr.5: 1240511-1240511 on Build GRCh38
rs532961369 Chr.5: 1240427-1240427 on Build GRCh38
rs1043347508 Chr.5: 1240577-1240577 on Build GRCh38
rs200088793 Chr.5: 1240510-1240510 on Build GRCh38
rs777019163 Chr.5: 1240561-1240561 on Build GRCh38
rs375296591 Chr.5: 1240528-1240528 on Build GRCh38
rs114838763 Chr.5: 1240404-1240404 on Build GRCh38
rs184008653 Chr.5: 1240538-1240538 on Build GRCh38
rs781203778 Chr.5: 1240523-1240523 on Build GRCh38
rs781108917 Chr.5: 1240600-1240600 on Build GRCh38
rs780428620 Chr.5: 1240517-1240517 on Build GRCh38
rs984791776 Chr.5: 1240438-1240438 on Build GRCh38
rs747992391 Chr.5: 1240601-1240601 on Build GRCh38
rs995604048 Chr.5: 1240544-1240544 on Build GRCh38
rs953627536 Chr.5: 1240416-1240416 on Build GRCh38
rs770421232 Chr.5: 1240541-1240541 on Build GRCh38
rs774004237 Chr.5: 1240547-1240547 on Build GRCh38
rs757849118 Chr.5: 1240508-1240508 on Build GRCh38
rs373234068 Chr.5: 1240594-1240594 on Build GRCh38
rs772544172 Chr.5: 1240535-1240535 on Build GRCh38
rs144574225 Chr.5: 1240606-1240606 on Build GRCh38
rs916328610 Chr.5: 1240534-1240534 on Build GRCh38
rs754329179 Chr.5: 1240502-1240502 on Build GRCh38
rs149285920 Chr.5: 1240591-1240591 on Build GRCh38
rs946211912 Chr.5: 1240473-1240473 on Build GRCh38
rs764437677 Chr.5: 1240552-1240552 on Build GRCh38
rs997977561 Chr.5: 1240386-1240386 on Build GRCh38
rs937369157 Chr.5: 1240436-1240436 on Build GRCh38
rs757590797 Chr.5: 1240408-1240408 on Build GRCh38
rs762179366 Chr.5: 1240562-1240562 on Build GRCh38
rs893431014 Chr.5: 1240470-1240470 on Build GRCh38
rs890838998 Chr.5: 1240492-1240492 on Build GRCh38
rs750538118 Chr.5: 1240514-1240514 on Build GRCh38
rs200879273 Chr.5: 1240519-1240519 on Build GRCh38
rs201308114 Chr.5: 1240576-1240576 on Build GRCh38
rs565008139 Chr.5: 1240421-1240421 on Build GRCh38
rs750404758 Chr.5: 1240583-1240583 on Build GRCh38
rs368251762 Chr.5: 1240551-1240551 on Build GRCh38
rs751831237 Chr.5: 1240593-1240593 on Build GRCh38
rs1039746566 Chr.5: 1240518-1240518 on Build GRCh38
rs777767601 Chr.5: 1240602-1240602 on Build GRCh38
rs372591269 Chr.5: 1240603-1240603 on Build GRCh38
rs947817624 Chr.5: 1240568-1240568 on Build GRCh38
rs548993851 Chr.5: 1240515-1240515 on Build GRCh38
rs190914195 Chr.5: 1240480-1240480 on Build GRCh38
rs909242175 Chr.5: 1240464-1240464 on Build GRCh38
rs551122975 Chr.5: 1240428-1240428 on Build GRCh38
rs766627247 Chr.5: 1240592-1240592 on Build GRCh38

View Full Product Details