Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100108478-100108711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF6
Entrez Gene ID
6878
Gene Name
TATA-box binding protein associated factor 6
Gene Aliases
MGC:8964, TAF(II)70, TAF(II)80, TAF2E, TAFII-70, TAFII-80, TAFII70, TAFII80, TAFII85
UniGene
Hs.489309
Gene Chromosome Location
Chr.7: 100107070-100119858 on Build GRCh38
Transcripts
NM_001190415.1 NM_005641.3 NM_139315.2 NR_033792.1 XM_006716100.2 XM_006716101.2 XM_006716102.2 XM_011516542.2 XM_011516543.2 XM_017012568.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
289130 c.1315C>T p.R439C Chr.7: 100108510-100108510 on Build GRCh38
39345 c.1347G>A p.R449R Chr.7: 100108478-100108478 on Build GRCh38
7148874 c.1327G>A p.D443N Chr.7: 100108498-100108498 on Build GRCh38
252859 c.1298C>A p.P433H Chr.7: 100108527-100108527 on Build GRCh38
6796427 c.1323G>A p.P441P Chr.7: 100108502-100108502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753585341 Chr.7: 100108567-100108570 on Build GRCh38
rs745474049 Chr.7: 100108565-100108565 on Build GRCh38
rs542521008 Chr.7: 100108646-100108648 on Build GRCh38
rs750135146 Chr.7: 100108634-100108634 on Build GRCh38
rs149977983 Chr.7: 100108529-100108529 on Build GRCh38
rs760633502 Chr.7: 100108559-100108559 on Build GRCh38
rs934943423 Chr.7: 100108552-100108555 on Build GRCh38
rs147878660 Chr.7: 100108503-100108503 on Build GRCh38
rs938643858 Chr.7: 100108647-100108648 on Build GRCh38
rs767049871 Chr.7: 100108613-100108613 on Build GRCh38
rs4134923 Chr.7: 100108680-100108680 on Build GRCh38
rs763953183 Chr.7: 100108502-100108502 on Build GRCh38
rs373378029 Chr.7: 100108580-100108608 on Build GRCh38
rs115360193 Chr.7: 100108595-100108595 on Build GRCh38
rs749666042 Chr.7: 100108552-100108552 on Build GRCh38
rs763892748 Chr.7: 100108546-100108546 on Build GRCh38
rs1057181664 Chr.7: 100108678-100108678 on Build GRCh38
rs368267898 Chr.7: 100108568-100108568 on Build GRCh38
rs746516738 Chr.7: 100108578-100108578 on Build GRCh38
rs766556536 Chr.7: 100108509-100108509 on Build GRCh38
rs140517175 Chr.7: 100108523-100108523 on Build GRCh38
rs766146041 Chr.7: 100108566-100108569 on Build GRCh38
rs755135145 Chr.7: 100108513-100108513 on Build GRCh38
rs760547433 Chr.7: 100108500-100108500 on Build GRCh38
rs926724129 Chr.7: 100108627-100108627 on Build GRCh38
rs371241346 Chr.7: 100108574-100108574 on Build GRCh38
rs112906003 Chr.7: 100108542-100108542 on Build GRCh38
rs776606499 Chr.7: 100108585-100108585 on Build GRCh38
rs1041642903 Chr.7: 100108682-100108682 on Build GRCh38
rs778461262 Chr.7: 100108564-100108564 on Build GRCh38
rs980799610 Chr.7: 100108605-100108605 on Build GRCh38
rs892506024 Chr.7: 100108641-100108641 on Build GRCh38
rs1054176833 Chr.7: 100108637-100108637 on Build GRCh38
rs373104578 Chr.7: 100108537-100108537 on Build GRCh38
rs751770909 Chr.7: 100108510-100108510 on Build GRCh38
rs757612084 Chr.7: 100108561-100108561 on Build GRCh38
rs761653566 Chr.7: 100108508-100108508 on Build GRCh38
rs773100801 Chr.7: 100108553-100108581 on Build GRCh38
rs768530981 Chr.7: 100108584-100108584 on Build GRCh38

View Full Product Details