Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48918511-48918766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIM2
Entrez Gene ID
11040
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
Gene Aliases
-
UniGene
Hs.727148
Gene Chromosome Location
Chr.X: 48913182-48919136 on Build GRCh38
Transcripts
NM_006875.3

SNP Detail

SNP ID Chromosome Location
rs782077114 Chr.X: 48918641-48918641 on Build GRCh38
rs782014876 Chr.X: 48918598-48918598 on Build GRCh38
rs111456785 Chr.X: 48918682-48918682 on Build GRCh38
rs782439008 Chr.X: 48918742-48918742 on Build GRCh38
rs782584800 Chr.X: 48918744-48918744 on Build GRCh38
rs201419725 Chr.X: 48918684-48918684 on Build GRCh38
rs781824342 Chr.X: 48918664-48918664 on Build GRCh38
rs782602485 Chr.X: 48918736-48918736 on Build GRCh38
rs782158356 Chr.X: 48918599-48918599 on Build GRCh38
rs893933061 Chr.X: 48918674-48918674 on Build GRCh38
rs782271303 Chr.X: 48918745-48918745 on Build GRCh38
rs782074676 Chr.X: 48918715-48918715 on Build GRCh38
rs928985123 Chr.X: 48918543-48918543 on Build GRCh38
rs782757616 Chr.X: 48918602-48918602 on Build GRCh38
rs782017459 Chr.X: 48918614-48918614 on Build GRCh38
rs1034273092 Chr.X: 48918617-48918617 on Build GRCh38
rs782740968 Chr.X: 48918657-48918657 on Build GRCh38
rs1011424498 Chr.X: 48918703-48918703 on Build GRCh38
rs202169102 Chr.X: 48918661-48918661 on Build GRCh38
rs782806614 Chr.X: 48918662-48918662 on Build GRCh38
rs781932883 Chr.X: 48918690-48918690 on Build GRCh38
rs782233920 Chr.X: 48918741-48918741 on Build GRCh38
rs781812758 Chr.X: 48918660-48918660 on Build GRCh38
rs782321972 Chr.X: 48918573-48918573 on Build GRCh38
rs782515557 Chr.X: 48918694-48918694 on Build GRCh38
rs966750827 Chr.X: 48918734-48918734 on Build GRCh38
rs781994307 Chr.X: 48918622-48918622 on Build GRCh38
rs148882154 Chr.X: 48918572-48918572 on Build GRCh38

View Full Product Details