Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32402797-32403038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
N4BP2L1
Entrez Gene ID
90634
Gene Name
NEDD4 binding protein 2 like 1
Gene Aliases
CG018
UniGene
Hs.732118
Gene Chromosome Location
Chr.13: 32400723-32429649 on Build GRCh38
Transcripts
NM_001079691.1 NM_001286459.1 NM_001286460.1 NM_001286461.1 NM_052818.2 XM_006719890.3 XM_011535301.2 XM_011535302.2 XM_011535303.2 XM_011535304.2 XM_017020836.1 XM_017020837.1 XM_017020838.1 XM_017020840.1 XM_017020841.1 XM_017020842.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2071591 c.670C>T p.H224Y Chr.13: 32403004-32403004 on Build GRCh38
6995843 c.662G>T p.R221L Chr.13: 32403012-32403012 on Build GRCh38
4427887 c.726_727insA p.Y243fs*>2 Chr.13: 32402947-32402948 on Build GRCh38
2071590 c.673G>A p.G225S Chr.13: 32403001-32403001 on Build GRCh38
5052126 c.661C>T p.R221W Chr.13: 32403013-32403013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567290551 Chr.13: 32402908-32402909 on Build GRCh38
rs569660230 Chr.13: 32402929-32402929 on Build GRCh38
rs193282815 Chr.13: 32402940-32402940 on Build GRCh38
rs906140130 Chr.13: 32402860-32402860 on Build GRCh38
rs747101340 Chr.13: 32402930-32402930 on Build GRCh38
rs573641521 Chr.13: 32402975-32402975 on Build GRCh38
rs368732329 Chr.13: 32402966-32402966 on Build GRCh38
rs746123400 Chr.13: 32402938-32402938 on Build GRCh38
rs752616019 Chr.13: 32403002-32403002 on Build GRCh38
rs934830009 Chr.13: 32402869-32402869 on Build GRCh38
rs754971431 Chr.13: 32402928-32402928 on Build GRCh38
rs760439916 Chr.13: 32402963-32402963 on Build GRCh38
rs977032150 Chr.13: 32402977-32402979 on Build GRCh38
rs759462544 Chr.13: 32402984-32402984 on Build GRCh38
rs371694391 Chr.13: 32403001-32403001 on Build GRCh38
rs766303483 Chr.13: 32402900-32402900 on Build GRCh38
rs769161117 Chr.13: 32402957-32402957 on Build GRCh38
rs138167620 Chr.13: 32402961-32402961 on Build GRCh38
rs761379595 Chr.13: 32402960-32402960 on Build GRCh38
rs745349511 Chr.13: 32403006-32403006 on Build GRCh38
rs781442890 Chr.13: 32402935-32402935 on Build GRCh38
rs774775326 Chr.13: 32402958-32402958 on Build GRCh38
rs749724555 Chr.13: 32402955-32402955 on Build GRCh38
rs540784451 Chr.13: 32403012-32403012 on Build GRCh38
rs753790181 Chr.13: 32402926-32402926 on Build GRCh38
rs770095742 Chr.13: 32402943-32402943 on Build GRCh38
rs570403210 Chr.13: 32402880-32402882 on Build GRCh38
rs757348552 Chr.13: 32402934-32402934 on Build GRCh38
rs149135496 Chr.13: 32402962-32402962 on Build GRCh38
rs750617706 Chr.13: 32403013-32403013 on Build GRCh38
rs757335831 Chr.13: 32403003-32403003 on Build GRCh38
rs949039096 Chr.13: 32402954-32402954 on Build GRCh38
rs780427619 Chr.13: 32402948-32402948 on Build GRCh38

View Full Product Details