Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230461161-230461432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP100
Entrez Gene ID
6672
Gene Name
SP100 nuclear antigen
Gene Aliases
lysp100b
UniGene
Hs.369056
Gene Chromosome Location
Chr.2: 230416156-230545602 on Build GRCh38
Transcripts
NM_001080391.1 NM_001206701.1 NM_001206702.1 NM_001206703.1 NM_001206704.1 NM_003113.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
720287 c.862T>C p.S288P Chr.2: 230461303-230461303 on Build GRCh38
6566722 c.954C>T p.N318N Chr.2: 230461395-230461395 on Build GRCh38
7391493 c.875G>A p.R292Q Chr.2: 230461316-230461316 on Build GRCh38
6787681 c.874C>T p.R292* Chr.2: 230461315-230461315 on Build GRCh38
3719578 c.924G>C p.E308D Chr.2: 230461365-230461365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772049350 Chr.2: 230461378-230461378 on Build GRCh38
rs769958142 Chr.2: 230461279-230461279 on Build GRCh38
rs750142442 Chr.2: 230461317-230461317 on Build GRCh38
rs751858461 Chr.2: 230461314-230461314 on Build GRCh38
rs776977782 Chr.2: 230461254-230461254 on Build GRCh38
rs574355188 Chr.2: 230461322-230461322 on Build GRCh38
rs201597647 Chr.2: 230461214-230461214 on Build GRCh38
rs779758329 Chr.2: 230461256-230461256 on Build GRCh38
rs779573676 Chr.2: 230461345-230461345 on Build GRCh38
rs543549365 Chr.2: 230461392-230461392 on Build GRCh38
rs142803787 Chr.2: 230461388-230461388 on Build GRCh38
rs779836216 Chr.2: 230461217-230461217 on Build GRCh38
rs753416593 Chr.2: 230461356-230461356 on Build GRCh38
rs116484154 Chr.2: 230461316-230461316 on Build GRCh38
rs757941812 Chr.2: 230461346-230461346 on Build GRCh38
rs199835321 Chr.2: 230461242-230461243 on Build GRCh38
rs35506327 Chr.2: 230461366-230461366 on Build GRCh38
rs924239827 Chr.2: 230461219-230461219 on Build GRCh38
rs546582941 Chr.2: 230461283-230461283 on Build GRCh38
rs571577080 Chr.2: 230461284-230461284 on Build GRCh38
rs989829568 Chr.2: 230461224-230461224 on Build GRCh38
rs778997734 Chr.2: 230461372-230461372 on Build GRCh38
rs749284583 Chr.2: 230461222-230461222 on Build GRCh38
rs927349968 Chr.2: 230461221-230461221 on Build GRCh38
rs143684378 Chr.2: 230461379-230461379 on Build GRCh38
rs759520604 Chr.2: 230461263-230461263 on Build GRCh38
rs79230352 Chr.2: 230461229-230461229 on Build GRCh38
rs762236298 Chr.2: 230461315-230461315 on Build GRCh38
rs75682703 Chr.2: 230461207-230461207 on Build GRCh38
rs1045754346 Chr.2: 230461216-230461216 on Build GRCh38
rs747952714 Chr.2: 230461373-230461373 on Build GRCh38
rs200256890 Chr.2: 230461239-230461239 on Build GRCh38
rs763026686 Chr.2: 230461407-230461407 on Build GRCh38
rs775466318 Chr.2: 230461281-230461281 on Build GRCh38
rs755545681 Chr.2: 230461342-230461342 on Build GRCh38
rs769151070 Chr.2: 230461408-230461408 on Build GRCh38
rs747966975 Chr.2: 230461237-230461237 on Build GRCh38

View Full Product Details