Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102707384-102707657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCOR1
Entrez Gene ID
23186
Gene Name
REST corepressor 1
Gene Aliases
COREST, RCOR
UniGene
Hs.510521
Gene Chromosome Location
Chr.14: 102592638-102730576 on Build GRCh38
Transcripts
NM_015156.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955819 c.582G>T p.V194V Chr.14: 102707443-102707443 on Build GRCh38
216311 c.526G>T p.E176* Chr.14: 102707387-102707387 on Build GRCh38
6764536 c.620A>G p.H207R Chr.14: 102707481-102707481 on Build GRCh38
6764533 c.600T>G p.F200L Chr.14: 102707461-102707461 on Build GRCh38
1368368 c.525C>T p.I175I Chr.14: 102707386-102707386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992080338 Chr.14: 102707616-102707616 on Build GRCh38
rs535690691 Chr.14: 102707630-102707630 on Build GRCh38
rs116647419 Chr.14: 102707611-102707611 on Build GRCh38
rs771574913 Chr.14: 102707522-102707523 on Build GRCh38
rs763718967 Chr.14: 102707515-102707515 on Build GRCh38
rs894842885 Chr.14: 102707591-102707591 on Build GRCh38
rs765565533 Chr.14: 102707495-102707495 on Build GRCh38
rs776001279 Chr.14: 102707523-102707533 on Build GRCh38
rs766707154 Chr.14: 102707555-102707558 on Build GRCh38
rs773148505 Chr.14: 102707438-102707438 on Build GRCh38
rs754818516 Chr.14: 102707563-102707563 on Build GRCh38
rs997362481 Chr.14: 102707428-102707428 on Build GRCh38
rs371811406 Chr.14: 102707553-102707553 on Build GRCh38
rs566201626 Chr.14: 102707557-102707557 on Build GRCh38
rs772257397 Chr.14: 102707434-102707434 on Build GRCh38
rs995295562 Chr.14: 102707569-102707569 on Build GRCh38
rs1024909652 Chr.14: 102707615-102707615 on Build GRCh38
rs779705436 Chr.14: 102707558-102707558 on Build GRCh38
rs142321093 Chr.14: 102707469-102707469 on Build GRCh38
rs374454637 Chr.14: 102707561-102707561 on Build GRCh38
rs544046280 Chr.14: 102707540-102707540 on Build GRCh38
rs144845698 Chr.14: 102707531-102707531 on Build GRCh38
rs376570636 Chr.14: 102707503-102707503 on Build GRCh38
rs759848181 Chr.14: 102707475-102707475 on Build GRCh38
rs750967629 Chr.14: 102707520-102707520 on Build GRCh38
rs763366058 Chr.14: 102707506-102707506 on Build GRCh38
rs750076340 Chr.14: 102707537-102707537 on Build GRCh38
rs747273034 Chr.14: 102707449-102707449 on Build GRCh38
rs550305555 Chr.14: 102707572-102707572 on Build GRCh38
rs776105969 Chr.14: 102707501-102707501 on Build GRCh38
rs747567409 Chr.14: 102707539-102707540 on Build GRCh38
rs369257488 Chr.14: 102707521-102707521 on Build GRCh38
rs149418829 Chr.14: 102707419-102707419 on Build GRCh38
rs372167310 Chr.14: 102707539-102707539 on Build GRCh38
rs777014780 Chr.14: 102707473-102707473 on Build GRCh38
rs749146784 Chr.14: 102707422-102707422 on Build GRCh38

View Full Product Details