Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148070364-148070589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINK5
Entrez Gene ID
11005
Gene Name
serine peptidase inhibitor, Kazal type 5
Gene Aliases
LEKTI, LETKI, NETS, NS, VAKTI
UniGene
Hs.331555
Gene Chromosome Location
Chr.5: 148063972-148137362 on Build GRCh38
Transcripts
NM_001127698.1 NM_001127699.1 NM_006846.3 XM_011537551.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3734244 c.186A>T p.K62N Chr.5: 148070427-148070427 on Build GRCh38
3776405 c.154C>G p.Q52E Chr.5: 148070395-148070395 on Build GRCh38
6470056 c.185A>G p.K62R Chr.5: 148070426-148070426 on Build GRCh38
1063958 c.191C>T p.A64V Chr.5: 148070432-148070432 on Build GRCh38
1063956 c.177C>A p.F59L Chr.5: 148070418-148070418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994493880 Chr.5: 148070535-148070535 on Build GRCh38
rs377031902 Chr.5: 148070423-148070423 on Build GRCh38
rs369887596 Chr.5: 148070551-148070551 on Build GRCh38
rs767861897 Chr.5: 148070462-148070465 on Build GRCh38
rs754555442 Chr.5: 148070471-148070471 on Build GRCh38
rs563162543 Chr.5: 148070524-148070524 on Build GRCh38
rs376634443 Chr.5: 148070484-148070484 on Build GRCh38
rs747871163 Chr.5: 148070480-148070480 on Build GRCh38
rs765120512 Chr.5: 148070442-148070442 on Build GRCh38
rs761129697 Chr.5: 148070484-148070488 on Build GRCh38
rs986646323 Chr.5: 148070391-148070391 on Build GRCh38
rs75257702 Chr.5: 148070420-148070420 on Build GRCh38
rs370397387 Chr.5: 148070436-148070436 on Build GRCh38
rs540712189 Chr.5: 148070408-148070408 on Build GRCh38
rs552548594 Chr.5: 148070435-148070435 on Build GRCh38
rs78216695 Chr.5: 148070482-148070482 on Build GRCh38
rs780665939 Chr.5: 148070478-148070478 on Build GRCh38
rs575743379 Chr.5: 148070547-148070547 on Build GRCh38
rs750310449 Chr.5: 148070453-148070453 on Build GRCh38
rs771729229 Chr.5: 148070415-148070415 on Build GRCh38
rs779710236 Chr.5: 148070466-148070466 on Build GRCh38
rs748409924 Chr.5: 148070407-148070407 on Build GRCh38
rs773557840 Chr.5: 148070414-148070414 on Build GRCh38
rs760304019 Chr.5: 148070517-148070517 on Build GRCh38
rs746456432 Chr.5: 148070468-148070468 on Build GRCh38
rs3752676 Chr.5: 148070522-148070522 on Build GRCh38
rs766432990 Chr.5: 148070421-148070421 on Build GRCh38
rs759615234 Chr.5: 148070429-148070429 on Build GRCh38
rs773569272 Chr.5: 148070482-148070482 on Build GRCh38
rs773683150 Chr.5: 148070497-148070497 on Build GRCh38
rs3752677 Chr.5: 148070465-148070465 on Build GRCh38

View Full Product Details