Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72688570-72688843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC1
Entrez Gene ID
117531
Gene Name
transmembrane channel like 1
Gene Aliases
DFNA36, DFNB11, DFNB7
UniGene
Hs.670211
Gene Chromosome Location
Chr.9: 72521801-72836351 on Build GRCh38
Transcripts
NM_138691.2 XM_017014256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3763989 c.45C>T p.D15D Chr.9: 72688737-72688737 on Build GRCh38
5963085 c.17-9C>T p.? Chr.9: 72688700-72688700 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776220877 Chr.9: 72688744-72688744 on Build GRCh38
rs1033776269 Chr.9: 72688775-72688775 on Build GRCh38
rs138738150 Chr.9: 72688621-72688621 on Build GRCh38
rs202182752 Chr.9: 72688663-72688663 on Build GRCh38
rs372383725 Chr.9: 72688715-72688715 on Build GRCh38
rs994984077 Chr.9: 72688622-72688622 on Build GRCh38
rs752499835 Chr.9: 72688765-72688765 on Build GRCh38
rs753008093 Chr.9: 72688802-72688802 on Build GRCh38
rs762858152 Chr.9: 72688767-72688767 on Build GRCh38
rs757765718 Chr.9: 72688723-72688723 on Build GRCh38
rs144387828 Chr.9: 72688756-72688756 on Build GRCh38
rs141858568 Chr.9: 72688783-72688783 on Build GRCh38
rs777874004 Chr.9: 72688706-72688706 on Build GRCh38
rs770832842 Chr.9: 72688778-72688778 on Build GRCh38
rs751191888 Chr.9: 72688669-72688669 on Build GRCh38
rs2589615 Chr.9: 72688737-72688737 on Build GRCh38
rs368677040 Chr.9: 72688700-72688700 on Build GRCh38
rs727504554 Chr.9: 72688714-72688714 on Build GRCh38
rs762917092 Chr.9: 72688668-72688668 on Build GRCh38
rs752846422 Chr.9: 72688703-72688703 on Build GRCh38
rs767879681 Chr.9: 72688797-72688797 on Build GRCh38
rs189479995 Chr.9: 72688751-72688751 on Build GRCh38
rs140437301 Chr.9: 72688738-72688738 on Build GRCh38
rs779209726 Chr.9: 72688736-72688736 on Build GRCh38
rs920608178 Chr.9: 72688792-72688792 on Build GRCh38
rs751389124 Chr.9: 72688670-72688670 on Build GRCh38
rs748339266 Chr.9: 72688782-72688782 on Build GRCh38
rs755253599 Chr.9: 72688691-72688691 on Build GRCh38
rs746805550 Chr.9: 72688822-72688822 on Build GRCh38
rs756106519 Chr.9: 72688704-72688704 on Build GRCh38
rs1049333804 Chr.9: 72688648-72688648 on Build GRCh38
rs774299542 Chr.9: 72688781-72688781 on Build GRCh38
rs1004890362 Chr.9: 72688684-72688684 on Build GRCh38

View Full Product Details