Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 124042456-124042705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAG2
Entrez Gene ID
10735
Gene Name
stromal antigen 2
Gene Aliases
SA-2, SA2, SCC3B, bA517O1.1
UniGene
Hs.496710
Gene Chromosome Location
Chr.X: 123960560-124102656 on Build GRCh38
Transcripts
NM_001042749.2 NM_001042750.1 NM_001042751.1 NM_001282418.1 NM_006603.4 XM_005262357.2 XM_005262358.2 XM_005262359.3 XM_005262360.2 XM_005262361.2 XM_006724727.1 XM_006724728.3 XM_011531253.2 XM_017029232.1 XM_017029233.1 XM_017029234.1 XM_017029235.1

SNP Detail

SNP ID Chromosome Location
rs760011591 Chr.X: 124042528-124042528 on Build GRCh38
rs150057715 Chr.X: 124042580-124042580 on Build GRCh38
rs964233454 Chr.X: 124042505-124042505 on Build GRCh38
rs751594711 Chr.X: 124042663-124042663 on Build GRCh38
rs987756014 Chr.X: 124042672-124042672 on Build GRCh38
rs184515751 Chr.X: 124042533-124042533 on Build GRCh38
rs145475901 Chr.X: 124042591-124042591 on Build GRCh38
rs768142839 Chr.X: 124042521-124042521 on Build GRCh38
rs764304764 Chr.X: 124042595-124042595 on Build GRCh38
rs750274139 Chr.X: 124042521-124042522 on Build GRCh38
rs753059756 Chr.X: 124042553-124042553 on Build GRCh38
rs199610347 Chr.X: 124042561-124042561 on Build GRCh38

View Full Product Details