Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26735147-26735420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDSS1
Entrez Gene ID
23590
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
Gene Aliases
COQ1, COQ10D2, DPS, SPS, TPRT, TPT, TPT 1, hDPS1
UniGene
Hs.558468
Gene Chromosome Location
Chr.10: 26697052-26746798 on Build GRCh38
Transcripts
NM_001321978.1 NM_001321979.1 NM_014317.4 XM_011519437.2 XM_017016011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5881896 c.832-1G>A p.? Chr.10: 26735239-26735239 on Build GRCh38
4137943 c.912+1G>T p.? Chr.10: 26735321-26735321 on Build GRCh38
1474571 c.874G>A p.A292T Chr.10: 26735282-26735282 on Build GRCh38
7152865 c.836C>G p.S279C Chr.10: 26735244-26735244 on Build GRCh38
6743480 c.852C>T p.P284P Chr.10: 26735260-26735260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748807129 Chr.10: 26735232-26735232 on Build GRCh38
rs369844695 Chr.10: 26735259-26735259 on Build GRCh38
rs376937544 Chr.10: 26735379-26735379 on Build GRCh38
rs771655954 Chr.10: 26735260-26735260 on Build GRCh38
rs147864501 Chr.10: 26735261-26735261 on Build GRCh38
rs982641022 Chr.10: 26735188-26735188 on Build GRCh38
rs771257367 Chr.10: 26735329-26735329 on Build GRCh38
rs570909836 Chr.10: 26735365-26735365 on Build GRCh38
rs772901240 Chr.10: 26735286-26735286 on Build GRCh38
rs367783149 Chr.10: 26735268-26735268 on Build GRCh38
rs768568415 Chr.10: 26735267-26735267 on Build GRCh38
rs778924698 Chr.10: 26735202-26735202 on Build GRCh38
rs199531574 Chr.10: 26735245-26735245 on Build GRCh38
rs748250156 Chr.10: 26735225-26735226 on Build GRCh38
rs1038764495 Chr.10: 26735290-26735290 on Build GRCh38
rs755480514 Chr.10: 26735200-26735200 on Build GRCh38
rs79708052 Chr.10: 26735394-26735394 on Build GRCh38
rs763165754 Chr.10: 26735294-26735294 on Build GRCh38
rs765310423 Chr.10: 26735282-26735282 on Build GRCh38
rs915938490 Chr.10: 26735344-26735344 on Build GRCh38
rs528693323 Chr.10: 26735342-26735342 on Build GRCh38
rs769459722 Chr.10: 26735181-26735181 on Build GRCh38
rs141486367 Chr.10: 26735371-26735371 on Build GRCh38
rs373761875 Chr.10: 26735357-26735357 on Build GRCh38
rs762662757 Chr.10: 26735302-26735302 on Build GRCh38
rs755457664 Chr.10: 26735328-26735328 on Build GRCh38
rs534121169 Chr.10: 26735293-26735293 on Build GRCh38
rs893043176 Chr.10: 26735366-26735366 on Build GRCh38
rs773432018 Chr.10: 26735334-26735336 on Build GRCh38
rs781778025 Chr.10: 26735231-26735231 on Build GRCh38
rs753310443 Chr.10: 26735353-26735353 on Build GRCh38
rs751955353 Chr.10: 26735311-26735311 on Build GRCh38
rs1023588581 Chr.10: 26735380-26735380 on Build GRCh38
rs756603214 Chr.10: 26735354-26735354 on Build GRCh38
rs772225677 Chr.10: 26735226-26735229 on Build GRCh38
rs778411730 Chr.10: 26735240-26735240 on Build GRCh38
rs770518092 Chr.10: 26735236-26735236 on Build GRCh38
rs766719266 Chr.10: 26735301-26735301 on Build GRCh38
rs556495037 Chr.10: 26735391-26735391 on Build GRCh38
rs761776627 Chr.10: 26735270-26735270 on Build GRCh38
rs745477727 Chr.10: 26735246-26735246 on Build GRCh38

View Full Product Details