Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4531688-4531941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLIN5
Entrez Gene ID
440503
Gene Name
perilipin 5
Gene Aliases
LSDA5, LSDP5, MLDP, OXPAT
UniGene
Hs.131034
Gene Chromosome Location
Chr.19: 4522531-4535196 on Build GRCh38
Transcripts
NM_001013706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998069 c.178G>A p.A60T Chr.19: 4531705-4531705 on Build GRCh38
1393658 c.83C>T p.A28V Chr.19: 4531800-4531800 on Build GRCh38
6395249 c.133G>T p.A45S Chr.19: 4531750-4531750 on Build GRCh38
998072 c.111C>T p.T37T Chr.19: 4531772-4531772 on Build GRCh38
998070 c.150C>T p.H50H Chr.19: 4531733-4531733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752887556 Chr.19: 4531820-4531820 on Build GRCh38
rs1042094637 Chr.19: 4531773-4531773 on Build GRCh38
rs747193089 Chr.19: 4531731-4531731 on Build GRCh38
rs998941522 Chr.19: 4531804-4531804 on Build GRCh38
rs766725113 Chr.19: 4531810-4531810 on Build GRCh38
rs374372886 Chr.19: 4531710-4531710 on Build GRCh38
rs769399556 Chr.19: 4531796-4531796 on Build GRCh38
rs773347466 Chr.19: 4531706-4531712 on Build GRCh38
rs761389712 Chr.19: 4531722-4531722 on Build GRCh38
rs950779750 Chr.19: 4531876-4531877 on Build GRCh38
rs959998941 Chr.19: 4531845-4531845 on Build GRCh38
rs778512170 Chr.19: 4531708-4531708 on Build GRCh38
rs771881511 Chr.19: 4531771-4531771 on Build GRCh38
rs754047368 Chr.19: 4531848-4531848 on Build GRCh38
rs868322899 Chr.19: 4531813-4531813 on Build GRCh38
rs375098526 Chr.19: 4531809-4531809 on Build GRCh38
rs769425840 Chr.19: 4531827-4531827 on Build GRCh38
rs376635614 Chr.19: 4531747-4531747 on Build GRCh38
rs752063787 Chr.19: 4531748-4531748 on Build GRCh38
rs779760982 Chr.19: 4531857-4531857 on Build GRCh38
rs773230794 Chr.19: 4531864-4531864 on Build GRCh38
rs776062542 Chr.19: 4531778-4531778 on Build GRCh38
rs551820561 Chr.19: 4531817-4531817 on Build GRCh38
rs35224272 Chr.19: 4531881-4531882 on Build GRCh38
rs528560540 Chr.19: 4531717-4531717 on Build GRCh38
rs772890622 Chr.19: 4531798-4531798 on Build GRCh38
rs781102630 Chr.19: 4531869-4531869 on Build GRCh38
rs927831077 Chr.19: 4531731-4531731 on Build GRCh38
rs537780687 Chr.19: 4531909-4531909 on Build GRCh38
rs756460745 Chr.19: 4531769-4531769 on Build GRCh38
rs781204109 Chr.19: 4531762-4531762 on Build GRCh38
rs572682599 Chr.19: 4531729-4531729 on Build GRCh38
rs1049276214 Chr.19: 4531730-4531730 on Build GRCh38
rs775000745 Chr.19: 4531812-4531812 on Build GRCh38
rs929548067 Chr.19: 4531858-4531858 on Build GRCh38
rs779882960 Chr.19: 4531772-4531772 on Build GRCh38
rs758161524 Chr.19: 4531850-4531850 on Build GRCh38
rs746963928 Chr.19: 4531862-4531862 on Build GRCh38
rs370914998 Chr.19: 4531718-4531718 on Build GRCh38
rs768161547 Chr.19: 4531777-4531777 on Build GRCh38
rs76461382 Chr.19: 4531907-4531907 on Build GRCh38
rs771309924 Chr.19: 4531762-4531784 on Build GRCh38
rs754968327 Chr.19: 4531866-4531866 on Build GRCh38
rs768121130 Chr.19: 4531819-4531819 on Build GRCh38
rs969743638 Chr.19: 4531825-4531825 on Build GRCh38
rs752979298 Chr.19: 4531763-4531763 on Build GRCh38
rs980291688 Chr.19: 4531826-4531826 on Build GRCh38
rs755138773 Chr.19: 4531751-4531751 on Build GRCh38
rs541598275 Chr.19: 4531915-4531915 on Build GRCh38
rs370675802 Chr.19: 4531770-4531770 on Build GRCh38
rs1030539612 Chr.19: 4531805-4531806 on Build GRCh38
rs897975377 Chr.19: 4531719-4531719 on Build GRCh38
rs747987953 Chr.19: 4531779-4531779 on Build GRCh38
rs963348975 Chr.19: 4531840-4531840 on Build GRCh38
rs377467819 Chr.19: 4531709-4531709 on Build GRCh38
rs927851559 Chr.19: 4531908-4531908 on Build GRCh38
rs943432611 Chr.19: 4531917-4531917 on Build GRCh38
rs375141454 Chr.19: 4531735-4531735 on Build GRCh38
rs746911390 Chr.19: 4531775-4531775 on Build GRCh38

View Full Product Details