Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25603605-25603878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL39
Entrez Gene ID
54148
Gene Name
mitochondrial ribosomal protein L39
Gene Aliases
C21orf92, L39mt, MRP-L5, MRPL5, MSTP003, PRED22, PRED66, RPML5
UniGene
Hs.420696
Gene Chromosome Location
Chr.21: 25585656-25607858 on Build GRCh38
Transcripts
NM_017446.3 NM_080794.3 XM_006724026.3 XM_011529651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2841251 c.416A>T p.N139I Chr.21: 25603800-25603800 on Build GRCh38
3549949 c.414G>A p.V138V Chr.21: 25603802-25603802 on Build GRCh38
7429656 c.394G>C p.D132H Chr.21: 25603822-25603822 on Build GRCh38
3549951 c.403C>T p.P135S Chr.21: 25603813-25603813 on Build GRCh38
4134847 c.386C>A p.T129N Chr.21: 25603830-25603830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145584095 Chr.21: 25603682-25603682 on Build GRCh38
rs186802528 Chr.21: 25603721-25603721 on Build GRCh38
rs772303475 Chr.21: 25603751-25603751 on Build GRCh38
rs199838152 Chr.21: 25603757-25603757 on Build GRCh38
rs112541853 Chr.21: 25603678-25603678 on Build GRCh38
rs750486484 Chr.21: 25603669-25603669 on Build GRCh38
rs762325514 Chr.21: 25603641-25603641 on Build GRCh38
rs181107715 Chr.21: 25603702-25603702 on Build GRCh38
rs371255612 Chr.21: 25603760-25603760 on Build GRCh38
rs725493 Chr.21: 25603683-25603683 on Build GRCh38
rs768395646 Chr.21: 25603761-25603761 on Build GRCh38
rs375688137 Chr.21: 25603775-25603775 on Build GRCh38
rs778913318 Chr.21: 25603851-25603852 on Build GRCh38
rs115932466 Chr.21: 25603837-25603837 on Build GRCh38
rs149452993 Chr.21: 25603836-25603836 on Build GRCh38
rs369538658 Chr.21: 25603736-25603736 on Build GRCh38
rs574362792 Chr.21: 25603635-25603635 on Build GRCh38
rs776628965 Chr.21: 25603768-25603768 on Build GRCh38
rs572029241 Chr.21: 25603823-25603823 on Build GRCh38
rs116331755 Chr.21: 25603832-25603832 on Build GRCh38
rs764430887 Chr.21: 25603844-25603844 on Build GRCh38
rs538753515 Chr.21: 25603698-25603698 on Build GRCh38
rs1027163849 Chr.21: 25603674-25603674 on Build GRCh38
rs769327598 Chr.21: 25603787-25603787 on Build GRCh38
rs923573656 Chr.21: 25603750-25603750 on Build GRCh38
rs541469036 Chr.21: 25603649-25603649 on Build GRCh38
rs748195777 Chr.21: 25603726-25603726 on Build GRCh38
rs73899034 Chr.21: 25603645-25603645 on Build GRCh38
rs377561797 Chr.21: 25603803-25603803 on Build GRCh38
rs762658061 Chr.21: 25603810-25603810 on Build GRCh38
rs1002861295 Chr.21: 25603629-25603629 on Build GRCh38
rs535901780 Chr.21: 25603657-25603657 on Build GRCh38
rs373523504 Chr.21: 25603780-25603780 on Build GRCh38

View Full Product Details