Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28535009-28535264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCC1
Entrez Gene ID
1104
Gene Name
regulator of chromosome condensation 1
Gene Aliases
CHC1, RCC1-I, SNHG3-RCC1
UniGene
Hs.469723
Gene Chromosome Location
Chr.1: 28505943-28539196 on Build GRCh38
Transcripts
NM_001048194.2 NM_001048195.2 NM_001048199.2 NM_001269.4 NR_030725.1 NR_030726.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751133 c.609G>A p.V203V Chr.1: 28535124-28535124 on Build GRCh38
464397 c.465G>A p.L155L Chr.1: 28535073-28535073 on Build GRCh38
1223443 c.538G>T p.G180* Chr.1: 28535146-28535146 on Build GRCh38
1195403 c.544G>A p.G182S Chr.1: 28535059-28535059 on Build GRCh38
1667320 c.493C>T p.P165S Chr.1: 28535101-28535101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767438460 Chr.1: 28535192-28535192 on Build GRCh38
rs909069779 Chr.1: 28535054-28535054 on Build GRCh38
rs772602606 Chr.1: 28535128-28535128 on Build GRCh38
rs375601359 Chr.1: 28535116-28535116 on Build GRCh38
rs750499626 Chr.1: 28535176-28535176 on Build GRCh38
rs11584475 Chr.1: 28535208-28535208 on Build GRCh38
rs751604308 Chr.1: 28535042-28535042 on Build GRCh38
rs764224573 Chr.1: 28535140-28535140 on Build GRCh38
rs757675613 Chr.1: 28535152-28535152 on Build GRCh38
rs769356283 Chr.1: 28535091-28535091 on Build GRCh38
rs148399552 Chr.1: 28535103-28535103 on Build GRCh38
rs780886920 Chr.1: 28535059-28535059 on Build GRCh38
rs36087670 Chr.1: 28535199-28535200 on Build GRCh38
rs764324439 Chr.1: 28535029-28535029 on Build GRCh38
rs891359176 Chr.1: 28535217-28535217 on Build GRCh38
rs777350942 Chr.1: 28535097-28535097 on Build GRCh38
rs779128221 Chr.1: 28535040-28535040 on Build GRCh38
rs773841273 Chr.1: 28535122-28535122 on Build GRCh38
rs754902191 Chr.1: 28535181-28535181 on Build GRCh38
rs373741597 Chr.1: 28535058-28535058 on Build GRCh38
rs2066726 Chr.1: 28535124-28535124 on Build GRCh38
rs757334897 Chr.1: 28535038-28535038 on Build GRCh38
rs765455765 Chr.1: 28535169-28535169 on Build GRCh38
rs200976400 Chr.1: 28535226-28535226 on Build GRCh38
rs1022676094 Chr.1: 28535074-28535074 on Build GRCh38
rs747933048 Chr.1: 28535062-28535062 on Build GRCh38
rs1000576421 Chr.1: 28535234-28535234 on Build GRCh38
rs775752504 Chr.1: 28535131-28535131 on Build GRCh38
rs778437799 Chr.1: 28535233-28535233 on Build GRCh38
rs558634776 Chr.1: 28535044-28535044 on Build GRCh38
rs761040336 Chr.1: 28535139-28535139 on Build GRCh38
rs756885070 Chr.1: 28535228-28535228 on Build GRCh38
rs755872535 Chr.1: 28535220-28535220 on Build GRCh38
rs371074587 Chr.1: 28535223-28535223 on Build GRCh38
rs141534722 Chr.1: 28535073-28535073 on Build GRCh38
rs753978304 Chr.1: 28535037-28535037 on Build GRCh38
rs201559412 Chr.1: 28535154-28535154 on Build GRCh38
rs201498588 Chr.1: 28535233-28535234 on Build GRCh38
rs1010262467 Chr.1: 28535221-28535221 on Build GRCh38

View Full Product Details