Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 102867202-102867475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAM
Entrez Gene ID
5066
Gene Name
peptidylglycine alpha-amidating monooxygenase
Gene Aliases
PAL, PHM
UniGene
Hs.369430
Gene Chromosome Location
Chr.5: 102754783-103031105 on Build GRCh38
Transcripts
NM_000919.3 NM_001177306.1 NM_001319943.1 NM_138766.2 NM_138821.2 NM_138822.2 NR_033440.1 XM_011543419.2 XM_017009495.1 XM_017009496.1 XM_017009497.1 XM_017009498.1 XM_017009499.1 XM_017009500.1 XM_017009501.1 XM_017009502.1 XM_017009503.1 XM_017009504.1 XM_017009505.1 XM_017009506.1 XM_017009507.1 XM_017009508.1 XM_017009509.1 XM_017009510.1 XM_017009511.1 XM_017009512.1 XM_017009513.1 XM_017009514.1 XM_017009515.1 XM_017009516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1059247 c.173C>T p.A58V Chr.5: 102867356-102867356 on Build GRCh38
5750956 c.110C>A p.P37Q Chr.5: 102867293-102867293 on Build GRCh38
2989735 c.145G>A p.V49I Chr.5: 102867328-102867328 on Build GRCh38
6737146 c.90-9delT p.? Chr.5: 102867264-102867264 on Build GRCh38
4920142 c.124T>A p.C42S Chr.5: 102867307-102867307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055319357 Chr.5: 102867292-102867292 on Build GRCh38
rs114014768 Chr.5: 102867275-102867275 on Build GRCh38
rs776407655 Chr.5: 102867367-102867367 on Build GRCh38
rs760548024 Chr.5: 102867266-102867266 on Build GRCh38
rs757392901 Chr.5: 102867386-102867386 on Build GRCh38
rs766256728 Chr.5: 102867358-102867358 on Build GRCh38
rs890638433 Chr.5: 102867265-102867265 on Build GRCh38
rs761335947 Chr.5: 102867232-102867232 on Build GRCh38
rs1005988460 Chr.5: 102867426-102867426 on Build GRCh38
rs751643681 Chr.5: 102867316-102867316 on Build GRCh38
rs780352855 Chr.5: 102867407-102867407 on Build GRCh38
rs941671119 Chr.5: 102867427-102867427 on Build GRCh38
rs201199380 Chr.5: 102867355-102867355 on Build GRCh38
rs772566153 Chr.5: 102867332-102867332 on Build GRCh38
rs994652163 Chr.5: 102867394-102867394 on Build GRCh38
rs750713150 Chr.5: 102867392-102867392 on Build GRCh38
rs371258308 Chr.5: 102867327-102867327 on Build GRCh38
rs774941196 Chr.5: 102867338-102867338 on Build GRCh38
rs749679855 Chr.5: 102867416-102867416 on Build GRCh38
rs780957345 Chr.5: 102867383-102867385 on Build GRCh38
rs764225314 Chr.5: 102867286-102867286 on Build GRCh38
rs756282251 Chr.5: 102867405-102867405 on Build GRCh38
rs1037402543 Chr.5: 102867441-102867441 on Build GRCh38
rs200231533 Chr.5: 102867336-102867336 on Build GRCh38
rs750280392 Chr.5: 102867411-102867411 on Build GRCh38
rs903605923 Chr.5: 102867256-102867256 on Build GRCh38
rs2230458 Chr.5: 102867328-102867328 on Build GRCh38
rs368627926 Chr.5: 102867442-102867442 on Build GRCh38
rs537531279 Chr.5: 102867377-102867377 on Build GRCh38
rs200926087 Chr.5: 102867277-102867277 on Build GRCh38
rs767467212 Chr.5: 102867390-102867390 on Build GRCh38
rs368145272 Chr.5: 102867287-102867287 on Build GRCh38
rs779593570 Chr.5: 102867432-102867432 on Build GRCh38
rs767211611 Chr.5: 102867242-102867242 on Build GRCh38
rs200066052 Chr.5: 102867343-102867343 on Build GRCh38
rs979885641 Chr.5: 102867340-102867340 on Build GRCh38
rs539783478 Chr.5: 102867290-102867290 on Build GRCh38
rs760224132 Chr.5: 102867357-102867357 on Build GRCh38
rs907784206 Chr.5: 102867403-102867403 on Build GRCh38
rs757230186 Chr.5: 102867324-102867324 on Build GRCh38
rs367562349 Chr.5: 102867435-102867435 on Build GRCh38
rs756027307 Chr.5: 102867415-102867415 on Build GRCh38
rs754897780 Chr.5: 102867282-102867282 on Build GRCh38
rs749994149 Chr.5: 102867263-102867263 on Build GRCh38
rs145666758 Chr.5: 102867378-102867378 on Build GRCh38
rs769287452 Chr.5: 102867337-102867337 on Build GRCh38
rs755345474 Chr.5: 102867418-102867418 on Build GRCh38
rs759259940 Chr.5: 102867370-102867370 on Build GRCh38
rs140503241 Chr.5: 102867334-102867334 on Build GRCh38
rs1027037798 Chr.5: 102867413-102867413 on Build GRCh38
rs766055359 Chr.5: 102867270-102867270 on Build GRCh38

View Full Product Details