Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45890238-45890497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSA
Entrez Gene ID
5476
Gene Name
cathepsin A
Gene Aliases
GLB2, GSL, NGBE, PPCA, PPGB
UniGene
Hs.609336
Gene Chromosome Location
Chr.20: 45890144-45898820 on Build GRCh38
Transcripts
NM_000308.3 NM_001127695.2 NM_001167594.2 NR_133656.1 NM_001278535.1 NM_080749.3

Gene Symbol
NEURL2
Entrez Gene ID
140825
Gene Name
neuralized E3 ubiquitin protein ligase 2
Gene Aliases
C20orf163, OZZ, OZZ-E3
UniGene
Hs.517094
Gene Chromosome Location
Chr.20: 45888472-45891287 on Build GRCh38
Transcripts
NM_000308.3 NM_001127695.2 NM_001167594.2 NR_133656.1 NM_001278535.1 NM_080749.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6724795 c.577C>T p.L193F Chr.20: 45890415-45890415 on Build GRCh38
1286576 c.719G>T p.R240L Chr.20: 45890273-45890273 on Build GRCh38
3841147 c.668C>T p.A223V Chr.20: 45890324-45890324 on Build GRCh38
1196868 c.682G>T p.A228S Chr.20: 45890310-45890310 on Build GRCh38
6724790 c.603G>T p.T201T Chr.20: 45890389-45890389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780412328 Chr.20: 45890438-45890438 on Build GRCh38
rs577241918 Chr.20: 45890354-45890354 on Build GRCh38
rs759545007 Chr.20: 45890379-45890379 on Build GRCh38
rs760597081 Chr.20: 45890464-45890464 on Build GRCh38
rs757328038 Chr.20: 45890320-45890320 on Build GRCh38
rs772110184 Chr.20: 45890463-45890463 on Build GRCh38
rs771797053 Chr.20: 45890319-45890320 on Build GRCh38
rs778987917 Chr.20: 45890414-45890414 on Build GRCh38
rs775660814 Chr.20: 45890263-45890263 on Build GRCh38
rs747935206 Chr.20: 45890348-45890348 on Build GRCh38
rs753980880 Chr.20: 45890318-45890318 on Build GRCh38
rs530672133 Chr.20: 45890451-45890451 on Build GRCh38
rs371582207 Chr.20: 45890440-45890440 on Build GRCh38
rs747472921 Chr.20: 45890439-45890439 on Build GRCh38
rs750431114 Chr.20: 45890422-45890422 on Build GRCh38
rs376732514 Chr.20: 45890403-45890403 on Build GRCh38
rs767364546 Chr.20: 45890303-45890303 on Build GRCh38
rs763893268 Chr.20: 45890475-45890475 on Build GRCh38
rs752391348 Chr.20: 45890308-45890308 on Build GRCh38
rs766456407 Chr.20: 45890367-45890369 on Build GRCh38
rs1045626288 Chr.20: 45890395-45890395 on Build GRCh38
rs940200985 Chr.20: 45890305-45890305 on Build GRCh38
rs909960503 Chr.20: 45890304-45890304 on Build GRCh38
rs772411777 Chr.20: 45890332-45890332 on Build GRCh38
rs1052969252 Chr.20: 45890367-45890367 on Build GRCh38
rs746401969 Chr.20: 45890327-45890327 on Build GRCh38
rs773090786 Chr.20: 45890326-45890327 on Build GRCh38
rs747304562 Chr.20: 45890344-45890344 on Build GRCh38
rs748968671 Chr.20: 45890443-45890443 on Build GRCh38
rs762537945 Chr.20: 45890298-45890298 on Build GRCh38
rs752590233 Chr.20: 45890394-45890394 on Build GRCh38
rs1034237882 Chr.20: 45890421-45890421 on Build GRCh38
rs761549706 Chr.20: 45890266-45890266 on Build GRCh38
rs760521678 Chr.20: 45890339-45890346 on Build GRCh38
rs764757463 Chr.20: 45890278-45890278 on Build GRCh38
rs115355487 Chr.20: 45890314-45890314 on Build GRCh38
rs770575809 Chr.20: 45890360-45890360 on Build GRCh38
rs775593066 Chr.20: 45890315-45890316 on Build GRCh38
rs758984311 Chr.20: 45890437-45890437 on Build GRCh38
rs780499100 Chr.20: 45890338-45890338 on Build GRCh38
rs778900027 Chr.20: 45890323-45890323 on Build GRCh38
rs892577789 Chr.20: 45890377-45890377 on Build GRCh38
rs1001455876 Chr.20: 45890406-45890406 on Build GRCh38
rs766490144 Chr.20: 45890299-45890299 on Build GRCh38
rs932273399 Chr.20: 45890326-45890326 on Build GRCh38
rs142373830 Chr.20: 45890381-45890381 on Build GRCh38
rs573188133 Chr.20: 45890405-45890405 on Build GRCh38
rs367770819 Chr.20: 45890311-45890311 on Build GRCh38
rs1014990420 Chr.20: 45890465-45890465 on Build GRCh38
rs772898181 Chr.20: 45890290-45890290 on Build GRCh38
rs35342327 Chr.20: 45890361-45890361 on Build GRCh38
rs757414186 Chr.20: 45890413-45890413 on Build GRCh38
rs754812339 Chr.20: 45890301-45890301 on Build GRCh38
rs986305470 Chr.20: 45890336-45890336 on Build GRCh38
rs770663921 Chr.20: 45890449-45890449 on Build GRCh38
rs754067203 Chr.20: 45890408-45890408 on Build GRCh38
rs763171912 Chr.20: 45890317-45890318 on Build GRCh38
rs1005907488 Chr.20: 45890456-45890456 on Build GRCh38
rs753818477 Chr.20: 45890466-45890477 on Build GRCh38
rs758943658 Chr.20: 45890452-45890452 on Build GRCh38
rs762625310 Chr.20: 45890355-45890355 on Build GRCh38

View Full Product Details