Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55954839-55955086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP8
Entrez Gene ID
126205
Gene Name
NLR family pyrin domain containing 8
Gene Aliases
CLR19.2, NALP8, NOD16, PAN4
UniGene
Hs.446925
Gene Chromosome Location
Chr.19: 55947832-55988629 on Build GRCh38
Transcripts
NM_001317000.1 NM_176811.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563025 c.953G>A p.S318N Chr.19: 55955011-55955011 on Build GRCh38
4563336 c.969G>A p.T323T Chr.19: 55955027-55955027 on Build GRCh38
6477978 c.791G>A p.W264* Chr.19: 55954849-55954849 on Build GRCh38
349668 c.802C>T p.Q268* Chr.19: 55954860-55954860 on Build GRCh38
6727265 c.973C>A p.L325I Chr.19: 55955031-55955031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774857045 Chr.19: 55954961-55954961 on Build GRCh38
rs753842717 Chr.19: 55955027-55955027 on Build GRCh38
rs763873285 Chr.19: 55954888-55954888 on Build GRCh38
rs199475836 Chr.19: 55954908-55954908 on Build GRCh38
rs562393097 Chr.19: 55954891-55954891 on Build GRCh38
rs752969286 Chr.19: 55955045-55955045 on Build GRCh38
rs750845471 Chr.19: 55954996-55954996 on Build GRCh38
rs544368279 Chr.19: 55954893-55954893 on Build GRCh38
rs927432618 Chr.19: 55955028-55955028 on Build GRCh38
rs143237796 Chr.19: 55954980-55954980 on Build GRCh38
rs951858087 Chr.19: 55954944-55954944 on Build GRCh38
rs773891559 Chr.19: 55954909-55954909 on Build GRCh38
rs145359925 Chr.19: 55955026-55955026 on Build GRCh38
rs770878136 Chr.19: 55954933-55954933 on Build GRCh38
rs950339354 Chr.19: 55955023-55955023 on Build GRCh38
rs370475583 Chr.19: 55954917-55954917 on Build GRCh38
rs1010163656 Chr.19: 55954862-55954862 on Build GRCh38
rs117657068 Chr.19: 55954868-55954868 on Build GRCh38
rs775150687 Chr.19: 55954878-55954878 on Build GRCh38
rs929649077 Chr.19: 55955058-55955058 on Build GRCh38
rs761086105 Chr.19: 55955013-55955013 on Build GRCh38
rs746748995 Chr.19: 55954925-55954925 on Build GRCh38
rs745809585 Chr.19: 55954945-55954945 on Build GRCh38
rs762189376 Chr.19: 55954892-55954892 on Build GRCh38
rs1050994020 Chr.19: 55955065-55955065 on Build GRCh38
rs781205586 Chr.19: 55954934-55954934 on Build GRCh38
rs776731099 Chr.19: 55954866-55954866 on Build GRCh38
rs779989262 Chr.19: 55954902-55954902 on Build GRCh38
rs7259764 Chr.19: 55954861-55954861 on Build GRCh38
rs768169112 Chr.19: 55954975-55954975 on Build GRCh38
rs755110708 Chr.19: 55955035-55955035 on Build GRCh38
rs927043465 Chr.19: 55954976-55954976 on Build GRCh38
rs759451076 Chr.19: 55954958-55954958 on Build GRCh38
rs561042891 Chr.19: 55954905-55954905 on Build GRCh38
rs575075134 Chr.19: 55954883-55954883 on Build GRCh38
rs907603497 Chr.19: 55954973-55954973 on Build GRCh38
rs767485403 Chr.19: 55955061-55955061 on Build GRCh38
rs199538298 Chr.19: 55954989-55954989 on Build GRCh38
rs767883117 Chr.19: 55954991-55954991 on Build GRCh38
rs762099494 Chr.19: 55954988-55954988 on Build GRCh38
rs779234643 Chr.19: 55955044-55955044 on Build GRCh38
rs140931794 Chr.19: 55954869-55954869 on Build GRCh38
rs750935000 Chr.19: 55954895-55954895 on Build GRCh38
rs200445239 Chr.19: 55954969-55954969 on Build GRCh38
rs757057195 Chr.19: 55955055-55955055 on Build GRCh38
rs1038092405 Chr.19: 55955006-55955006 on Build GRCh38
rs373207228 Chr.19: 55954918-55954918 on Build GRCh38

View Full Product Details