Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50885187-50885444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS12
Entrez Gene ID
283471
Gene Name
transmembrane protease, serine 12
Gene Aliases
CT151
UniGene
Hs.125571
Gene Chromosome Location
Chr.12: 50842918-50887884 on Build GRCh38
Transcripts
NM_182559.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072780 c.764G>A p.G255D Chr.12: 50885357-50885357 on Build GRCh38
4898279 c.743C>T p.P248L Chr.12: 50885336-50885336 on Build GRCh38
940404 c.728A>G p.Y243C Chr.12: 50885321-50885321 on Build GRCh38
940403 c.705G>T p.E235D Chr.12: 50885298-50885298 on Build GRCh38
1299499 c.657C>T p.N219N Chr.12: 50885250-50885250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754129759 Chr.12: 50885420-50885421 on Build GRCh38
rs564001932 Chr.12: 50885294-50885294 on Build GRCh38
rs374684282 Chr.12: 50885251-50885251 on Build GRCh38
rs756252050 Chr.12: 50885324-50885324 on Build GRCh38
rs367743059 Chr.12: 50885293-50885293 on Build GRCh38
rs764701325 Chr.12: 50885399-50885400 on Build GRCh38
rs747785172 Chr.12: 50885387-50885387 on Build GRCh38
rs767827960 Chr.12: 50885322-50885322 on Build GRCh38
rs770627049 Chr.12: 50885406-50885406 on Build GRCh38
rs779172967 Chr.12: 50885354-50885354 on Build GRCh38
rs746570828 Chr.12: 50885280-50885280 on Build GRCh38
rs372309874 Chr.12: 50885250-50885250 on Build GRCh38
rs865827502 Chr.12: 50885372-50885372 on Build GRCh38
rs201837185 Chr.12: 50885326-50885326 on Build GRCh38
rs770964354 Chr.12: 50885282-50885282 on Build GRCh38
rs749648769 Chr.12: 50885232-50885232 on Build GRCh38
rs948114716 Chr.12: 50885309-50885309 on Build GRCh38
rs761291002 Chr.12: 50885391-50885393 on Build GRCh38
rs774568077 Chr.12: 50885298-50885298 on Build GRCh38
rs976595141 Chr.12: 50885299-50885300 on Build GRCh38
rs1043842270 Chr.12: 50885360-50885360 on Build GRCh38
rs746517747 Chr.12: 50885401-50885401 on Build GRCh38
rs754351643 Chr.12: 50885327-50885327 on Build GRCh38
rs751220041 Chr.12: 50885421-50885422 on Build GRCh38
rs201478278 Chr.12: 50885285-50885285 on Build GRCh38
rs757967515 Chr.12: 50885393-50885393 on Build GRCh38
rs753202427 Chr.12: 50885389-50885390 on Build GRCh38
rs767715782 Chr.12: 50885231-50885231 on Build GRCh38
rs755299136 Chr.12: 50885345-50885345 on Build GRCh38
rs182342296 Chr.12: 50885304-50885304 on Build GRCh38
rs903672917 Chr.12: 50885398-50885398 on Build GRCh38
rs755389442 Chr.12: 50885238-50885238 on Build GRCh38
rs777612545 Chr.12: 50885395-50885395 on Build GRCh38
rs556211153 Chr.12: 50885239-50885239 on Build GRCh38

View Full Product Details