Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68980147-68980394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGPRD
Entrez Gene ID
116512
Gene Name
MAS related GPR family member D
Gene Aliases
MRGD, TGR7
UniGene
Hs.527802
Gene Chromosome Location
Chr.11: 68980021-68980986 on Build GRCh38
Transcripts
NM_198923.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4036192 c.716T>C p.I239T Chr.11: 68980271-68980271 on Build GRCh38
6715042 c.640C>T p.R214W Chr.11: 68980347-68980347 on Build GRCh38
6715040 c.652C>T p.R218W Chr.11: 68980335-68980335 on Build GRCh38
3953737 c.652C>A p.R218R Chr.11: 68980335-68980335 on Build GRCh38
5604114 c.609C>T p.V203V Chr.11: 68980378-68980378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767675310 Chr.11: 68980308-68980308 on Build GRCh38
rs137989951 Chr.11: 68980269-68980269 on Build GRCh38
rs576529805 Chr.11: 68980334-68980334 on Build GRCh38
rs769003671 Chr.11: 68980302-68980303 on Build GRCh38
rs369321530 Chr.11: 68980335-68980335 on Build GRCh38
rs199619059 Chr.11: 68980285-68980285 on Build GRCh38
rs745676884 Chr.11: 68980370-68980370 on Build GRCh38
rs965102744 Chr.11: 68980186-68980186 on Build GRCh38
rs1057168016 Chr.11: 68980332-68980333 on Build GRCh38
rs375088455 Chr.11: 68980283-68980283 on Build GRCh38
rs370340360 Chr.11: 68980203-68980203 on Build GRCh38
rs756255622 Chr.11: 68980228-68980228 on Build GRCh38
rs150304482 Chr.11: 68980227-68980227 on Build GRCh38
rs868172152 Chr.11: 68980190-68980190 on Build GRCh38
rs766196399 Chr.11: 68980268-68980268 on Build GRCh38
rs760409878 Chr.11: 68980176-68980176 on Build GRCh38
rs368476984 Chr.11: 68980168-68980168 on Build GRCh38
rs753166976 Chr.11: 68980346-68980346 on Build GRCh38
rs780931228 Chr.11: 68980357-68980357 on Build GRCh38
rs146025023 Chr.11: 68980371-68980371 on Build GRCh38
rs966598823 Chr.11: 68980244-68980244 on Build GRCh38
rs759335972 Chr.11: 68980274-68980274 on Build GRCh38
rs900414568 Chr.11: 68980167-68980167 on Build GRCh38
rs1031028574 Chr.11: 68980202-68980202 on Build GRCh38
rs545567692 Chr.11: 68980294-68980294 on Build GRCh38
rs921561364 Chr.11: 68980359-68980359 on Build GRCh38
rs772771135 Chr.11: 68980238-68980238 on Build GRCh38
rs771566564 Chr.11: 68980237-68980237 on Build GRCh38
rs369796412 Chr.11: 68980208-68980208 on Build GRCh38
rs371981858 Chr.11: 68980246-68980246 on Build GRCh38
rs765259755 Chr.11: 68980189-68980189 on Build GRCh38
rs770470283 Chr.11: 68980262-68980262 on Build GRCh38
rs776537616 Chr.11: 68980188-68980188 on Build GRCh38
rs750447253 Chr.11: 68980326-68980326 on Build GRCh38
rs781495882 Chr.11: 68980233-68980233 on Build GRCh38
rs989403988 Chr.11: 68980195-68980195 on Build GRCh38
rs746490723 Chr.11: 68980349-68980349 on Build GRCh38
rs999333600 Chr.11: 68980192-68980192 on Build GRCh38
rs755548426 Chr.11: 68980234-68980234 on Build GRCh38
rs766507797 Chr.11: 68980332-68980332 on Build GRCh38
rs544096530 Chr.11: 68980350-68980350 on Build GRCh38
rs201081941 Chr.11: 68980177-68980177 on Build GRCh38
rs928254912 Chr.11: 68980279-68980279 on Build GRCh38
rs200393238 Chr.11: 68980229-68980229 on Build GRCh38
rs772867905 Chr.11: 68980174-68980174 on Build GRCh38
rs768822138 Chr.11: 68980224-68980224 on Build GRCh38
rs115772206 Chr.11: 68980330-68980330 on Build GRCh38
rs779419192 Chr.11: 68980343-68980343 on Build GRCh38
rs535121405 Chr.11: 68980261-68980261 on Build GRCh38
rs777395337 Chr.11: 68980347-68980347 on Build GRCh38

View Full Product Details